Search
| Cas9 format | Transfection reagent* | Electroporation** | |
|---|---|---|---|
| TrueCut Cas9 Protein v2 + gRNA | Lipofectamine CRISPRMAX Cas9 Transfection Reagent | For maximum efficiency in difficult-to-transfect cell types, use the Neon Transfection System | |
| * For best results, we recommend that you transfect your cells with TrueCut Cas9 Protein v2 + TrueGuide Synthetic gRNA (crRNA:tracrRNA duplex or sgRNA) using the Lipofectamine CRISPRMAX Cas9 Transfection Reagent. ** Use the Neon Transfection System 10 μL Kit (Cat. No. MPK1025). | |||
For Research Use Only. Not for use in diagnostic procedures.