Versatility meets functionality with multiparameter meters
Measure pH, ion concentration, conductivity, dissolved oxygen, and temperature with a single, powerful meter. Our benchtop and portable multiparameter meters are designed to increase your productivity in the lab and in the field. Choosing a multiparameter meter reduces the amount of equipment on your benchtop or in your hands while in the field, eliminates the need for multiple meters as the display showcases multiple parameters on one unit, and saves time in transferring electrodes from meter to meter for measurements. Combine your parameters today for simultaneous measurements saving space, time, and money. Browse our complete portfolio or let one of our experts help you make a selection.
Select your multiparameter bench or portable meter
Premium
Advanced
 Orion Versa
Star Pro pH/Ion, Conductivity & DO Bench Meter
(Versa Star Pro series)
Orion Versa
Star Pro pH/Conductivity
Bench Meter
(Versa Star Pro series)
Orion Pro Star
MP218 pH/Ion, Conductivity & DO Bench Meter
(Pro Star series)
Orion Pro Star PD216 pH/Ion &
DO Bench Meter
(Pro Star series)
Orion Pro Star PC215 pH/Ion & Conductivity
Bench Meter
(Pro Star series)
 Multiparameter
high-performance
laboratory meters
Dual parameter
high-performance
laboratory meters
Multiparameter easy to use, dependable laboratory metersMultiparameter easy to use, dependable laboratory metersMultiparameter easy to use, dependable laboratory meters
Ideal applicationsFood & beverage, industrial, waterPharmaceutical, biotechnology, food & beverageEnvironmental,
water/ wastewater,
food & beverage, industrial
Environmental,
water/ wastewater, food & beverage, industrial
Environmental,
water/ wastewater, food & beverage, industrial
pH range-2.000 to 20.000-2.000 to 20.000-2.000 to 20.000-2.000 to 20.000-2.000 to 20.000
pH resolution0.001 / 0.01 / 0.10.001 / 0.01 / 0.10.001 / 0.01 / 0.10.001 / 0.01 / 0.10.001 / 0.01 / 0.1
pH calibration pointsUp to 6Up to 6Up to 5Up to 5Up to 5
mV, RmV, ORP modesYesYesYesYesYes
Ion concentration modeYesNoYesYesYes
Conductivity modeYesYesYesNoYes
Additional modesTDS, salinity,
resistivity
TDS, salinity,
resistivity
TDS, salinity,
resistivity
—TDS, salinity, resistivity
Dissolved oxygen modeYesNoYesYesNo
Compatible probesOptical,
polarographic
—Optical,
polarographic
Optical,
polarographic
—
Temperature range-5 to 105°C
23 to 221°F
-5 to 105°C
23 to 221°F
-30 to 130ºC
-22 to 266ºF
-30 to 130ºC
-22 to 266ºF
-30 to 130ºC
-22 to 266ºF
Number of channels 4, customizable
with modules
 4, customizable
with modules
322
Stirrer probe inputs22222
Display5.3-inch, backlit
color display
5.3-inch, backlit
color display
7-inch, high-
resolution, backlit
color display
7-inch, high-resolution, backlit color display7-inch, high-resolution, backlit color display
TouchscreenNoNoYesYesYes
Onscreen text promptsStep-by-step
calibration and
guided setup
instructions
Step-by-step
calibration and
guided setup
instructions
Step-by-step
calibration and
guided setup
instructions
Step-by-step calibration and
guided setup instructions
Step-by-step calibration and
guided setup instructions
Data log2000 with date/time2000 with date/time10000 with date/time10000 with date/time10000 with date/time
Calibration log30 per parameter30 per parameter20 per parameter20 per parameter20 per parameter
Identification optionsElectrode ID,
sample ID, user ID
Electrode ID,
sample ID, user ID
Electrode ID,
sample ID, user ID
Electrode ID,
sample ID, user ID
Electrode ID,
sample ID, user ID
Data export optionsUSB (2), RS232USB (2), RS232Computer, printer,
USB flash drive
Computer, printer,
USB flash drive
Computer, printer,
USB flash drive
Power supplyUniversal,
100–240 VAC,
50–60 Hz
(Cat. No. STARA-PWR)
Universal,
100–240 VAC,
50–60 Hz
(Cat. No. STARA-PWR)
Universal,
100-240 VAC,
50-60 Hz
(Cat. No. PSTAR-PWR),
9 DC adapter, 2.0A
Universal,
100-240 VAC,
50-60 Hz,
(Cat. No. PSTAR-PWR),
9 DC adapter, 2.0A
Universal,
100-240 VAC,
50-60 Hz
(Cat. No. PSTAR-PWR),
9 DC adapter, 2.0A 
Premium
Premium
Premium
 Orion Star A325
Multiparameter meter
(STARA3250 series)
Orion Star A326
Multiparameter meter
(STARA3260 series)
Orion Star A329
Multiparameter meter
(STARA3290 series)
 Premium STARA3250 series Orion multiparameter metersPremium STARA3260 series Orion multiparameter metersPremium STARA3290 series Orion multiparameter meters
 Rugged, dependable multiparameter meterRugged, versatile, and dependable multiparameter meterVersatile, powerful, multiparameter meter
Ideal for
applications in:
Industrial—cooling towers, wastewater; Environmental—ponds, lakesIndustrial—wastewater; Environmental—ponds, lakesIndustrial—wastewater, drinking water, food & beverage; Environmental—ponds, lakes
pH range–2.000 to 20.000–2.000 to 20.000–2.000 to 20.000
pH resolution0.1/0.01/0.0010.1/0.01/0.0010.1/0.01/0.001
pH relative accuracy±0.002±0.002±0.002
pH calibration pointsUp to 5Up to 5Up to 5
mV, RmV range±2,000.0 mV±2,000.0 mV±2,000.0 mV
EH/ORP modeYesYesYes
Dissolved oxygen modeNoYesYes
TDS relative accuracy±0.5% reading ±1 digit--±0.5% reading ±1 digit
Temperature range–5 to 105°C, 23 to 221°F–5 to 105ºC, 23 to 221ºF–5 to 105ºC, 23 to 221ºF
Number of datalogs5000, with time and date stamp5000, with time and date stamp5000, with time and date stamp
Data export optionsRS232, USBRS232, USBRS232, USB
AC adapterOptional—universal, 100–240VACOptional—universal, 100–240VACOptional—universal, 100–240VAC
Battery powerIncluded—4AAsIncluded—4AAsIncluded—4AAs
  Enhance your productivity with accessories for your benchtop and portable meters. Meter Accessories ›
Advanced
 Eutech PC2700 pH/Conductivity Bench Meter
 Economical, routine laboratory meters
pH range-2.000 to 20.000
pH resolution0.001 / 0.01 / 0.1
pH calibration pointsUp to 6
mV, RmV modesYes
Ion concentration modeYes
Conductivity modeYes
Additional modesTDS, salinity, resistivity
Range (temperature)0.0 to 100.0 °C, 32.0 to 212.0 °F
Backlit displayYes
Number of channels2
Data log500 with date/time
Calibration logMost recent viewable on meter and stored in meter memory
Data export optionsRS232
Power supplyUniversal, 100-240VAC, 50/60 Hz, Cat. No. 60X426401
Premium
Premium
Advanced
Advanced
Advanced
Advanced
 Eutech PD 450 Portable Multiparameter MeterEutech PC 450 Portable Multiparameter MeterEutech PCD 650 Portable Multiparameter MeterEutech PC 650 Portable Multiparameter MeterEutech PD 650 Portable Multiparameter MeterEutech CD 650 Portable Multiparameter Meter
pH range-2.00 to 16.00-2.00 to 16.00-2.00 to 20.00-2.00 to 20.00-2.00 to 20.00---
pH resolution0.01 pH0.01 pH0.1 / 0.01 Ph0.1 / 0.01 Ph0.1 / 0.01 Ph---
pH relative accuracy±0.01 pH±0.01 pH±0.01 pH±0.01 pH±0.01 pH---
pH calibration pointsUp to 6Up to 6Up to 6Up to 6Up to 6---
mV, RmV modesYesYesYesYesYes---
EH/ORP mode------YesYesYes---
Conductivity mode---YesYesYes---Yes
Dissolved oxygen modeYes---Yes---YesYes
TDS relative accuracy---±1 % full scale±1 % full scale±1 % full scale±1 % full scale±1 % full scale +
Ion concentration mode------YesYesYes---
Temperature range-10.0 to 110.0 °C (14.0 to 230.0 °F)-10.0 to 110.0 °C (14.0 to 230.0 °F)-10.0 to 110.0 °C (14.0 to 230.0 °F)-10.0 to 110.0 °C (14.0 to 230.0 °F)-10.0 to 110.0 °C (14.0 to 230.0 °F)-10.0 to 110.0 °C (14.0 to 230.0 °F)
Number of datalogs500 with date and time stamp500 with date and time stamp500 with date and time stamp500 with date and time stamp500 with date and time stamp500 with date and time stamp
Data export optionsUSB or RS232USB or RS232RS232C through LED, IrDARS232C through LED, IrDARS232C through LED, IrDARS232C through LED, IrDA
AC adapter9 V DC adapter (optional)9 V DC adapter (optional)9 V DC adapter9 V DC adapter9 V DC adapter9 V DC adapter
Battery power2 AA batteries (included)2 AA batteries (included)4 AA batteries (included)4 AA batteries (included)4 AA batteries (included)4 AA batteries (included)
Premium
Advanced
 Orion Versa
Star Pro pH/Ion, Conductivity & DO Bench Meter
(Versa Star Pro series)
Orion Versa
Star Pro pH/Conductivity
Bench Meter
(Versa Star Pro series)
Orion Pro Star
MP218 pH/Ion, Conductivity & DO Bench Meter
(Pro Star series)
Orion Pro Star PD216 pH/Ion &
DO Bench Meter
(Pro Star series)
Orion Pro Star PC215 pH/Ion & Conductivity
Bench Meter
(Pro Star series)
 Multiparameter
high-performance
laboratory meters
Dual parameter
high-performance
laboratory meters
Multiparameter easy to use, dependable laboratory metersMultiparameter easy to use, dependable laboratory metersMultiparameter easy to use, dependable laboratory meters
Ideal applicationsFood & beverage, industrial, waterPharmaceutical, biotechnology, food & beverageEnvironmental,
water/ wastewater,
food & beverage, industrial
Environmental,
water/ wastewater, food & beverage, industrial
Environmental,
water/ wastewater, food & beverage, industrial
pH range-2.000 to 20.000-2.000 to 20.000-2.000 to 20.000-2.000 to 20.000-2.000 to 20.000
pH resolution0.001 / 0.01 / 0.10.001 / 0.01 / 0.10.001 / 0.01 / 0.10.001 / 0.01 / 0.10.001 / 0.01 / 0.1
pH calibration pointsUp to 6Up to 6Up to 5Up to 5Up to 5
mV, RmV, ORP modesYesYesYesYesYes
Ion concentration modeYesNoYesYesYes
Conductivity modeYesYesYesNoYes
Additional modesTDS, salinity,
resistivity
TDS, salinity,
resistivity
TDS, salinity,
resistivity
—TDS, salinity, resistivity
Dissolved oxygen modeYesNoYesYesNo
Compatible probesOptical,
polarographic
—Optical,
polarographic
Optical,
polarographic
—
Temperature range-5 to 105°C
23 to 221°F
-5 to 105°C
23 to 221°F
-30 to 130ºC
-22 to 266ºF
-30 to 130ºC
-22 to 266ºF
-30 to 130ºC
-22 to 266ºF
Number of channels 4, customizable
with modules
 4, customizable
with modules
322
Stirrer probe inputs22222
Display5.3-inch, backlit
color display
5.3-inch, backlit
color display
7-inch, high-
resolution, backlit
color display
7-inch, high-resolution, backlit color display7-inch, high-resolution, backlit color display
TouchscreenNoNoYesYesYes
Onscreen text promptsStep-by-step
calibration and
guided setup
instructions
Step-by-step
calibration and
guided setup
instructions
Step-by-step
calibration and
guided setup
instructions
Step-by-step calibration and
guided setup instructions
Step-by-step calibration and
guided setup instructions
Data log2000 with date/time2000 with date/time10000 with date/time10000 with date/time10000 with date/time
Calibration log30 per parameter30 per parameter20 per parameter20 per parameter20 per parameter
Identification optionsElectrode ID,
sample ID, user ID
Electrode ID,
sample ID, user ID
Electrode ID,
sample ID, user ID
Electrode ID,
sample ID, user ID
Electrode ID,
sample ID, user ID
Data export optionsUSB (2), RS232USB (2), RS232Computer, printer,
USB flash drive
Computer, printer,
USB flash drive
Computer, printer,
USB flash drive
Power supplyUniversal,
100–240 VAC,
50–60 Hz
(Cat. No. STARA-PWR)
Universal,
100–240 VAC,
50–60 Hz
(Cat. No. STARA-PWR)
Universal,
100-240 VAC,
50-60 Hz
(Cat. No. PSTAR-PWR),
9 DC adapter, 2.0A
Universal,
100-240 VAC,
50-60 Hz,
(Cat. No. PSTAR-PWR),
9 DC adapter, 2.0A
Universal,
100-240 VAC,
50-60 Hz
(Cat. No. PSTAR-PWR),
9 DC adapter, 2.0A 
Premium
Premium
Premium
 Orion Star A325
Multiparameter meter
(STARA3250 series)
Orion Star A326
Multiparameter meter
(STARA3260 series)
Orion Star A329
Multiparameter meter
(STARA3290 series)
 Premium STARA3250 series Orion multiparameter metersPremium STARA3260 series Orion multiparameter metersPremium STARA3290 series Orion multiparameter meters
 Rugged, dependable multiparameter meterRugged, versatile, and dependable multiparameter meterVersatile, powerful, multiparameter meter
Ideal for
applications in:
Industrial—cooling towers, wastewater; Environmental—ponds, lakesIndustrial—wastewater; Environmental—ponds, lakesIndustrial—wastewater, drinking water, food & beverage; Environmental—ponds, lakes
pH range–2.000 to 20.000–2.000 to 20.000–2.000 to 20.000
pH resolution0.1/0.01/0.0010.1/0.01/0.0010.1/0.01/0.001
pH relative accuracy±0.002±0.002±0.002
pH calibration pointsUp to 5Up to 5Up to 5
mV, RmV range±2,000.0 mV±2,000.0 mV±2,000.0 mV
EH/ORP modeYesYesYes
Dissolved oxygen modeNoYesYes
TDS relative accuracy±0.5% reading ±1 digit--±0.5% reading ±1 digit
Temperature range–5 to 105°C, 23 to 221°F–5 to 105ºC, 23 to 221ºF–5 to 105ºC, 23 to 221ºF
Number of datalogs5000, with time and date stamp5000, with time and date stamp5000, with time and date stamp
Data export optionsRS232, USBRS232, USBRS232, USB
AC adapterOptional—universal, 100–240VACOptional—universal, 100–240VACOptional—universal, 100–240VAC
Battery powerIncluded—4AAsIncluded—4AAsIncluded—4AAs
  Enhance your productivity with accessories for your benchtop and portable meters. Meter Accessories ›
Advanced
 Eutech PC2700 pH/Conductivity Bench Meter
 Economical, routine laboratory meters
pH range-2.000 to 20.000
pH resolution0.001 / 0.01 / 0.1
pH calibration pointsUp to 6
mV, RmV modesYes
Ion concentration modeYes
Conductivity modeYes
Additional modesTDS, salinity, resistivity
Range (temperature)0.0 to 100.0 °C, 32.0 to 212.0 °F
Backlit displayYes
Number of channels2
Data log500 with date/time
Calibration logMost recent viewable on meter and stored in meter memory
Data export optionsRS232
Power supplyUniversal, 100-240VAC, 50/60 Hz, Cat. No. 60X426401
Premium
Premium
Advanced
Advanced
Advanced
Advanced
 Eutech PD 450 Portable Multiparameter MeterEutech PC 450 Portable Multiparameter MeterEutech PCD 650 Portable Multiparameter MeterEutech PC 650 Portable Multiparameter MeterEutech PD 650 Portable Multiparameter MeterEutech CD 650 Portable Multiparameter Meter
pH range-2.00 to 16.00-2.00 to 16.00-2.00 to 20.00-2.00 to 20.00-2.00 to 20.00---
pH resolution0.01 pH0.01 pH0.1 / 0.01 Ph0.1 / 0.01 Ph0.1 / 0.01 Ph---
pH relative accuracy±0.01 pH±0.01 pH±0.01 pH±0.01 pH±0.01 pH---
pH calibration pointsUp to 6Up to 6Up to 6Up to 6Up to 6---
mV, RmV modesYesYesYesYesYes---
EH/ORP mode------YesYesYes---
Conductivity mode---YesYesYes---Yes
Dissolved oxygen modeYes---Yes---YesYes
TDS relative accuracy---±1 % full scale±1 % full scale±1 % full scale±1 % full scale±1 % full scale +
Ion concentration mode------YesYesYes---
Temperature range-10.0 to 110.0 °C (14.0 to 230.0 °F)-10.0 to 110.0 °C (14.0 to 230.0 °F)-10.0 to 110.0 °C (14.0 to 230.0 °F)-10.0 to 110.0 °C (14.0 to 230.0 °F)-10.0 to 110.0 °C (14.0 to 230.0 °F)-10.0 to 110.0 °C (14.0 to 230.0 °F)
Number of datalogs500 with date and time stamp500 with date and time stamp500 with date and time stamp500 with date and time stamp500 with date and time stamp500 with date and time stamp
Data export optionsUSB or RS232USB or RS232RS232C through LED, IrDARS232C through LED, IrDARS232C through LED, IrDARS232C through LED, IrDA
AC adapter9 V DC adapter (optional)9 V DC adapter (optional)9 V DC adapter9 V DC adapter9 V DC adapter9 V DC adapter
Battery power2 AA batteries (included)2 AA batteries (included)4 AA batteries (included)4 AA batteries (included)4 AA batteries (included)4 AA batteries (included)
Browse additional multiparameter products
Meter Accessories
find the right benchtop meter for your pH and ORP measurement needs
Shop our comprehensive range of meter accessories designed to boost productivity, safeguard your investment and ensure optimal performance. Our assortment of accessories for bench and portable meters helps streamline your workflow while enhancing your user experience.
Electrodes and Probes
Electrodes, probes, and sensors for use with conductivity meters
Obtain accurate results with electrodes and probes developed to support the requirements of your sample testing. Choose from a wide variety of parameters, body materials, body styles, fill types, reference systems, junctions and more to meet the needs of your application.
Standards and Solutions
Orion standards and solutions
Ensure the reliability of your measurements with our premium buffers, standards and solutions. Our high-quality calibration and maintenance solutions are essential for precise and consistent results. Shop a variety of buffers, buffer kits, standards, electrode filling solutions, electrode storage solutions and more.
Support
Knowledge Base: Certificates of Analysis and SDS for Orion, ROSS, AQUAfast, AquaMate, and AquaSensors products.
Orion Meters Software and Upgrades
Thermo Scientific Orion benchtop and portable meters are designed to help deliver accurate, consistent results, day after day. Boost productivity by upgrading your software today.
Find your software
Style Sheet for Global Design System