Search
Enhance your productivity while protecting your investment. We offer an assortment of meter accessories for our benchtop and portable meters.
Complement your benchtop meter with key accessories that make you more productive.
Uniformly mix solutions using five selectable stirring speeds with the meter-controlled Orion Star stirrer probe. Designed to fit into benchtop electrode stands, this stirrer probe can be easily positioned and quickly rinsed between samples. Our premium Versa Star Pro benchtop meters can operate two stirrer probes simultaneously, so you can measure your samples faster.
Included with every Thermo Scientific Orion Star A and Versa Star Pro benchtop meter, the electrode stand holds a variety of sensors for easy placement in and out of samples, and allows for consolidated rinsing between samples.
Because anything can happen in the field, add extra protection with durable portable meter armor. Included with all Orion Star A portable meter kits, the armor features dedicated holders for pH, conductivity, and dissolved oxygen sensors. An adjustable wrist/belt strap can secure the meter to fixtures such as pipes. The strap also converts into a built-in meter stand.
Our field case is specially designed to efficiently transport everything you need for field measurements. Sensors can stay connected with the meter while in the case. A removable door makes it easy to use the cable management system. The field case, which is included with all Orion Star A portable meter kits, helps you stay organized while protecting your instrumentation.
| Accessory category | Cat No. | Description | Meter type | Compatibility |
|---|---|---|---|---|
| 090043 | Orion Swing Arm and Electrode Holder, Swing and Base | Orion Dual Star, Star A111, Star A211, Star A214, Star A215, Star A216 and Versa Star Pro Meters | ||
| STARA-BEA | Orion Star A Series Benchtop Holder, Stand and Meter-attachment Bracket | Orion Dual Star, Star A111, Star A112, Star A113, Star A211, Star A212, Star A213, Star A214, Star A215, Star A216 and Versa Star Pro Meters | ||
| STARA-ELHD | Orion Star A Series Electrode Holder for Stands | Orion 090043 and STARA-BEA Stands | ||
| STARA-HB | Orion Star A Series Weighted Base | Orion 090043 and STARA-BEA Stands |
| Accessory category | Cat No. | Description | Meter type | Compatibility |
|---|---|---|---|---|
| Field carrying cases and armor | 1210005 | Orion Portable Soft-Sided Field Case | Orion Star A121, Star A122, Star A123, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, Star A325, Star A326 and Star A329 Meters | |
| STARA-AR | Orion Star A Series Portable Meter Protective Armor | Orion Star A121, Star A122, Star A123, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, Star A325, Star A326 and Star A329 Meters | ||
| STARA-CS | Orion Star A Series Portable Meter Hard Field Case | Orion Star A121, Star A122, Star A123, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, Star A325, Star A326 and Star A329 Meters | ||
| STARA-ESCD | Orion Star A Series Portable Holder for Conductivity & DO Probes | Orion STARA-AR Meter Armor | ||
| STARA-ESPH | Orion Star A Series Portable Holder for pH Electrode | Orion STARA-AR Meter Armor |
| Accessory category | Cat No. | Description | Meter type | Compatibility |
|---|---|---|---|---|
| Interface cables and computer software | 1010004 | Orion RS232 Computer and Printer Cables for Star A Series Meters | Orion Dual Star, Star A211, Star A212, Star A213, Star A214, Star A215, Star A216, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, Star A325, Star A326, Star A329 and Versa Star Pro Meters | |
| 1010005 | Orion RS232-to-USB Cable and Adapter for Star A Series Meters | Orion Dual Star, Star A211, Star A212, Star A213, Star A214, Star A215, Star A216, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, Star A325, Star A326, Star A329 and Versa Star Pro Meters | ||
| 1010053 | Orion RS232 Computer Cable for Star A Series Meters | Orion Dual Star, Star A211, Star A212, Star A213, Star A214, Star A215, Star A216, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, Star A325, Star A326, Star A329 and Versa Star Pro Meters | ||
| VSTAR-NPCD | Orion Navigator Pro Software on CD for Versa Star Pro Meters | Orion Versa Star Pro Meters | ||
| VSTAR-NPUSB | Navigator Pro Software on USB for Versa Star Pro Meters | Orion Versa Star Pro Meters | ||
| Power adapters and replacement parts | 1010003 | Orion Universal Power Adapter for Star A Meters | Orion Dual Star, Star A111, Star A112, Star A113, Star A121, Star A122, Star A123, Star A211, Star A212, Star A213, Star A214, Star A215, Star A216, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, Star A325, Star A326 and Star A329 Meters | |
| STARA-PWR | Orion Universal Power Adapter for Versa Star Pro Meters | Orion Versa Star Pro Meters | ||
| 090045 | Orion BNC Shorting Plug for pH Meters | Orion Dual Star, Star A111, Star A121, Star A211, Star A214, Star A215, Star A216, Star A221, Star A321, Star A324, Star A325, Star A326, Star A329 and Versa Star Pro pH Meters | ||
| Accessory category | Cat No. | Description | Meter type | Compatibility |
|---|---|---|---|---|
| Printers and accessories | STARA-106 | Orion Star A Series Ink Ribbon Printer 110–240V | Orion Dual Star, Star A211, Star A212, Star A213, Star A214, Star A215, Star A216, Star A221, Star A222, Star A223, Star A321, Star A322, Star A323, Star A324, Star A325, Star A326, Star A329 and Versa Star Pro Meters | |
| STARA-108 | Orion Ink Ribbons for Star A Series Printer, 6 Pack | Orion STARA-106 Printer | ||
| STARA-109 | Orion Paper Rolls for Star A Series Printer, 5 Pack | Orion STARA-106 Printer | ||
| 1010008 | Orion Ink Cartridge for 1010006 Inkjet Printer | Orion 1010006 Printer | ||
| 1010009 | Orion Paper Roll for 1010006 Inkjet Printer | Orion 1010006 Printer | ||
| PRT300 | Orion PRT300 Printer with Cable, 110V | Legacy Orion Aplus and PerpHecT Meters | ||
| PRT301 | Orion PRT300 Printer with Cable, 220V | Legacy Orion Aplus and PerpHecT Meters | ||
| PRT302 | Orion Printer Ribbon for PRT300 Printers | Orion PRT 300 Printer |
Find the pH benchtop or portable meter you need for your measurements. Orion pH meter selector tool
Download the Orion solutions brochure to learn about our premium, advanced, and basic meters and help identify the best match based on your required features, performance specifications, and budget.
Knowledge Base: Certificates of Analysis and SDS for Orion, ROSS, AQUAfast, AquaMate, and AquaSensors products.
Orion Meters Software and Upgrades
Thermo Scientific Orion benchtop and portable meters are designed to help deliver accurate, consistent results, day after day. Boost productivity by upgrading your software today.