On this page:
Designed to maximize process efficiency
Advanced Granulation Technology (AGT) supports connected cell culture workflows by offering a simple approach to media preparation, contributing to process reliability from development through manufacturing.
Using advanced granulation techniques, the AGT format facilitates the production of complete formulations in serum-free, protein-free, and chemically defined media. Each lot is pH- and osmolality pre-adjusted for rapid dissolution, helping shorten preparation time compared with conventional dry-powder media (DPM). By adopting AGT, you can gain the productivity of a ready-to-hydrate medium with the storage and transport efficiency of a dry format, supporting consistency and performance across bioprocessing workflows.
Benefits of using AGT in your workflow
Ordering information
Catalog # Name Size Price Qty
12529020 Each
129.00
12529012 Each
1,234.00
12529001 Each
8,570.00
12529003 Each
26,190.00
12559001 Each
6,450.00
12559003 Each
21,510.00
12559027 Each
108.00
12559019 Each
940.00
Supporting diverse applications and modalities
Gene therapy
AGT media support consistent viral vector production through rapid hydration and reproducible media performance from development to commercial scale.
Monoclonal antibodies (mAbs)
AGT media help maintain reliable cell growth and yield in mAb production, supporting scalability and consistency across bioprocessing workflows.
Vaccines
AGT media aim to simplify vaccine media preparation and reduce variability, promoting reproducible viral growth and efficient manufacturing from research to production.
Explore other media for consistent cell growth
CHO media
CHO media support reliable growth and productivity for biologics manufacturing. With serum-free, chemically defined formulations, these media help maintain controlled osmolality and nutrient delivery for performance across scales.
Diploid growth and production media
Support vaccine development by minimizing process variability and serum use. Designed for scalability, these formulations promote reliable cell growth and viral yield.
Virus production media and reagents
Facilitate scalable vaccine manufacturing through serum-reduced formulations that promote viral growth and reproducibility from research to production.
Discover other cell culture solutions to scale processes
Advancing cell culture processes depends on bioprocessing solutions that support growth, productivity, and control. Our solutions include feeds and supplements, functional additives and reagents, and cell culture equipment to build a connected workflow that supports consistent scale-up and reliable performance.
Find the right media formulation for your process
Every bioprocess has distinct performance needs based on cell line, scale, and workflow design. Custom media development seeks to optimize growth, productivity, and quality across the entire research-to-manufacturing spectrum. Our specialists collaborate with you to adapt existing formulations or design media for advanced applications, helping reduce variability and improve scalability across production stages.
Frequently asked questions
AGT is a granulated dry-medium format produced using advanced granulation techniques designed to simplify cell culture preparation and improve process consistency. Each AGT formulation contains complete, pre-adjusted nutrients in granular form that dissolve quickly in water for uniform hydration. Produced under cGMP conditions, AGT supports reliable growth across development and manufacturing workflows.
AGT media replace conventional dry powder media (DPM) blends with a single, complete granular medium, eliminating the need for multi-component weighing and mixing. The granules are pre-adjusted for pH and osmolality, designed to dissolve rapidly to help shorten preparation time and reduce variability. Incorporating AGT media into your workflow allows for lower storage and handling needs because the format requires fewer containers, and incoming QC is simplified. Hydration behavior is predictable across scales, supporting consistent performance.
AGT media make it easy to move from small-scale liquid processes to large-scale production. This format offers the same performance as liquid media with the added benefit of being simpler to store, ship, and mix - just add water. This facilitates faster, cleaner, and more efficient scaling without changing your process.
AGT media are designed for quick preparation. The granules are dissolved directly in water for injection or cell culture-grade water, eliminating the need for separate pH and osmolality adjustments. Once fully dissolved, the solution is sterile filtered to create ready-to-use liquid media. This process helps reduce preparation time and handling steps while maintaining outcomes across batches and scales.
AGT media are manufactured under cGMP conditions and meet rigorous quality control standards for traceability, documentation, and reproducibility. Each lot is thoroughly tested to confirm its physical, chemical, and functional consistency, making it suitable for use in regulated bioprocessing environments. The dry format simplifies inventory management and preparation within cGMP-compliant facilities.
Cell culture resources
Access technical resources curated to optimize cell culture performance and scale processes confidently. Review guidance on workflow design, media selection, and process optimization to support productivity and consistency across applications.
Enable accelerated bioprocessing workflows with AGT media
Gibco Cell Culture Media
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.