Rhodium • Transition Metal
Primary XPS region: Rh3d
Overlapping regions: Mg KLL
Binding energies of common chemical states:
Chemical stateBinding energy Rh3d5/2
Rh metal307.6 eV
Native oxide308.8 eV
Experimental information
N/A
Interpretation of XPS spectra
  • Rh3d region has well-separated spin-orbit components (Δmetal=4.7eV).
    • The Coster-Kronig effect is observed, broadening the Rh3d3/2 component compared to Rh3d5/2, and making the Rh3d3/2 component shorter than expected.
  • There is a very strong overlap between the Ru3d and C1s regions.
    • RhO2 may also have asymmetric peak shapes in some crystalline phases [1].
element-rhodium-xpsspectra-300x450
Reference
  • [1] Y. Abe et al., Surface Science Spectra, Vol. 8, No. 2, 2001.
About this element
Element Crystal cub
Symbol: Rh
Date of discovery: 1803
Name origin: Greek rhodon
Appearance: silver-white
Discoverer: William Wollaston
Obtained from: platinum ores
Melting point: 2237 K
Boiling point: 3968 K
Density[kg/m3]: 12450
Molar volume: 8.28 × 10-6 m3/mol
Protons/Electrons: 45
Neutrons: 58
Shell structure: 2,8,18,16,1
Electron configuration: [Kr]4d85s1
Oxidation state: 2,3,4
Crystal structure: cubic
This silvery-white hard transition metal belongs to the platinum group. It has high reflectance and is hard and durable. Rhodium is primarily used as an alloying agent with platinum and iridium to improve high temperature strength and oxidatlion resistance. These metal alloys are employed in furnace windings, electrical contacts, and high temperature resistance wires. As the pure metal, rhodium’s brilliance and resistance to tarnish makes it well suited for plating jewelry. This metal was discovered in 1803 by W. Wollaston and is named from the Greek origin "rhono", meaning rose, after the rosy-colored solution yielded by its salts.
element_rhodium
Products
Nexsa G2 XPS
  • Micro-focus X-ray sources
  • Unique multi-technique options
  • Dual-mode ion source for monoatomic & cluster ion depth profiling
Style Sheet for Instrument Cards Original
K-Ahttps://lpha XPS
High resohttps://lution XPS Fast, efficient, automated workfhttps://low Ion source for depth profihttps://ling
ESCALAB QXi XPS
  • High spectral resolution
  • Multi-technique surface analysis
  • Extensive sample preparation and expansion options
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.