Zirconium • Transition Metal
Primary XPS region: Zr3d
Overlapping regions: B1s
Binding energies of common chemical states:
Chemical stateBinding energy Zr3d5/2
Zr metal178.9 eV
Zr sub-oxides179-180.5 eV
ZrO2182.3 eV
Zr(IV) silicate182.7 eV
Experimental information
N/A
Interpretation of XPS spectra
  • Zr3d region has well-resolved spin-orbit components (Δmetal=2.4eV).
    • Spin-orbit splitting changes with chemical state.
  • Zr3d peaks for Zr metal have an asymmetric peak shape.
    • The same is true for conducting zirconium compounds, such as ZrB2.
Zr3d Spectrum Sputter Damaged ZrO2
General comments
  • Zr is a reactive metal and usually found as oxide, ZrO2.
  • The oxide is reduced to sub-oxides by argon ion bombardment.
  • Commonly seen as minor components in Hf-containing compounds.
Element Crystal Hex
About this element
Symbol: Zr
Date of discovery: 1789
Name origin: Arabic zarkûn
Appearance: silvery white
Discoverer: Martin Heinrich Klaproth
Obtained from: zircon
Melting point: 1852 K
Boiling point: 4490 K
Density[kg/m3]: 6.52
Molar volume: 14.02 × 10-6 m3/mol
Protons/Electrons: 40
Neutrons: 51
Shell structure: 2,8,18,10,2
Electron configuration: [Kr]4d25s2
Oxidation state: 4
Crystal structure: hexagonal
At elevated temperatures, this silvery white metal can ignite spontaneously in air. It is found in S-type stars and has been identified in our own sun, as well. Zirconium is used in flash bulbs for photography, explosive primers, and lamp filaments. It is also used as the main ingredient of cubic zirconium stones. Similar in appearance to diamonds, cubic zirconium stones are sold as an affordable alternative to the more precious diamond. Although zirconium does not play a biological role, human tissue does assimilate the element, making it suitable for some artificial joints and limbs.
Zirconium • Transition Metal
Products
Nexsa XPS
  • Módulo de inclinação para medições ARXPS
  • Fonte de íons de modo duplo para recursos de perfil de profundidade expandida
  • Análise de isolantes
Folha de estilo para cartões de instrumentos original
K-Alpha XPS
  • Espectroscopia de área selecionável
  • Monocromador microfocalizado
  • Espectroscopia de estado químico de alta resolução
ESCALAB Xi+ XPS
  • Espectroscopia de alta sensibilidade
  • XPS com raios X não monocromáticos
  • Analisador de energia hemisférica de 180 graus
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Folha de estilo para especificações de tabela de produtos
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Folha de estilo para especificações de tabela de produtos
Fix for left nav
Folha de estilo para Rodapé de suporte e serviço
Folha de estilo para Fontes
Folha de estilo para Cartões
Serviços de microscopia eletrônica para
a ciência dos materiais
Para garantir o desempenho ideal do sistema, fornecemos acesso a uma rede de especialistas em serviços de campo, suporte técnico e peças de reposição certificadas.