Unlabeled Glycan Workflow
Label-free analysis of N-glycans and O-glycans
Glycans are often derivitized prior to analysis, but it is also possible to analyze both N-glycans and O-Glycans in a label-free form, typically using ion chromatography (IC) or liquid chromatography with charged aerosol detection. IC technique is sensitive and selective and resolves glycans according to charge, size, composition, isomers and linkage. As an alternative, liquid chromatography with the correct choice of detector can also be used.
Featured unlabeled glycan analysis products
Unlabeled glycan analysis workflow
Unlabeled glycan analysis workflow
Unlabeled N-glycosylation and O-glycosylation analysis categories
Universal native released glycan detection
The new Thermo Scientific Vanquish Flex UHPLC platform offers the ultimate in biocompatible state-of-the-art binary or quaternary solvent blending. The Vanquish Flex platform is designed to deliver new benchmarks in accuracy, precision, and sensitivity in peptide chromatography. With rock steady retention times, you can be confident in the identity of every peptide in discovery, development and QC environments.
Ion chromatography for N-linked and O-linked glycan analysis
High performance anion-exchange chromatography with pulsed amperometric detection (HPAE-PAD) on systems such as the Dionex ICS-6000 Hybrid HPIC System, is a well-established analysis method to characterize released Glycans. The separation is exceptionally selective for sugars and resolves glycans based on charge, size, composition, isomers and linkages. It is even possible to separate glycans based on sialic acid linkage, providing valuable information not only about the sugar sequence of a glycan, but also the subtle linkage differences that may indicate disease states.
Featured videos