Search
Thermo Fisher Scientific offers a range of instruments to support your characterization workflow from sample preparation to data analysis. With focused ion beam scanning electron microscopes that quickly and precisely prepare even your most delicate samples, transmission electron microscopes that collect detailed structural and chemical data, and intuitive software that ties them all together, you have a suite of tools that can help you look deeper into the nano-universe.
Sample preparation is one of the most critical tasks in nanoscale materials analysis.
Thermo Scientific Scios and Helios DualBeam Focused Ion Beam Scanning Electron Microscopes automatically prepare a wide range of samples—including air-, moisture-, temperature-, and beam-sensitive materials—in as little as 45 minutes. With high-quality samples free of defects and contamination, you can be sure you will collect accurate, reliable nanoscale data. With over 30 years of experience and more than 2,000 systems installed worldwide, our DualBeams lead the market with innovative features and comprehensive application knowledge.
Explore an optimum solution for your application from our versatile range of DualBeam FIB-SEMs that offer:
![]() |
![]() |
![]() |
![]() |
![]() |
|
| Instrument | Scios 2 FIB-SEM | Helios 5 FIB-SEM | Helios 5 PFIB-SEM | Helios 5 Hydra PFIB-SEM | Helios 5 Laser PFIB-SEM |
| Milling | Ga+ | Ga+ | Xe+ | Xe+, N+, O+, Ar+ | Xe+, N+, O+, Ar+, Laser |
| Imaging | UHR, LoVac | XHR, HiVac | XHR, HiVac | XHR, HiVac | XHR, HiVac |
| Cross-sectioning and 3D volumes | ≥ 10 µm | ≥ 10 µm | ≥ 100 µm | ≥ 100 µm | ≥ 1 mm |
| TEM sample prep | AutoTEM 5 Basic | AutoTEM 5 Auto | AutoTEM 5, Ga-free | AutoTEM 5, Ga-free | AutoTEM 5, Ga-free |
| Analytical capabilities | EDS, EBSD, SIMS, CL, Raman | ||||
| View product | View product | View product | View product | View product | |
Transmission electron microscopes deliver unparalleled resolution to help you see nanoscale structures and chemical information within your materials.
Thermo Scientific Talos and Spectra TEMs include enhanced electron sources and advanced detectors that deliver accurate, high-quality images along with chemical and structural data that provide unambiguous results. Explore optimum solutions for your application from our unmatched range of transmission electron microscopes.
![]() |
![]() |
![]() |
![]() |
![]() |
![]() |
|
| Instrument | Talos 12 TEM | Talos F200C TEM | Talos F200 i/X TEM |
Spectra 200 S/TEM | Spectra 300 TEM | Spectra Ultra S/TEM |
| Electron gun | Tungsten, LaB6 | S-FEF, X-FEG | S-FEF, X-FEG, X-CFEG | X-CFEG | X-CFEG, X-FEG / Mono, X-FEG / UltiMono | X-CFEG, X-FEG / Mono, X-FEG / UltiMono |
| Cs Correctors | No | No | No | S/TEM | S/TEM, TEM | S/TEM, TEM |
| HR STEM (nm) | 1 | 0.2 | 0.16 / 0.14 | 0.06 (ST) / 0.07 XT) | 0.05 (ST) / 0.06 (XT) | 0.05 |
| Pole piece distance (mm) | 11 | 11 | 5.5 | 5.4 (ST) / 6.4 (XT) | 5.4 / 6.4 | 5.4 |
| EDX capabilities (solid angle) | Bruker 30 / 100 (0.18 / 0.8) | Bruker 30 / 100 (0.18 / 0.8) | Super-X / DualX (0.9 / 1.65) | Super-X / DualX (0.7 / 1.76) | Super-X / DualX (0.7 / 1.76) | Ultra X (4.05) |
| EELS capabilities | No | Yes | Yes | Yes | Yes | Yes |
| Cryo capabilities | Cryo box | Cryo box | Cryo holders (XT) | Cryo holders | Cryo box (XT) | Cryo holders |
| Room dimensions (H x L x W) | 2.6 m x 3.5 m x 2.96 m 2.6 m x 2.85 m x 2.96 m |
3.03 m x 4.9 m x 3.9 m | Talos i: 2.8 m x 4 m x 3.6 m Talos X: 3.03 m x 4.9 m x 3.9 m |
3/3.3 m x 6.5 x 6.1 | 3/3.3/3.7 m x 6.5 m x 5.1 m | 3/3.3/3.7 m x 6.5 m x 5.1 m |
| Thermo Scientific Velox Software, oil-free vacuum system, remote control, scripting interface | ||||||
| View product | View product | View product | View product | View product | ||
For Research Use Only. Not for use in diagnostic procedures.