Search
TOPO cloning kits with Mach1-T1R competent cells make cloning fast and simple. Mach1 cells deliver robust growth for miniprep analysis in just four hours, saving up to 2 days compared to restriction enzyme and ligation cloning.
We offer a suite of TOPO cloning kits with Mach1-T1R competent cells for basic cloning of Taq-amplified fragments, blunt-ended fragments, sequencing, long-fragment PCR cloning, and entry into the Gateway system. Simply add your PCR product (blunt-ended fragment or amplified using Taq polymerase) to the appropriate vector, incubate for 5 minutes and transform into our fast growing Mach1 competent cells.
Invitrogen's Mach1-T1RE. coli achieve sufficient growth for miniprep analysis from an overnight colony in just four hours (Figure 1) compared to conventional strains which require an overnight incubation.
Mach1-T1R cells offer several advantages:
Mach1 competent cells are also available with Zero Blunt and TOPO XL vectors:
Have any questions on competent cells or transformation? Click on the resources listed below to access overviews, videos, genotype guides, and educational resources.
Review rich educational content on transformation workflow, troubleshooting, strategies, and selection guides.
Stay up-to-date on industry trends in molecular cloning with on-demand webinars.
Access white papers and technical and application notes on molecular cloning.
Watch instructional videos on troubleshooting tips, how-to videos, and explainers related to molecular cloning.
For Research Use Only. Not for use in diagnostic procedures.