Search
Gel Electrophoresis Image Analysis |
Invitrogen iBright Analysis Software is precast gel electrophoresis analysis software that offers a convenient way to analyze the DNA bands on E-Gel precast agarose gels. Images captured as .g2i format can be imported directly from E-Gel Power Snap Plus device or from a computer for analysis. iBright Analysis Software supports image analysis of both 96-well and 48-well precast gels.
iBright Analysis Software, compatible with both Mac and Windows operating systems, is available as a cloud-based solution via the Thermo Fisher Connect platform and as a free desktop application.
Automatic well detection for deconvolution.
Automatic band detection for intensity analysis.
The iBright Analysis Software has image deconvolution. This allows the user to align the lanes and read the results better. The 96-well E-Gels have a staggered well distribution for optimal running distance; with the iBright Analysis Software users can reconstruct the 96-well E-Gel images into a linear format for readability. The user can also adjust the frames utilized for deconvolution, to get the best fit for things like like contrast and brightness.
iBright Analysis Software detects the DNA bands on the gel and analyzes their intensity. The software has multiple user adjustment settings, and the data on band intensities can be exported in a spreadsheet.
The intuitive and user-friendly iBright Analysis Software includes features such as, organizing and editing the reconstructed images, storing data from precast gel electrophoresis image analysis, and creating reports.
E-Editor 2.0 Software is gel electrophoresis analysis software for analyzing images of stained precast gels and blots of E-PAGE gels and E-Gels. The E-Editor 2.0 software is available as a desktop application compatible with both Mac and Windows operating systems.
E-Editor 2.0 software is included with the purchase of E-Gel or E-PAGE 96 gels and related equipment. Download a copy of the E-Editor 2.0 software and manual below.
Windows E-Editor Version 2.0 for Windows, for configuring E-Gel and E-PAGE Gel Images: Compatible with Windows 98, NT, 2000 and XP. | Macintosh E-Gel 96 Editor Version 1.5 for Macintosh, for configuring E-Gel 96 Gels: Compatible with OS 8.5 and above (OS X with OS 9 emulation only)
|
After installation on your computer, open your image file (.tif, .jpg, or .bmp) in the E-Editor 2.0 program, align the lanes, and arrange. Save the reconfigured image or copy and paste selected lanes into other applications for further analysis.
The E-Editor 2.0 includes image editing features, such as:
By your use of the E-Editor or E-Gel 96 Editor Software, you accept and acknowledge the following terms and conditions:
You agree not to de-compile, disassemble, reverse-engineer, or in any way modify the software or merge it into another program.
THIS SOFTWARE IS PROVIDED "AS IS", AND ANY EXPRESS OR IMPLIED WARRANTIES INCLUDING, BUT NOT LIMITED TO, THE IMPLIED WARRANTIES OF MERCHANTABILITYAND FITNESS FOR A PARTICULAR PURPOSE ARE DISCLAIMED. IN NO EVENT SHALL INVITROGEN CORPORATION BE LIABLE FOR ANY DIRECT, INDIRECT, INCIDENTAL, SPECIAL, EXEMPLARY, OR CONSEQUENTIAL DAMAGES (INCLUDING, BUT NOT LIMITED TO, PROCUREMENT OF SUBSTITUTE GOODS OR SERVICES, LOSS OF USE, DATA, OR PROFITS, OR BUSINESS INTERRUPTION) HOWEVER CAUSED AND ON ANY THEORY OF LIABILITY, WHETHER IN CONTRACT, STRICT LIABILITY, OR TORT (INCLUDING NEGLIGENCE OR OTHERWISE) ARISING IN ANY WAY OUT OF THE USE OF THIS SOFTWARE, EVEN IF ADVISED OF THE POSSIBILITY OF SUCH DAMAGE. IF ANY PROVISION OF THIS AGREEMENT IS FOUND TO BE LEGALLY UNENFORCEABLE, THEN THAT PROVISION WILL BE DELETED FROM THIS AGREEMENT, AND THE REMAINDER OF THE AGREEMENT WILL CONTINUE IN FULL FORCE AND EFFECT.
Watch educational and insightful videos on molecular biology techniques.
View live and recorded webinars on research trends in molecular biology.
Access white papers, application notes, and technical notes on molecular biology.
Find the right E-Gel precast gels and DNA ladders for your applications.
For Research Use Only. Not for use in diagnostic procedures.