Search
Ribonuclease (RNase) Free Supplies
|
Trace amount of ribonuclease (RNase) contamination can sabotage your laboratory experiments. RNA is known to be difficult to work with because it is readily degraded by RNases that are ubiquitous in the environment, including on our hands. Utilizing certified RNase-free supplies and reagents can help protect your experimental results.
Manufactured with rigorous nuclease (includes RNase) contamination controls that include stringent quality specifications, our products are designed to meet the needs of researchers working with RNA. Our nuclease-free products are tested to help ensure that no traces of contaminating nonspecific endonuclease, exonuclease, and RNase activities are present.
Presence of nucleases such as DNase and RNase can degrade a molecular sample, interfering with downstream analysis. Researchers working with RNA know that RNA is unstable and degrades rapidly. Even trace quantities of RNases can lead to lower yields from in vitro transcription reactions, degradation during RNA purification protocols, and produce variable results after RT-qPCR. It is vital to protect targeted molecular samples from the RNases that can break them down. All labware should be RNase-free before commencing molecular research; this includes instruments, surfaces, protective wear, and consumables involved.
When used to describe reagents or labware items, RNase-free means that the item is free of enzymes called ribonucleases, which degrade ribonucleic acid (RNA). In molecular biology and biochemistry, maintaining an RNase-free environment is crucial for experiments involving RNA, such as RNA extraction, reverse transcription, and RNA sequencing.
To achieve an RNase-free environment, special precautions must be taken including the use of RNase-free reagents, consumables, and equipment, as well as rigorous cleaning and handling protocols to prevent contamination.
Surfaces including benchtops, pipettes, glassware, and benchtop instruments are common sources of RNase contamination in the lab and should be treated with a surface decontamination agent. For instance, if sterile gloves have contacted bare skin, touched a refrigerator handle, door handle, pipettor, pen or pencil, or any phone, the gloves should subsequently not be considered RNase-free. For more details, you can learn about the basics of RNase control or explore the surface decontamination products provided by Thermo Fisher Scientific.
RNase-free products can be broken down into four categories: RNase inhibition, RNase detection, RNase decontamination, RNase stabilization.
Because of the importance of keeping lab spaces nuclease-free, there are many products to help inhibit, detect, and decontaminate RNases, as well as preserve and stabilize RNA. Products that can help make your laboratory RNase-free:
To determine if RNases are present in samples or solutions, RNase detection kits are used. These kits typically monitor RNase activity.
Decontamination reagents eliminate nuclease contamination (particularly ribonuclease) on work surfaces.
RNase stabilizers are reagents that help stabilize and preserve the integrity of RNA at the point of collection or post-collection.
Another way to keep your laboratory RNase-free is to utilize certified nuclease-free and RNase-free supplies and reagents. View our catalog of nuclease-free tubes, tips, water, buffers, and reagents:
Certified nuclease-free (RNase- and DNase-free) tubes and pipette tips designed for handling RNA and DNA with confidence.
Certified nuclease-free water products available in a wide range of sizes and packaging formats for molecular biology research.
RNA and DNA storage buffers, general-use buffers, and molecular biology-grade reagents—all rigorously tested and shown to be nuclease-free.
In support of our commitment to providing the highest-quality products, information, and services, our site in Austin, Texas has obtained ISO 9001:2000 registration. The achievement of ISO 9001 registration assures that quality systems and procedures have been developed and implemented in product design, production, and support operations. ISO 9001 certification is a strategic milestone in our ongoing efforts to effectively support customers. BSI, Inc., the world's largest registration body for management systems, is the registrar for Ambion quality systems.
Our experts are here to help with application support, method development, and any other sample preparation questions you may have. Don't work in isolation. Contact us directly at techsupport@thermofisher.com.
Technical bulletins
TechNotes articles
For Research Use Only. Not for use in diagnostic procedures.