Trust your samples to an outstanding, reliable shaker
Benefit from around-the-clock operation with the reliable, flexible Thermo Scientific MaxQ Orbital Shakers. These easy-to-operate shakers deliver outstanding performance, accuracy, and reproducible results – especially when working within controlled temperature environments. While your cells flourish and your cultures agitate you may find yourself with less worry over managing your shakers and more time to focus on your 
Features of MaxQ Orbital Shakers
Reliable operation
Our outstanding drive mechanisms provide continuous 24-hour operation and allow you to shake large and uneven loads, smoothly and uniformly. These drive mechanisms are available on all MaxQ Orbital Shakers and come with a 10-year warranty.
Easy to configure
Easy-to-use controls are designed to precisely adjust individual displays for temperature, speed and time. View all parameters simultaneously to prevent risk of error while establishing running conditions.
Designed for flexibility
Incubation, refrigeration, and CO₂ resistance are hallmarks of these lab workhorses. They include configurable platforms for maximum flexibility and capacity. Platforms allow the interchangeability of clamps that hold a variety of lab consumables.
MaxQ incubated and refrigerated shakers
The MaxQ 8000 models are stackable and come in either incubated or incubated and refrigerated options.
Features include:
  • Stack two to three units high for space savings with a speed range of 25 to 400 rpm ±1 rpm
  • HEPA filter evenly distributes air inside the chamber and reduces cross contamination improving culturing conditions; rated at 95% efficient at 0.3 microns for entrapment of airborne particles
  • Ergonomically designed fold down door and a platform that fully slides out
    - Maximum platform height: 154.9 cm / 61 in.
    - Interior chamber comfortably fits 2L Erlenmeyer flasks
  • Maintenance can be performed from the front of the unit without moving or unstacking.
  • Stainless steel interior and built-in drain make cleaning easy
  • Remote alarm comes standard
MaxQ incubated and refrigerated shakers specifications
Shaker Model
MaxQ 8000 Incubated Shaker MaxQ 8000 Refrigerated & Incubated Shaker
Cat. Nos.
SHKE8000 (120V)
SHKE8000-1CE (240V)
SHKE8000-7 (120V)
SHKE8000-8CE (240V)
Orbital diameter
1-in. / 25.4 mm
1 in. / 25.4 mm
Platform size
29.5 x 18 in. / 74.9 x 45.7 cm
29.5 x 18 in. / 74.9 x 45.7 cm
Unit weight
658 lbs. / 298.5 kg
723 lbs. / 328 kg
Largest flask size (using adhesive mats)
2.8 L
2.8 L
Max capacity of largest flask (using adhesive mats)
6
6
Max load capacity
71 lbs. / 32. 2 kg
71 lbs. / 32. 2 kg
Speed range
25 – 400 ± 1 rpm
25 – 400 ± 1 rpm
Temperature range
Ambient +10 – 60 °C
Ambient -5 – 60 °C
View the selection guide for additional orbital shaker models
MaxQ CO₂ resistant shaker
The Thermo Scientific MaxQ 416 High Performance Table Top Orbital Shaker is designed for long-term use on the bench or inside a controlled temperature environment, such as a CO2 incubator.
Features include:
  • Choice of two platform sizes maximizes capacity
  • Low heat motor with moisture resistant electronics provides continuous maintenance-free operation on the benchtop or in a chamber
  • Operates within temperature range of 4°C to +40°C and humidity conditions from 50 to 80% non-condensing
  • Durable tri-eccentric balanced drive systems for optimum handling of full or unbalanced loads regardless of vessel placement completely eliminates walking
  • Oversized, adjustable suction cup feet minimize vibration
  • Remote alarm comes standard
Shop for MaxQ 416 HP Shaker and Platform
View more details on the MaxQ HP Table Top Orbital Shaker or add to cart now. Platforms and clamps sold separately. 
View the selection guide for additional orbital shaker models
Shop for MaxQ Shaker Accessories
Use the tabs below to shop for MaxQ Shaker Accessories. The MaxQ 8000 Models and MaxQ 416HP Table Top Shaker use the same clamps, test tube and rack trays and screws. All of which can be ordered below. 
Catalog # Name Size Price Qty
238000 Each
1,413.00
Online Exclusive
1,570.00 Save 157.00 (10%)
238008 Each
1,548.00
Online Exclusive
1,720.00 Save 172.00 (10%)
236099 Each
864.00
Online Exclusive
960.00 Save 96.00 (10%)
236025 Each
47.07
Online Exclusive
52.30 Save 5.23 (10%)
236011 Each
42.30
Online Exclusive
47.00 Save 4.70 (10%)
236012 Each
41.94
Online Exclusive
46.60 Save 4.66 (10%)
236013 Each
45.09
Online Exclusive
50.10 Save 5.01 (10%)
236014 Each
76.05
Online Exclusive
84.50 Save 8.45 (10%)
236015 Each
89.46
Online Exclusive
101.00 Save 11.54 (11%)
236016 Each
112.50
Online Exclusive
125.00 Save 12.50 (10%)
236019 Each
188.10
Online Exclusive
209.00 Save 20.90 (10%)
300349 Each
78.21
Online Exclusive
86.90 Save 8.69 (10%)