Search
Cell Therapy SoftwareGibco CTS Cellmation Software for DeltaV System |
Gibco CTS Cellmation software is an off-the-shelf solution designed to help simplify and optimize cell therapy manufacturing processes by providing digital connectivity for modular cell therapy instrumentation offered by Thermo Fisher Scientific. The software’s open platform provides a flexible system for managing and integrating multiple instruments, streamlining the cell therapy workflow, and enabling 21 CFR Part 11/Annex 11 regulatory compliance. CTS Cellmation Software is powered by DeltaV Distributed Control System (DCS) and has been developed following GAMP5 methods to help ensure compatibility with cGMP-compliant processes. CTS Cellmation Software is optimized to easily scale from process development through clinical manufacturing and commercialization.
In this webinar, we introduce our closed, modular, end-to-end manufacturing process, as well as demonstrate how our instruments can be integrated and automated into existing workflows. Gain insights into current challenges in the field and how automation can help overcome these to improve the current state-of-the-art workflows.
The CTS Cellmation Software helps to enable automation of your cell therapy manufacturing processes by connecting multiple Thermo Fisher Scientific instruments across your workflow. Below are the instruments the CTS Cellmation Software can connect to and control:
The Cellmation software is compatible with Emerson DeltaV DCS and specific Thermo Fisher Scientific cell therapy instruments. See if this solution is the right fit for your workflow.
To review Release Notes and download the latest software for any of the Cellmation modules, visit the software download page.
In this video, you will see an overview of how the Gibco CTS Cellmation Software automates and controls the workflow on day 3 of the manufacturing process, leveraging CTS Xenon Electroporation System, CTS Rotea Counterflow Centrifugation System, and CTS DynaCellect Magnetic Separation System.
The multipurpose Gibco CTS Rotea Counterflow Centrifugation System offers exceptional flexibility in a cell therapy development and manufacturing system. The Cellmation Software is now available for CTS Rotea Counterflow Centrifugation System.
The CryoMed CRF features enhanced data traceability via a touch screen user interface and offers customizable freezing profiles. CTS Cellmation Software enables 21 CFR Part 11 compliance and allows you to connect your cell therapy workflow.
Contact us to discuss available service contracts that fit your specific needs. Service contracts provide both onsite and remote support. Installation is performed by Thermo Fisher Scientific representatives with remote support offered 12 hours per day, 5 days a week. Technical Support can be reached Monday-Friday between 8 AM to 8 PM EST.
To get the latest software for the Cellmation Software, see our software download page.
Intended use of the products mentioned on this page vary. For specific intended use statements please refer to the product label.