Designed by cell sorting experts for high-performance, high-throughput cell sorting, the Invitrogen Bigfoot Spectral Cell Sorter is a fluorescence-activated cell sorter (FACS) tailored to enhance your cell sorting capabilities and fit easily into your workflow. Equipped with up to 9 lasers and 60 detectors, it allows you to perform your existing experiments using conventional compensation or leverage its spectral analysis flow cytometry capabilities to resolve even more cell populations. Integrated biocontainment and aerosol management help ensure the protection of both you and your samples.
The Bigfoot Spectral Cell Sorter offers high-speed and high-throughput sorting, capable of processing a 96-well plate in just 11 seconds and a 384-well plate in 20 seconds. It is designed to be simple enough for an individual lab and robust enough for a core facility. The Bigfoot Spectral Cell Sorter empowers you to master a wide range of sorting experiments, including deep immunophenotyping, genomics applications, cell and gene therapy research, single-cell sorting, and other high-performance, high-throughput cell sorting applications.
Conventional or spectral analysis
Migrate your flow cytometry experiments and protocols directly using conventional compensation, or apply spectral unmixing to expand your parameters and discovery.
Flexible cell types and sorting applications
Sample from your choice of tubes and sort into tubes, plates, slides, or a genomics chip. Especially well suited for applications requiring high performance and high-throughput.
How cell sorting is becoming an even more powerful tool
Cell sorting using flow cytometry is important in immunology research, with spectral unmixing enhancing the precision and depth of cell analysis.
Spectral Flow Cytometry Fundamentals
Learn about the history of spectral flow cytometry, the players in the marketplace, and the technological differences among spectral instrumentation. Whether you are a beginner or an advanced user, these fundamentals will help you conceptualize advances in spectral flow cytometry and how to think about designing experiments for a spectral cytometer.
Experience exceptional precision and speed with the Invitrogen Bigfoot Spectral Cell Sorter from Thermo Fisher Scientific. Designed for advanced flow cytometry applications, this innovative instrument offers high-throughput sorting, enhanced sensitivity, and spectral analysis capabilities. An excellent choice for researchers focused on cell sorting and analysis, the Bigfoot Spectral Cell Sorter helps ensure sample integrity and reproducibility. Discover how this advanced technology can elevate your lab's performance in cell analysis.
Flow Cytometry Panel Design Service & Panel Builder Tool
Immunology at Work Resource Center
Antibodies for Flow Cytometry
Antibodies for Flow Cytometry
Register for flow webinar series
Latest promotions and deals
For Research Use Only. Not for use in diagnostic procedures.