Search
Each benchtop KingFisher purification instrument is designed for processing at different throughput levels, volume ranges, and samples per run. Decide which automated purification system is right for your laboratory research or testing.
| KingFisher instrument |
|
|
|
|
| Instrument size | Compact benchtop | Benchtop | Benchtop | Benchtop—integrates with robotic liquid handler |
| Throughput level | Low to medium | Medium to high | Medium to high | Ultrahigh |
| Processing volume range |
|
|
|
|
| Samples per run | 6 or 12 | 24 or 96 | 24 or 96 | 24 or 96 |
| Customizable protocols | Yes, with PC software | Yes, with PC software | Yes, with touchscreen or PC software | Yes, with PC software |
| Heating/cooling |
|
|
|
|
| Ultraviolet lamp | 8 watts (up to 16 hr) | No | 2 UV lamps, max 23 h 59 min | No |
| Size | 40 x 46 x 34 cm; 15.7 x 18.1 x 13.4 in. (W x D x H) | 68 x 60 x 38 cm; 26.77 x 23.6 x 14.96 in. (W x D x H) | 78 x 59 x 45 cm; 31 x 23 x 18 in. (W x D x H) | 36 x 46.5 x 40 cm; 14.2 x 18.3 x 15.7 in. (W x D x H) |
|---|---|---|---|---|
| Weight | 17 kg; 37.5 lbs. | 28 kg; 62 lbs. | 56 kg; 123 lbs. | 24.0 kg; 52.9 lbs. |
| Ordering information |
|
The main differences are throughput, the number of samples per run, and the way the models handle the plates and process.
KingFisher instruments are versatile sample preparation instruments in the lab, and are elegantly designed to support multiple applications.
Various throughputs—process 6–96 samples per run depending on the instrument model
Interchangeable formats—choose 24- or 96-well plates so you can process a wide range of input volumes
Protocol customization—easily edit, modify, or create new protocols
Optimized reagents—compatible with multiple magnetic-bead reagents
When you buy a refurbished KingFisher automated sample purification instrument from Thermo Fisher Scientific, you're not only getting the speed, accuracy, and reliability you trust, you're also get the confidence that comes from knowing you have our highly experienced service and support team behind you.
Refurbished KingFisher instruments purchased directly from Thermo Fisher Scientific are restored to meet original manufacturer specifications and include:
Supplies are limited, so don’t miss this chance to enhance your lab's capabilities. Start the conversation with a sales representative today!
For Research Use Only. Not for use in diagnostic procedures.