artistic composition of a software screen placed next to an image of stained cultured cells placed next to a server tower  
Seamless and scalable
Thermo Scientific™ Store™ Image and Database Management Software is a SQL™ or Oracle™ relational database used for storing and managing the images and data automatically transferred from an HCS instrument or imported from another imaging source. It allows for rapid, highly scalable, global access to both data and images. Use the Store™ Image and Database Management Software after you have collected data and images.
Request a quote
Features
  • Automatic transfer of images and data—Centralized data storage allowing data and image access from multiple locations
  • Accessible and linked—Data is seamlessly linked to its associated image
  • Visualize—View images and data linked together and easily compare images
  • Export—Built in export formats for software such as Spotfire™, Excel™, and GeneData Screener™ programs
Cell Analysis Software and Informatics Products Brochure
Workflow diagram showing how data moves from an HCS Admininstrator to an HCS Application Server to the Store Database. Through this workflow, new plate images and plate data are are accepted into the system, inserted into the database, and stored.
Be productive from your first day of use with Store™ Express Image and Database Management Software
Every Thermo Scientific™ High Content Platform is configured with a base version of Thermo Scientific™ Store™ Express Image and Database Management Software. With 10 GB of storage, Store™ Express Image and Database Management Software helps ensure the security of your data and allows you to be productive from your first day of use. Although it is not a scalable solution, you always have the option to upgrade to the full solution, Thermo Scientific™ Store™ Image and Database Management Software as your requirements change.
  • Store™ Express Image and Database Software included with all high-content platforms
  • Sufficient for moderate use with active data management program
  • Out-of-the-box use with no IT overhead
  • Fully functional with no capabilities hit
Scalable storage
Store™ Image and Database Management Software’s optional configuration includes:
  • Scalable for use with a SQL or Oracle database to accommodate screens
  • Installed on servers for capacity and management
  • Expert guidance for IT implementation
  • Preferred for industry partners
Expands for higher-throughput screening capabilities
Store™ Image and Database Management Software expands to a server-based platform, allowing huge amounts of images and data to be managed. With Store™ Image and Database Management Software, import, convert and spool images from any platform or client running HCS Studio™ Cell Analysis Software. Share data and images across your organization for a fully collaborative work environment.
Ordering Information
Product Quantity Catalog # 
Thermo Scientific Store™ Informatics Software Standard Edition 1 each S0100015D Request a quote
Thermo Scientific Store™ Informatics Software Academic Edition 1 each S0100015A Request a quote
Thermo Scientific Store™ Informatics Software Enterprise Edition 1 each S0100015 Request a quote