Search
Invitrogen Super Bright polymer dye antibody conjugates offer enhanced flexibility in marker and clone selection, offering more options for violet laser–excitable antibody conjugates and facilitating expansion of your flow cytometry panels.
| Violet (Ex/Em) |
| Super Bright 436 dye Super Bright 436 antibodies (413/431 nm) |
| Super Bright 600 dye Super Bright 600 antibodies (414/601 nm) |
| Super Bright 645 dye Super Bright 645 antibodies (414/645 nm) |
| Super Bright 702 dye Super Bright 702 antibodies (413/702 nm) |
| Super Bright 780 dye Super Bright 780 antibodies (413/780 nm) |
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and is used under license.
For Research Use Only. Not for use in diagnostic procedures.