Search
Primary XPS region: Ta4f
Overlapping regions: O2s
Binding energies of common chemical states:
| Chemical state | Binding energy Ta4f7/2 |
|---|---|
| Ta metal | 21.8 eV |
| TaN | 23.0 eV |
| Ta2O5 | 26.2 eV |
N/A
Symbol: Ta
Date of discovery: 1802
Name origin: King Tantalus (Greek mythology)
Appearance: gray-blue
Discoverer: Anders Ekeberg
Obtained from: columbite-tantalite
Melting point: 3290 K
Boiling point: 5731 K
Density[kg/m3]: 16650
Molar volume: 10.85 × 10-6 m3/mol
Protons/Electrons: 73
Neutrons: 108
Shell structure: 2,8,18,32,11,2
Electron configuration: [Xe]4f145d36s2
Oxidation state: 5,4,3
Crystal structure: cubic
This high-melting point transition metal was separated from niobium in 1802 by A. Ekeberg. When pure, tantalum can be drawn into a fine wire for use as a filament for evaporating metals. Due to its high melting point, ductility, and corrosion resistance, tantalum is often incorporated into a variety of metal alloys for use in chemical process equipment, capacitors and aircraft parts. The metal alloy tantalum carbide graphite is one of the hardest metals known, with a melting point of 3738°C. Tantalum is a non-irritating metal and is immune to body liquids, making it well-suited for use in surgical applications.
Products