Femtosecond laser-assisted FIB-SEM for large-volume material removal
The Thermo Scientific Helios 5 Laser System integrates an ultrafast femtosecond laser with FIB and SEM technology, enabling material removal up to 15,000× faster than FIB alone. By combining laser ablation, plasma or Ga FIB, and electron imaging in one chamber, with SEM coincidence at the sample region of interest, this advanced configuration enables exceptional throughput for large-volume material processing and subsurface analysis.
Available on the Helios 5, Helios Hydra, and Helios PFIB platforms, the laser option provides a powerful tool for laboratories that need both speed and nanoscale precision, allowing researchers to move seamlessly from bulk material removal to fine FIB polishing and atomic-resolution S/TEM analysis.
Extreme speed
Millimeter-scale volumes in minutes with femtosecond laser.
Integrated workflow
Laser, FIB, and SEM share one coincident point.
Big + small in one tool
Rapid laser cuts followed by nanometer FIB polishing.
Ready-to-analyze surfaces
Clean laser-ablated surfaces suitable for SEM/EBSD.
Flexible platform option
Configurable on Helios 5, Helios Hydra, or Helios PFIB systems.
Accelerated insights
Shortens failure analysis and 3D tomography cycles.
For Research Use Only. Not for use in diagnostic procedures.