Search
Streamline your Luminex workflow and increase efficiency. Generate consistent and high-quality data using our assays designed for 384-well high-throughput format for both protein and gene expression studies. Leverage our lab automation solutions, whether automating a single step or your entire workflow for Invitrogen ProcartaPlex assays or QuantiGene Plex assays. The combination of multiplexing assays and high throughput maximizes data per sample and number of samples processed, potentially saving weeks compared to single plex and lower throughput assays.
Invitrogen ProcartaPlex and QuantiGene Plex assays offer the distinct ability of a multi-omics high-throughput approach by utilizing the Luminex platform. Genomic and proteomic workflows can be combined without compromising data interpretation or sensitivity. Investigate cell functions and responses by simultaneously interrogating large sets of RNA or protein targets in single samples in the 384-well format.
Examination of both RNA and protein expression variations (see Workflow and Sample data, below) can provide a robust and accurate indication of cell states. RNA levels can be transient and may not correlate with protein levels, which are likely to be maintained for longer periods of time to maintain cellular functions. Due to differences in timing and expression levels, one assay may detect changes that the other does not. Alternatively, the two data sets can further support key findings. The ability to perform both RNA and protein measurements using simple, unified high-throughput workflows can make the Invitrogen ProcartaPlex and QuantiGene Plex assays effective screening tools for large-scale screening studies that benefit from automation workflows.
In a typical workflow, cells are treated with various stimulants or compounds. Cell lysates (for RNA, membrane and intracellular proteins) or cell supernatants (for cytokines or other secreted proteins) are collected. Samples are interrogated with kits corresponding to the targets of interest. Results are generated on a Luminex Instrument.
Reduce time at the bench by ~80% by automating the plate preparation and pipetting of Invitrogen ProcartaPlex multiplex immunoassays to only 45 minutes with the Thermo Scientific KingFisher Apex instrument. KingFisher instruments provide easy-to-follow protocols that remove manual steps and reduce overall processing time and errors associated with sample preparation and handling while increasing the reproducibility of your results compared to manual workflows (Figure 2 and Figure 3).
The Thermo Scientific inSPIRE Collaborative Automation Platform offers a configurable and intuitive interface for the high-throughput ProcartaPlex and QuantiGene Plex assays. Each step of the workflow for these assays is automated by various instruments that are displayed here on the inSPIRE platform. The Momentum Scheduling software integrates all steps and allows for automation of the manual protocol to exceptional detail, like incubation temperature monitoring and control, protocol specified sample handling and well level sample tracking.
Figure 1. Total protein quantitation (bar graphs) and normalized gene expression analysis (line graphs) showing an excellent correlation between gene expression and protein translation for representative targets, MCP-1/CCL2 and IL-8/CXCL8, for the stimulants, LPS and PMA.
Figure 2. High degree of consistency between manual and KingFisher Apex system prepared plates were observed, indicated by the great correlation of standard curve values with R2 value >0.9.
Figure 3. Cytokine production and concentrations were measured during target cell killing of functional CAR-T cells with the ProcartaPlex™ Human Th1/Th2/Th9/Th17 Cytokine Panel, 18plex. Quantitation of target cytokines using the KingFisher Apex instrument showed increased sensitivity compared to manual operation.
For Research Use Only. Not for use in diagnostic procedures.