Search
Central to our Thermo Scientific Vanquish Analytical Purification LC Systems is the fully integrated, high-performance Thermo Scientific Vanquish Fraction Collector (VFC), which allows you to achieve pinpoint purification by fractionation with precision. The Vanquish Analytical Purification LC Systems support a broad range of applications for both upstream (e.g., raw materials and synthetic products) and downstream processes (e.g., protein purification, oligonucleotide synthesis). They match the separation power of the LC systems with compelling fractionation for high-purity and high-yield compound collection. Built with Thermo Scientific Vanquish HPLC and UHPLC system components, they deliver unrivaled performance, robustness, and user-friendliness.
Confidently collect fractions with the highest purity. Choose from a variety of options that fit your application needs, including versatile fractionation modes and customizable needle heights to achieve high recovery and low carryover. Deliver the ultimate in precision across your whole analytical workflow.
The combined power of having a modern LC system with an integrated fraction collector and automated workflows, like delay volume determination, allows you to optimize the fractionation process. Fast fraction collection valve switching and exactly measured delay volume preserve the separation quality by minimized delay times for the fractionation step after analyte detection.
The modularity of our Vanquish LC systems makes it easy to build a system to best suit your application requirements. Upgrade a standard HPLC or UHPLC instrument to a Vanquish Analytical Purification LC System by adding an integrated fraction collector that facilitates precise isolation of your peaks of interest at a wide range of flow rates while applying time- or peak-based fractionation triggers. Choose from a variety of sample containers including vials as well as shallow and deep well plates, to help meet your application requirements.
Choose an LC system based on your needs. Select the Vanquish Core HPLC System for high flow rates. For low analytical flow rates, and if biocompatibility is a must, choose either the Vanquish Flex or Vanquish Horizon UHPLC System. The optimized delay volumes are essential for transferring the high peak capacity of a UHPLC separation into high-resolution fractionation. Our Vanquish Analytical Purification LC System comes with Thermo Scientific Viper Fingertight Fitting systems, which support near-zero dead volume connections and enable sharper peaks and tool-free setup.
Our Vanquish Analytical Purification Systems protect the sample integrity through precise control of fractionation needle movement and minimizing contamination through the use of different wash, flush and rinse options. Features like barcode reading and enclosed sample storage under temperature and light controlled conditions preserve the integrity of the fractionated samples until your are ready for the next step.
The Vanquish Analytical Purification LC System seamlessly integrates with our Thermo Scientific Chromeleon Chromatography Data System (CDS) to deliver superior compliance and data integrity, networking capabilities, automation features like qualification processed, instrument wizards, and fractionation simulation as well as data processing.
The typical flow path of the Vanquish Analytical Purification LC System with integrated Vanquish Fraction Collector. System can be a one- or two-stack configuration.
The specification table below provides an overview of the Vanquish Analytical Purification LC Systems for a standard setup. Not all specifications are listed. Your specifications may differ based on the individual components of your chosen system. Refer to the specification sheets for the system components in the Component specifications, below.
| Category | Vanquish Analytical Purification LC Systems |
| System pressure, maximum | 700, 1000 or 1500 bar, depends on system configuration |
| Flow range | 0-5, 0-8 or 0-10 mL/min, depends on pumps selected |
| Pump options (default delay volume) | Binary (35, 200 or 400 µL) Quaternary (679 µL) Dual (679 µL) Isocratic Delay volume depends on pump(s) selected |
| Biocompatible | Yes, depends on system configuration |
| pH range | 2-12, chloride concentration up to 1 mol/L or 1-13 (chloride concentration up to 0.1 mol/L), depends on system configuration |
| System design | Modular |
| Touchscreen user interface display compatible (optional) | Yes, depends on system configuration |
| Solvent monitoring system compatible (optional) | Yes, depends on system configuration |
| Customizable gradient delay volume to simplify method transfer | Yes, depends on system configuration |
| SmartInject technology for maximum injection precision and improved column lifetime | Yes |
| Injection volume range | Standard: 0.01-25 µL (Vanquish Flex/Vanquish Horizon) or 0.01-100 µL (Vanquish Core) Optional: 0.01-100 µL or up to 1000 µL with Multidraw (Vanquish Flex/Vanquish Horizon); 0.01-250 µL or up to 1000 µL with Multidraw (Vanquish Core) Depends on sampler selected |
| Sample cooling | Yes, 4-40 °C Depends on sampler selected |
| Sample capacity (standard) Based on standard 12 mm OD vials (≤ 1.5 mL) |
4 racks of 54 vials (≤ 1.5mL) each, other rack formats available + capacity of 12 x 22.5 mm OD vials (≤ 10 mL) in the carousel |
| Sampler extension available | Yes, with Vanquish Charger |
| Column compartment thermostatting | Yes, 5-120 °C, depends on column compartment selected Still air and forced air |
| Column compartment pre-heater | Active and passive Depends on system configuration |
| Column compartment post-column cooler | Yes Depends on column compartment selected |
| Detector choices | Vanquish Charged Aerosol Detector Vanquish Diode Array Detector Vanquish Fluorescence Detector Vanquish Multiple Wavelength Detector Vanquish Variable Wavelength Detector Refractive Index Detector Mass Spectrometer |
| Fraction collector | Integral Fraction Collector FT |
| Fraction cooling | Yes, 4-40 °C |
| Fraction vessel capacity |
Any four of the following sample racks or well plates with SBS footprint format:
Example of total fraction vessel capacity:
|
| Fraction collection modes | Triggered by peak signals or time frames; (saw) vertical, (saw) horizontal; interrupt and continuous collection; above or inside vessel collection including septum piercing; manual collection, pooling, wrapping and concatenating |
Standard Vanquish Analytical Purification LC Systems are made up of the following components. Download the associated specifications sheet for more detail on each component.
| Vanquish Horizon Analytical Purification LC System | Vanquish Flex Analytical Purification LC System | Vanquish Core Analytical Purification LC System | |
Pump
|
Vanquish Binary Pump H |
or or |
or |
Autosampler
|
or |
or |
or |
Extended sample capacity (optional)
|
Vanquish Charger (optional) |
||
Column compartment
|
Up to 3 Vanquish Column Compartment H or Up to 3 Vanquish Column Compartment C |
||
| Detectors | Visit our HPLC and UHPLC Detectors page | ||
Fraction collector
|
Vanquish Integral Fraction Collector FT |
||
For Research Use Only. Not for use in diagnostic procedures.