Search
Gene expression analysis is a real-time PCR application that plays a critical role in various SARS-CoV-2 research areas, from basic research to future vaccine and therapeutic development. Identifying key gene expression pathways and networks are crucial to understanding coronavirus pathogenesis, tropism, and transmission.
Gene expression analysis requires high-quality RNA for real-time RT-PCR. Select the RNA isolation method that is best suited to your sample type and throughput needs.
Learn more ›
Our Applied Biosystem TaqMan Gene Expression Assays consist of target primers and an optimized sequence-specific probe (with no extra design, optimization, or melt-curve analysis needed). Explore predesigned assays for the curated list of genes associated with SARS-CoV-2 viral and host target research below, or search our database of more than 2.3 million gene expression assays for additional targets of interest. TaqMan Assays are available in four format options to meet your experiment through-put needs: single-tube, 96-well plates (0.1 and 0.2 mL), TaqMan Array Cards or OpenArray plates. For simultaneous interrogation of multiple genes related to in SARS-CoV-2 infection and pathogenesis, we also offer curated panels of assays to streamline your workflow.
For virus detection and quantitation, we currently offer three predesigned gene expression assays for SARS-CoV-2 viral gene targets.
| Viral gene target | Assay ID |
|---|---|
| SARS-CoV-2N (N gene) | vi07918637_s1 |
| SARS-CoV-2S (S gene) | vi07918636_s1 |
| Orf1ab gene | APH6D4Y |
The first two assays listed are also available as part of the following inventoried panels:
| Panel | Description |
|---|---|
| TrueMark Respiratory Panel 2.0, OpenArray plate | Characterization of 32 respiratory tract viral and bacterial microbes |
| TrueMark Respiratory Panel 2.0, TaqMan Array card | Characterization of 41 respiratory tract viral, bacterial, and fungal microbes |
To study differential gene expression of host targets, our predesigned TaqMan Gene Expression assay portfolio includes over 2.3 million pre-designed primer/probe sets covering more than 30 species.
A selection of some the most-cited genes related to SARS-CoV-2 research are listed in the table below. Many of these genes are also available in our curated panels for simultaneous analysis of multiple host genes implicated in SARS-CoV-2 infection.
| Viral entry factors | Restriction factors | Cytokines | Pro-inflammatory & cytotoxic mediators | Chemokines & receptors | Additional targets |
|---|---|---|---|---|---|
| ACE2 (232) | BST2 (36) | IL1B (77) | GNLY (21) | CCL2 (20) | HGF (217) |
| ANPEP (220) | CH25H (18) | IL2 (51) | GZMA (51) | CCL3 (12) | IFIT1 (13) |
| BSG (85) | CLEC4D (44) | IL4 (55) | GZMB (18) | CCL5 (36) | IFNGR1 (91) |
| CLEC4G (78) | DNAJC6 (220) | IL5 (46) | GZMK (43) | CCL11 (23) | IFNGR2 (61) |
| CTSB (118) | ELF1 (118) | IL6 (58) | CXCL3 (11) | CCL20 (44) | IFT43 (133) |
| CTSL (50) | IFIT3 (15) | IL7 (66) | CXCL9 (38) | CCL27 (29) | ISG15 (15) |
| DPP4 (340) | IFITM1 (11) | IL8 (53) / CXCL8 (40) | IFNB1 (16) | CCR1 (21) | KRT4 (63) |
| FURIN (173) | IFITM2 (5) | IL10 (66) | IFNG (48) | CCR5 (22) | KRT7 (57) |
| NRP1 (227) | IFITM3 (11) | IL12 (186) | KLRB1 (27) | CXCL1 (17) | SCGF (8) |
| TMPRSS11A (51) | LY6E (28) | IL13 (45) | KLRC1 (31) | CXCL3(11) | STAT1 (292) |
| TMPRSS11B (47) | REC8 (218) | IL16 (164) | KLRC2 (10) | CXCL9 (38) | VEGF (166 |
| TMPRSS2 (124) | ZBP1 (76) | IL18 (39) | KLRD1 (44) | CXCL10 (41) | |
| TMPRSS4 (136) | ZC3HAV1 (149) | m-CSF (63) | CXCL12 (48) | ||
| MIF (26) | |||||
| PRF1 (21) | |||||
| TGFB1 (83) | |||||
| TGFB2 (86) | |||||
| TNF (58) | |||||
| TNFSF10 (63) |
Don’t see your gene of interest? Search for other predesigned assays by gene or assay ID ›
TaqMan flexible arrays include a curated and scientifically selected list of gene expression assays from the most-cited genes related to specific signaling pathways or disease states. Panels are available in multiple configurations and species, and are modifiable to easily add or delete assays/genes of interest, or to change your desired layout.
We currently offer three TaqMan® flexible content panels specific to coronavirus research:
Panel | Description | Genes | Host species |
|---|---|---|---|
Coronavirus Entry Factor | 13 of the most-cited host factors that SARS-CoV-2 and other coronaviruses hijack to infect a cell | ACE2, TMPRSS2, TMPRSS4, TMPRSS11A, TMPRSS11B, BSG, ANPEP, CLEC4G, FURIN, CTSL, CTSB, DPP4, and NRP1 | Human Mouse Rat |
Coronavirus Restriction Factors | 13 of the most-cited antiviral host factors that are known to inhibit various stages of the coronavirus life cycle | LY6E, IFITM1, IFITM2, IFITM3, ZAP / ZC3HAV1, BST2, CLEC4D, ELF1, REC8, IFIT3, DNAJC6, ZBP1, CH25H | Human Mouse Rat |
Coronavirus Immune Signaling | 29 cytokines, chemokines, and growth factors involved in the immune response to SARS-CoV-2 and other coronaviruses | IL1B, IL2, IL4, IL5, IL6, IL7, IL10, IL12, IL13, IL16, IL18, IFNB1, IFNg, TNF, TGFB1, M-CSF/CSF1R, MIF, VEGF, SCGF, HGF, TRAIL/TNFSF10, CCL2, CCL3 / MIP1a, CCL5, CCL11, CCL27, CXCL1, CXCL10, CXCL12 | Human Mouse Rat |
In addition to Coronavirus flexible content panels, we offer various inventoried TaqMan panels in both card and plate formats for gene expression analysis of immune signaling pathways.
Learn more ›
You can also browse our comprehensive database for other applicable flexible panels.
Learn more ›
If you are unble to find a predesigned TaqMan Assay for your target of interest, we have two options that enable customization while retaining the quality, reproducibility, and gold-standard chemistry you have come to expect from TaqMan reagents.
For gene expression analysis, we recommend the following assays as positive controls to monitor RNA purification efficacy and ensure the absence of real-time PCR inhibitors
| Positive control | Assay ID |
|---|---|
| Xeno Control | Ac00010014_a1 |
| Human RNase P Control | Hs04930436_g1 |
The master mix used for gene expression analysis depends on whether you want to perform 1-step real-time RT-PCR or 2-step real-time RT-PCR. A 1-step workflow reduces contamination and speeds up overall RT-PCR run time, whereas a 2-step workflow allows you to save your cDNA. If you perform 2-step real-time RT-PCR, you will also need to buy an additional master mix for the reverse-transcription reaction.
Learn more about 1-step and 2-step real-time RT-PCR ›
Learn more about reverse transcription products for 2-step real-time PCR ›
We recommend the following master mixes for most gene expression analysis needs. But you should also review the documentation provided with your assay for any specific workflow requirements.
| Basic research | Translational research* | |
|---|---|---|
| 1-step RT-PCR | TaqMan Fast Virus 1-Step Master Mix | TaqPath 1-Step Multiplex Master Mix or TaqPath 1-Step RT-qPCR Master Mix |
| 2-step RT-PCR | SuperScript IV Reverse Transcriptase + TaqMan Fast Advanced Master Mix | SuperScript IV Reverse Transcriptase + TaqPath qPCR Master Mix, CG |
Try our best performing TaqMan Fast Advanced Master Mix. Request a sample ›
Request a sample for translational research ›
*General purpose reagents manufactured in an ISO13485 facility and are labelled for laboratory use.
We offer real-time PCR instruments for all research needs, from simplicity on a budget to high throughput with maximum productivity.
Learn more ›
Once you order your assays, we provide the protocols and documentation specific to your assay format for download.
Learn more ›
For data analysis, we recommend the Quantstudio Design & Analysis (DA2) app on Connect, our cloud-based platform.
Gene expression analysis requires high-quality RNA for real-time RT-PCR. Select the RNA isolation method that is best suited to your sample type and throughput needs.
Learn more ›
Our Applied Biosystem TaqMan Gene Expression Assays consist of target primers and an optimized sequence-specific probe (with no extra design, optimization, or melt-curve analysis needed). Explore predesigned assays for the curated list of genes associated with SARS-CoV-2 viral and host target research below, or search our database of more than 2.3 million gene expression assays for additional targets of interest. TaqMan Assays are available in four format options to meet your experiment through-put needs: single-tube, 96-well plates (0.1 and 0.2 mL), TaqMan Array Cards or OpenArray plates. For simultaneous interrogation of multiple genes related to in SARS-CoV-2 infection and pathogenesis, we also offer curated panels of assays to streamline your workflow.
For virus detection and quantitation, we currently offer three predesigned gene expression assays for SARS-CoV-2 viral gene targets.
| Viral gene target | Assay ID |
|---|---|
| SARS-CoV-2N (N gene) | vi07918637_s1 |
| SARS-CoV-2S (S gene) | vi07918636_s1 |
| Orf1ab gene | APH6D4Y |
The first two assays listed are also available as part of the following inventoried panels:
| Panel | Description |
|---|---|
| TrueMark Respiratory Panel 2.0, OpenArray plate | Characterization of 32 respiratory tract viral and bacterial microbes |
| TrueMark Respiratory Panel 2.0, TaqMan Array card | Characterization of 41 respiratory tract viral, bacterial, and fungal microbes |
To study differential gene expression of host targets, our predesigned TaqMan Gene Expression assay portfolio includes over 2.3 million pre-designed primer/probe sets covering more than 30 species.
A selection of some the most-cited genes related to SARS-CoV-2 research are listed in the table below. Many of these genes are also available in our curated panels for simultaneous analysis of multiple host genes implicated in SARS-CoV-2 infection.
| Viral entry factors | Restriction factors | Cytokines | Pro-inflammatory & cytotoxic mediators | Chemokines & receptors | Additional targets |
|---|---|---|---|---|---|
| ACE2 (232) | BST2 (36) | IL1B (77) | GNLY (21) | CCL2 (20) | HGF (217) |
| ANPEP (220) | CH25H (18) | IL2 (51) | GZMA (51) | CCL3 (12) | IFIT1 (13) |
| BSG (85) | CLEC4D (44) | IL4 (55) | GZMB (18) | CCL5 (36) | IFNGR1 (91) |
| CLEC4G (78) | DNAJC6 (220) | IL5 (46) | GZMK (43) | CCL11 (23) | IFNGR2 (61) |
| CTSB (118) | ELF1 (118) | IL6 (58) | CXCL3 (11) | CCL20 (44) | IFT43 (133) |
| CTSL (50) | IFIT3 (15) | IL7 (66) | CXCL9 (38) | CCL27 (29) | ISG15 (15) |
| DPP4 (340) | IFITM1 (11) | IL8 (53) / CXCL8 (40) | IFNB1 (16) | CCR1 (21) | KRT4 (63) |
| FURIN (173) | IFITM2 (5) | IL10 (66) | IFNG (48) | CCR5 (22) | KRT7 (57) |
| NRP1 (227) | IFITM3 (11) | IL12 (186) | KLRB1 (27) | CXCL1 (17) | SCGF (8) |
| TMPRSS11A (51) | LY6E (28) | IL13 (45) | KLRC1 (31) | CXCL3(11) | STAT1 (292) |
| TMPRSS11B (47) | REC8 (218) | IL16 (164) | KLRC2 (10) | CXCL9 (38) | VEGF (166 |
| TMPRSS2 (124) | ZBP1 (76) | IL18 (39) | KLRD1 (44) | CXCL10 (41) | |
| TMPRSS4 (136) | ZC3HAV1 (149) | m-CSF (63) | CXCL12 (48) | ||
| MIF (26) | |||||
| PRF1 (21) | |||||
| TGFB1 (83) | |||||
| TGFB2 (86) | |||||
| TNF (58) | |||||
| TNFSF10 (63) |
Don’t see your gene of interest? Search for other predesigned assays by gene or assay ID ›
TaqMan flexible arrays include a curated and scientifically selected list of gene expression assays from the most-cited genes related to specific signaling pathways or disease states. Panels are available in multiple configurations and species, and are modifiable to easily add or delete assays/genes of interest, or to change your desired layout.
We currently offer three TaqMan® flexible content panels specific to coronavirus research:
Panel | Description | Genes | Host species |
|---|---|---|---|
Coronavirus Entry Factor | 13 of the most-cited host factors that SARS-CoV-2 and other coronaviruses hijack to infect a cell | ACE2, TMPRSS2, TMPRSS4, TMPRSS11A, TMPRSS11B, BSG, ANPEP, CLEC4G, FURIN, CTSL, CTSB, DPP4, and NRP1 | Human Mouse Rat |
Coronavirus Restriction Factors | 13 of the most-cited antiviral host factors that are known to inhibit various stages of the coronavirus life cycle | LY6E, IFITM1, IFITM2, IFITM3, ZAP / ZC3HAV1, BST2, CLEC4D, ELF1, REC8, IFIT3, DNAJC6, ZBP1, CH25H | Human Mouse Rat |
Coronavirus Immune Signaling | 29 cytokines, chemokines, and growth factors involved in the immune response to SARS-CoV-2 and other coronaviruses | IL1B, IL2, IL4, IL5, IL6, IL7, IL10, IL12, IL13, IL16, IL18, IFNB1, IFNg, TNF, TGFB1, M-CSF/CSF1R, MIF, VEGF, SCGF, HGF, TRAIL/TNFSF10, CCL2, CCL3 / MIP1a, CCL5, CCL11, CCL27, CXCL1, CXCL10, CXCL12 | Human Mouse Rat |
In addition to Coronavirus flexible content panels, we offer various inventoried TaqMan panels in both card and plate formats for gene expression analysis of immune signaling pathways.
Learn more ›
You can also browse our comprehensive database for other applicable flexible panels.
Learn more ›
If you are unble to find a predesigned TaqMan Assay for your target of interest, we have two options that enable customization while retaining the quality, reproducibility, and gold-standard chemistry you have come to expect from TaqMan reagents.
For gene expression analysis, we recommend the following assays as positive controls to monitor RNA purification efficacy and ensure the absence of real-time PCR inhibitors
| Positive control | Assay ID |
|---|---|
| Xeno Control | Ac00010014_a1 |
| Human RNase P Control | Hs04930436_g1 |
The master mix used for gene expression analysis depends on whether you want to perform 1-step real-time RT-PCR or 2-step real-time RT-PCR. A 1-step workflow reduces contamination and speeds up overall RT-PCR run time, whereas a 2-step workflow allows you to save your cDNA. If you perform 2-step real-time RT-PCR, you will also need to buy an additional master mix for the reverse-transcription reaction.
Learn more about 1-step and 2-step real-time RT-PCR ›
Learn more about reverse transcription products for 2-step real-time PCR ›
We recommend the following master mixes for most gene expression analysis needs. But you should also review the documentation provided with your assay for any specific workflow requirements.
| Basic research | Translational research* | |
|---|---|---|
| 1-step RT-PCR | TaqMan Fast Virus 1-Step Master Mix | TaqPath 1-Step Multiplex Master Mix or TaqPath 1-Step RT-qPCR Master Mix |
| 2-step RT-PCR | SuperScript IV Reverse Transcriptase + TaqMan Fast Advanced Master Mix | SuperScript IV Reverse Transcriptase + TaqPath qPCR Master Mix, CG |
Try our best performing TaqMan Fast Advanced Master Mix. Request a sample ›
Request a sample for translational research ›
*General purpose reagents manufactured in an ISO13485 facility and are labelled for laboratory use.
We offer real-time PCR instruments for all research needs, from simplicity on a budget to high throughput with maximum productivity.
Learn more ›
Once you order your assays, we provide the protocols and documentation specific to your assay format for download.
Learn more ›
For data analysis, we recommend the Quantstudio Design & Analysis (DA2) app on Connect, our cloud-based platform.
For Research Use Only. Not for use in diagnostic procedures.