Anti-Clogging Technology of the Attune NxT Flow Cytometer
Protect your precious samples
The Attune NxT is designed for improved sample processing and on-time project runs, and greater instrument endurance. Expand flow cytometry use to include previously troublesome samples including larger cells and dissociated tissue masses.
The technology behind less clogging includes:
- Syringe-based system for controlled sample volume and pressure entry at 75 PSI (compared to a max pressure of
15 PSI for traditional flow cytometers) - Larger flow cell: 200 µm in diameter
- Rapid acquisition speeds even with diluted samples, reducing the likelihood of cell-cell interactions that can clog the system
Invitrogen Attune NxT Flow Cytometer Demo Video
Designed for faster experimental run times, the Invitrogen Attune NxT compact flow cytometer offers rapid and accurate analysis for a broad range of sample types. Experience the difference.
Is Clogging Getting in the Way of Pizza?!
Scientists racing to get pizza before it is done are clogging at doors. Clogging is not a problem with the Invitrogen Attune NxT Flow Cytometer as it is with other cytometers that leads to sample loss, project delays, and the need to monitor.
Clogging is good in Holland, not in Labs. The Attune NxT Flow Cytometer
Researchers frustrated by clog dancing in the lab. Clogging is great in Holland, not in the lab. The Attune NxT Flow Cytometer.
Flow Cytometry - Attune NxT Non-clogging flow cytometer - Rube Goldberg
Traditionally, flow cytometers clog with large clumpy samples. This leads to sample loss, project delays, and the need to monitor. Invitrogen Attune NxT flow cytometer is designed to reduce the risk of clogging and protecting your precious samples.
Principles behind clog resistant technology
Volumetric sample entry works like a pipette. Tension is created with a plunger pushing down (Figure 1A). The tension is released with the plunger coming up with the sample (Figure 1B). Clogging is less likely to occur under this high and controlled pressure environment.
Figure 1. The syringe-based system for sample introduction is one of anti-clogging design features of the Attune NxT Flow Cytometer.
Educational resources
The Attune NxT Flow Cytometer is designed to minimize the hassle, frustration and sample loss associated with a clogged instrument and bring high-throughput statistical analysis to an increasing number of sample types. Investigate difficult samples and large, clumpy cells including cancer cells, cardiomyocytes and adipocytes, previously not amenable to flow cytometry.
See examples of different cell types analyzed on the Attune NxT Flow Cytometer below (Table 1).
Table 1. Cell types successfully analyzed on the Attune NxT Flow Cytometer
Sample type | Demonstrated example | Published References |
---|---|---|
Algae | Nannochloropsis oculata | |
Bacteria | Escherichia coli | |
Bacillus subtilis | ||
Legionella strains | ||
Staphyloccus aureus | ||
Diatom | Phaeodactylum tricornutum | |
Human | Human adenocarcinoma | |
Human bone marrow (PBMCs) | ||
Human bone osteosarcoma cells (U2OS) | ||
Human cardiomyocytes | ||
Human corneal epithelial cells | ||
Human embryonic kidney (293FT) | ||
Human endothelial cells | ||
Human leukocytes | ||
Human mesenchymal stem cell (hMSC) | ||
Human red blood cells (RBC) | ||
Human T cell leukemia (Jurkat) | ||
Human whole blood | ||
Mouse | Mouse splenocytes | |
Mouse T and B regulatory cells | ||
Mouse cardiomyocytes | ||
Mouse bone marrow | ||
Mouse lung tissue | ||
Mouse tumor cells | ||
Mouse skeletal muscle | ||
Parasites | Plasmodium falciparum | |
L. amazonensis, T. brucei and T. cruzi | ||
Trypanosoma cruzi | ||
Picophytoplankton | Prochlorococcus spp; Synechococcus spp |
|
Plants | Arabidopsis thaliana | |
Phalaris arundinacea | ||
Solidago canadensis | ||
Yeast | Candida albicans | |
Saccharomyces cerevisiae | ||
Zebra fish | Zebra fish organs |
Customer stories
See how Dr. Bruno Sainz from Madrid Autónoma University describes his no-clog experience. Learn the benefits of Attune technology to analyze tumor masses.
Invitrogen Attune NxT Flow Cytometer Demo Video
Designed for faster experimental run times, the Invitrogen Attune NxT compact flow cytometer offers rapid and accurate analysis for a broad range of sample types. Experience the difference.
Is Clogging Getting in the Way of Pizza?!
Scientists racing to get pizza before it is done are clogging at doors. Clogging is not a problem with the Invitrogen Attune NxT Flow Cytometer as it is with other cytometers that leads to sample loss, project delays, and the need to monitor.
Clogging is good in Holland, not in Labs. The Attune NxT Flow Cytometer
Researchers frustrated by clog dancing in the lab. Clogging is great in Holland, not in the lab. The Attune NxT Flow Cytometer.
Flow Cytometry - Attune NxT Non-clogging flow cytometer - Rube Goldberg
Traditionally, flow cytometers clog with large clumpy samples. This leads to sample loss, project delays, and the need to monitor. Invitrogen Attune NxT flow cytometer is designed to reduce the risk of clogging and protecting your precious samples.
Principles behind clog resistant technology
Volumetric sample entry works like a pipette. Tension is created with a plunger pushing down (Figure 1A). The tension is released with the plunger coming up with the sample (Figure 1B). Clogging is less likely to occur under this high and controlled pressure environment.
Figure 1. The syringe-based system for sample introduction is one of anti-clogging design features of the Attune NxT Flow Cytometer.
Educational resources
The Attune NxT Flow Cytometer is designed to minimize the hassle, frustration and sample loss associated with a clogged instrument and bring high-throughput statistical analysis to an increasing number of sample types. Investigate difficult samples and large, clumpy cells including cancer cells, cardiomyocytes and adipocytes, previously not amenable to flow cytometry.
See examples of different cell types analyzed on the Attune NxT Flow Cytometer below (Table 1).
Table 1. Cell types successfully analyzed on the Attune NxT Flow Cytometer
Sample type | Demonstrated example | Published References |
---|---|---|
Algae | Nannochloropsis oculata | |
Bacteria | Escherichia coli | |
Bacillus subtilis | ||
Legionella strains | ||
Staphyloccus aureus | ||
Diatom | Phaeodactylum tricornutum | |
Human | Human adenocarcinoma | |
Human bone marrow (PBMCs) | ||
Human bone osteosarcoma cells (U2OS) | ||
Human cardiomyocytes | ||
Human corneal epithelial cells | ||
Human embryonic kidney (293FT) | ||
Human endothelial cells | ||
Human leukocytes | ||
Human mesenchymal stem cell (hMSC) | ||
Human red blood cells (RBC) | ||
Human T cell leukemia (Jurkat) | ||
Human whole blood | ||
Mouse | Mouse splenocytes | |
Mouse T and B regulatory cells | ||
Mouse cardiomyocytes | ||
Mouse bone marrow | ||
Mouse lung tissue | ||
Mouse tumor cells | ||
Mouse skeletal muscle | ||
Parasites | Plasmodium falciparum | |
L. amazonensis, T. brucei and T. cruzi | ||
Trypanosoma cruzi | ||
Picophytoplankton | Prochlorococcus spp; Synechococcus spp |
|
Plants | Arabidopsis thaliana | |
Phalaris arundinacea | ||
Solidago canadensis | ||
Yeast | Candida albicans | |
Saccharomyces cerevisiae | ||
Zebra fish | Zebra fish organs |
Customer stories
See how Dr. Bruno Sainz from Madrid Autónoma University describes his no-clog experience. Learn the benefits of Attune technology to analyze tumor masses.
For Research Use Only. Not for use in diagnostic procedures.