Search Thermo Fisher Scientific
Dye-Labeled Phosphoramidites |
Thermo Fisher Scientific offers DyLight fluorescent–labeled phosphoramidites that feature absorption and emission spectra suitable for a broad range of applications, including microscopy and PCR.
These phosphoramidites may be used on all automated DNA synthesizers using standard synthesis procedures
Our expertise includes the development of custom dye chemistries, linkers, and labeling strategies. Fluorescent tags can be incorporated at the 5′ end of an oligonucleotide using DyLight phosphoramidites with standard synthesis and deprotection procedures. In addition, these direct-labeled phosphoramidites can be used in research applications without the constraints of licensing and royalty fees.
For more than 40 years, our OEM and custom capabilities in research, development, and manufacturing of nucleic acids continue to be key resources for our global partners at molecular diagnostic, pharmaceutical, and life science research companies.
DyLight DY547 CEP | DyLight DY647 CEP | DyLight DY677 CEP | |
---|---|---|---|
Cat. No. | SY6332 | SY6334 | SY6336 |
Formula | C38H54IN4O2P | C47H63N4O5PS | C55H67N4O5PS |
Molecular weight | 756.74 g/mol | 827.07 g/mol | 927.18 g/mol |
HPLC purity | ≥ 90% at 547 nm | ≥ 85% at 647 nm | ≥ 85% at 677 nm |
Identity | UV-Vis spectrum conforms to that of the authentic sample | ||
Shipping / Storage condition | Dry ice / -20 °C | ||
Request a quote |
DyLight Fluorescein can be further purified to remove the 5-isomer and leaves ≥95% of the 6-isomer (FAM-6), making it ideal for applications requiring higher sensitivity.
Click images to enlarge
DyLight FAM-6 CEP | DyLight Fluorescein CEP | DyLight HEX CEP | DyLight TET CEP | ||
---|---|---|---|---|---|
Cat. No. | 27-1608 | 27-1606 | SY6342 | SY6341 | |
Phosphoramidite Description | Fluorescein dipivalate phosphoramidite, 6-isomer | [(3′,6′-dipivaloylfluoresceinyl)-5,6-carboamidohexyl]-1-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite | 6-(4,7,2′,4′,5′,7′-Hexachloro-3′,6′-dipivaloylfluoresceinyl-6-carboxamido)-hexyl-1-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite | 5′-Tetrachloro-Fluorescein Phosphoramidite | |
Formula | C46H58N3O10P | C46H58N3O10P | C46H52N3O10Cl6P | C46H54N3O10Cl4P | |
Molecular weight | 843.95g/mol | 843.94g/mol | 1050.63 g/mol | 981.73 g/mol | |
CAS / PubChem# | 204697-37-0 / 58114954 | N/A | 1360547-55-2 | 877049-90-6 | |
HPLC purity | ≥ 90% | ≥ 90% | ≥ 95% | ≥ 95% | |
31P NMR purity | ≥ 95% (6-isomer) | 85% | N/A | N/A | |
Shipping / Storage condition | Dry ice / -20 °C | ||||
Request a quote |
Not finding what you are looking for? We also offer DNA phosphoramidites, RNA phosphoramidites, specialty phosphoramidites, 2′-modifications, linker and spacer phosphoramidites, and succinates and CPG support synthesis supports. Contact us to learn more.