Mr-Frosty-Freezing-Container
Designed for use with the entire portfolio of Thermo Scientific Nunc and Nalgene cryogenic storage tubes, Nalgene™ Mr. Frosty™ freezing containers provide simple cooling of samples at the optimal rate to preserve cells.
Download the Mr. Frosty specification sheet
Features of the Mr. Frosty Freezing Container
Optimal cooling
Provides critical, repeatable -1˚C/ minute cooling rate required for successful cell cryopreservation and recovery.
Easy and safe
Fill to the designated line with 100% isopropyl alcohol and place in your mechanical freezer. Samples never touch the alcohol - no risk of label or printing removal.
Flexibility
Units and inserts available for 1.0, 1.2, 1.8, 2.0, 3.6, 4.5 and 5.0mL tubes, the Mr. Frosty freezing container provides the widest tube compatibility available. Holds 12 or 18 tubes.
Available Thermo Scientific Mr. Frosty Freezing Containers
Cat. #5100-0001
Cat. #5100-0036
Cat. #5100-0050
Tube compatibility1.0 - 2.0mL tubes3.6mL tubes4.5 - 5.0mL tubes
Capacity18 tubes12 tubes12 tubes
Vol. Isopropyl Alcohol (mL)250500500
Height (mm)86151151
Diameter (mm)117117117
 Order nowOrder nowOrder now
Mr. Frosty Freezing Container Protocol Video
Our freezing containers are easy to use—requires only 100% isopropyl alcohol and a mechanical freezer
  • Screw top lid secures samples inside in the event of accidental container tipping
  • No contact with the alcohol: no wicking, no label removal and no compromise of tube hand printing