Ion AmpliSeq Community Panels for Targeted NGS Testing
Custom NGS panels designed with leading researchers to help you get up and running quickly and conveniently
Ion AmpliSeq community panels, designed with input from leading disease researchers and verified for performance, provide targeted next-generation sequencing (NGS) for your research in cancer, inherited disease, and microbial and infectious disease. These pre-designed custom panels are customizable to fit your unique project requirements and advance clinical research, giving you the flexibility to add or remove genes and amplicons.
Ion AmpliSeq Rapid Microbial Identification (RMID) Research Panel
The Ion AmpliSeq Rapid Microbial Identification (RMID) Research Panel is a pre-designed custom targeted NGS panel that is available to order on AmpliSeq.com as a made-to-order community panel product. 
It uses Ion AmpliSeq technology for targeted NGS to research bacterial 16S rRNA, fungal rRNA, and rpoB regions, allowing for pan-bacterial and pan-fungal metagenomic detection, along with species-specific identification of 88 microbial species in a single assay.
Fast and simple NGS workflow with walkaway automation
Get NGS test results in as little as 24 hours with just 10 minutes of hands-on time for library preparation through sequencing
Comprehensive panel design
Simultaneously perform metagenomic and targeted species-specific analysis for bacteria, mycobacteria, and fungi in a single assay powered by Ion AmpliSeq technology
Verified performance
Reliably detect relevant microbial species using a panel designed and tested by community users with real-world clinical
research samples
Customize the panel to meet your needs
Add or remove targets in the panel design to develop the exact NGS test you need for your application
Browse Ion AmpliSeq Community Panels
Cancer
Ion AmpliSeq gene panels for research in breast, colon, and lung tumor
variant detection
Genetic disease
Ion AmpliSeq panels for research in Mendelian diseases
Custom needs
Customize any predesigned panel to meet your unique research needs
Video: OncoNetwork Consortium
Eight leading European researchers worked together with Thermo Fisher Scientific scientists to develop a 22-gene Ion AmpliSeq community panel for lung and colon cancer.
How to order an Ion AmpliSeq community panel
Ion AmpliSeq community panels, designed with input from leading researchers, are available for any lab to order through ampliseq.com.
  1. Visit Ion AmpliSeq™ Designer at ampliseq.com and sign in. If you do not have an account, follow the registration process to create a new account.
  2. After signing in, go to Pre-designed Panels > Community Panels located in the top navigation bar of the portal.
  3. Browse the Community panel designs or search your panel of interest using the search box on the upper left side.
  4. Add your panel to the cart and follow the checkout process.
For Research Use Only. Not for use in diagnostic procedures.
PMR: 001259