Neodymium • Lanthanide Rare Earth
Primary XPS region: Nd3d, Nd4d
Overlapping regions: O KLL
Binding energies of common chemical states:
Chemical stateBinding energy Nd3d5/2
Nd aluminum phosphate~980 eV
Experimental information
  • Collect both Nd3d and Nd4d regions to aid with data interpretation.
Interpretation of XPS spectra
  • There is a very strong overlap of Nd3d with O KLL peaks.
    • Use Nd4d instead (120eV), but this can also overlap with Al2s.
General comments
  • Common oxidation state: Nd(III).
  • Magnetic in metallic state.
About this element
Element Crystal hex
Symbol: Nd
Date of discovery: 1885
Name origin: Greek neos and didymos
Appearance: silver
Discoverer: C.F. Aver von Welsbach
Obtained from: monazite and bastnasite
Melting point: 1297 K
Boiling point: 3373 K
Density[kg/m3]: 6800
Molar volume: 20.59 × 10-6 m3/mol
Protons/Electrons: 60
Neutrons: 84
Shell structure: 2,8,18,22,8,2
Electron configuration: [Xe]4f46s2
Oxidation state: 3
Crystal structure: hexagonal
Neodymium is a rare, reactive, earth metal with a bright, silvery, metallic luster that quickly tarnishes in air. It was discovered in 1885 by Austrian chemist Baron Carl Auer von Welbach, who separated the metal from didymium with repeated fractionations. While the free element is in misch metal, long known and used as a pyrophoric alloy for light flints, the element was not isolated in its pure form until 1925. Neodymium is used to create strong permanent magnets used in loudspeakers and computer data storage devices. It is also used to color glass used in lasers and incandescent light bulbs.
element_neodymium
Products
Nexsa G2 XPS
  • Mikrofokus-Röntgenquellen
  • Einzigartige Optionen mit mehreren Verfahren
  • Dual-Mode-Ionenquelle für monoatomare und Cluster-Ionentiefenprofilierung
Formatvorlage für das Original der Instrumentenkarten
K-Alpha XPS
  • Hochauflösende XPS
  • Schneller, effizienter, automatisierter Arbeitsablauf
  • Ionenquelle für Tiefenprofilierung
ESCALAB Xi+ XPS
  • Hohe spektrale Auflösung
  • Oberflächenanalyse mit mehreren Verfahren
  • Umfangreiche Probenvorbereitungs- und Erweiterungsoptionen
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Formatvorlage für Produkttabellen-Spezifikationen
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Formatvorlage für Produkttabellen-Spezifikationen
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.