Search
Transformation is mainly performed in molecular cloning to maintain and propagate DNA sequences of interest in bacteria or other host cells. The DNA of interest is incorporated into plasmids or other vectors, which act as a vehicle to shuttle DNA of interest into host cells. The transformed colonies are analyzed after transformation to ensure propagation of the DNA insert. Some of the issues observed include absence of transformants, transformants with incorrect inserts or absence of inserts.
This article on bacterial transformation troubleshooting addresses some common problems and recommendations on how to solve them.
After overnight incubation following transformation, either no colonies or very few colonies are observed on the selective LB agar plate.
| Possible causes for very few or no transformants | Recommendations to optimize transformation and colony formation |
|---|---|
| Suboptimal transformation efficiency | To ensure good transformation efficiencies:
|
Suboptimal quality and/or quantity of transforming DNA
|
|
Cloned DNA or protein that is toxic to the cells |
|
Incorrect strain used to propagate a vector carrying a lethal gene for selection |
|
| Insufficient number of cells plated |
|
Suboptimal growth time and temperature |
|
Incorrect antibiotic or concentration in plates |
|
Improper use of cell-spreading tools |
|
After colony selection and analysis, vector contains an incorrect or truncated DNA fragment as identified by sequencing or restriction analysis (fragment size identified in the gel).
| Possible cause for transformants with incorrect or truncated DNA inserts | Recommendations to maximize propagation of correct DNA inserts |
|---|---|
Unstable DNA |
|
| DNA mutation |
|
| Cloned fragment truncated |
|
After colony selection and analysis, vector is empty.
| Possible cause | Recommendation |
|---|---|
| Cloned DNA or protein that is toxic to the cells |
|
| Improper colony selection method |
|
| Issues in upstream cloning steps |
|
Picked colonies do not grow in a selective media or after colony selection no DNA vector could be obtained.
| Possible cause | Recommendation to improve chances of transformation |
|---|---|
| Antibiotic-resistant strain |
|
| Vector plasmid recombined into the host chromosome |
|
| Satellite colonies |
|
| High number of cells plated |
|
| Cell contamination |
|
After overnight incubation, too many transformed E. coli colonies are observed making it difficult to pick single colonies (they might even fuse together). Or colonies are spread unevenly forming isolated clusters.
| Possible causes for poor colony formation | Recommendations to avoid overgrown and clumped colonies |
|---|---|
Large number of cells plated |
|
Long incubation |
|
| Improper spreading |
|
It takes unusually longer to grow cells in liquid media or purified DNA yields are not sufficient.
| Possible causes | Recommendations to optimize cell growth and improve DNA |
|---|---|
Wrong media |
|
Improper growth conditions or old colony |
|
Have any questions on competent cells or transformation? Click on the resources listed below to access overviews, videos, genotype guides, and educational resources.
Review rich educational content on transformation workflow, troubleshooting, strategies, and selection guides.
Stay up-to-date on industry trends in molecular cloning with on-demand webinars.
Access white papers and technical and application notes on molecular cloning.
Watch instructional videos on troubleshooting tips, how-to videos, and explainers related to molecular cloning.