Search
Custom DNA Synthesis for Large & Complex Genes |
Researchers who need long custom DNA constructs for pharmaceutical development, such as viral genomes, or for metabolic engineering, pathway engineering, or synthetic biology can avail our proprietary Invitrogen GeneArt Gene Synthesis services. Our scientists have experience generating longer constructs from 10 kb to several hundred kilobases or containing complex sequence features.
Using our advanced GeneArt technology, even relatively complex sequences can be synthesized and assembled rapidly and reliably. Complex genes and sequences may have challenging features such as complex secondary structures, repetitive sequences (direct or indirect), high (>80%) or low (<20%) GC content (Figure 1), or long polypurine/polypyrimidine runs that cannot be synthesized using our standard process. For these projects, we have developed a unique proprietary technology that employs specially modified and purified oligonucleotides, and more sophisticated gene assembly methods.
In many cases these projects can be completed and shipped with the same turnaround time as our conventional gene synthesis projects.
Figure 1. Example of sequence with very high GC content. The plot shows the GC content in a 40 bp window centered at the indicated nucleotide position. In this example, ~300 bp of the 5’ end of the sequence has a very high GC content.
For details on our Complex Gene Synthesis service, including a price and delivery estimate, upload your sequence for analysis by going to our secure online GeneArt Services Dashboard.
Our high-throughput gene synthesis capacity of >15 Mbp per month allows fast and reliable production of DNA pieces for assembly. Our other capabilities include:
We serve over 3,000 customers on over 12,000 gene constructs each month, so you can be sure we have the global capacity to meet your research needs.
Cloned genes ranging in sizes up to 1,000 bp can be completed in as fast as 5 business days.
| DNA type |
Genes/ORFs
|
|
|
|
|---|---|---|---|---|
| Length | 0.1–1 kb | 1–7 kb | 7 kb–100 kb | 100 kb–1.5 Mb |
| Production time | 5–10 business days | 12–22 business days | From 23 business days | On request |
| Service workflow & capabilities | High-throughput production
| Specialized manufacturing team
| Combined expert services and tools
| |
| How to order | Order online | Contact us | ||
Watch how-to videos and explainers on GeneArt Instant Designer.
Contact us
Request more information
North America and Asia: 1-800-955-6288
Europe: +49 (0)941 942 76-100
Order support:
Geneartsupport@thermofisher.com
Stay up to date on gene synthesis and protein synthesis service offerings.
Read white papers on de novo gene synthesis technologies.
![]()
For Research Use Only. Not for use in diagnostic procedures.