Search
Use your existing CO₂ incubator to investigate suspension culture and scale-up. Save space and achieve reliable around-the-clock operation with CO₂ resistant shakers ideally suited to maximize cell health within your existing CO₂ incubator.
| CO₂ Resistant Shaker 3mm Orbit |
CO₂ Resistant Shaker 19mm Orbit |
|
![]() |
![]() |
|
| Learn more | Learn more | |
| Maximum vessel size | 5L Erlenmeyer Flask | 5L Erlenmeyer Flask |
| Available platforms |
|
|
| Speed | 30 to 1200 rpm | 30 to 300 rpm |
| Environmental temperature range | 4º to 60º C | 4º to 60º C |
| Humidity limits | 20% to 95% | 20% to 95% |
| Exterior dimensions (H x W x D) |
|
|
| Orbit | 118 in. (0.3cm) | .75 in. (1.9) |
| Weight capabity | 22 lbs (10 kg)* | 13 lb. (6 kg) |
| Data output | 4-20 mA | 4-20 mA |
| Accessories | ||
*Maximum load when <150 rpm
Thermo Scientific CO₂ incubators and environmental chambers are ideal to pair with your Thermo Scientific CO₂ resistant shakers for the flexibility to scale up suspension cultures while continuing stationary cultures in one system.
| CO₂ Incubator | Maximum Shakers | Maximum 500mL Flasks | Shelf Dimension | Load Limit per Shelf |
|---|---|---|---|---|
| Forma 310 Direct Heat CO₂ Incubator | 1 | 9 | 470 x 470mm | 16kg slide in and out |
| Forma Steri-Cycle CO₂ Incubator | 1 | 9 | 470 x 470mm | 16kg slide in and out |
| Forma Series II 3110 Water-Jacketed CO₂ Incubator | 1 | 9 | 470 x 470mm | 13.6kg slide in and out |
| Large-Capacity Reach-In CO₂ Incubator | 8 units, 4 on each shelf | 72 | 777 X 655mm | 90kg fixed reinforced shelves* (1900005) |
| Heracell 150i CO₂ Incubators-Stainless | 1 | 9 | 423 x 465mm | 10kg |
| Heracell 150i CO₂ Incubators-Copper | 1 | 9 | 423 x 465mm | 10kg |
| Vios iDx 165 CO₂ Incubator-Stainless | 1 | 9 | 423 x 465mm | 10kg |
| Vios iDx 165 CO₂ Incubator-Copper | 1 | 9 | 423 x 465m | 10kg |
| Heracell 240i CO₂ incubator-Stainless | 1 | 18 | 560 x 500mm | 10kg |
| Heracell 240i CO₂ incubator-Copper | 1 | 18 | 560 x 500mm | 10kg |
| Vios iDx 255 CO₂ Incubator-Stainless | 1 | 18 | 560 x 500mm | 10kg |
| Vios iDx 255 CO₂ Incubator-Copper | 1 | 18 | 560 x 500mm | 10kg |
| CO₂ Incubator | Maximum Shakers | Maximum 500mL Flasks | Shelf Dimension | Load Limit per Shelf |
|---|---|---|---|---|
| Forma 310 Direct Heat CO₂ Incubator | 0 | 0 | 470 x 470mm | 16kg slide in and out |
| Forma Steri-Cycle CO₂ Incubator | 0 | 0 | 470 x 470mm | 16kg slide in and out |
| Forma Series II 3110 Water-Jacketed CO₂ Incubator | 0 | 0 | 470 x 470mm | 13.6kg slide in and out |
| Large-Capacity Reach-In CO₂ Incubator | 8 units, 4 on each shelf | 80 | 777 X 655mm | 90kg fixed reinforced shelves* (1900005) |
| Heracell 150i CO₂ Incubators-Stainless | 0 | 0 | 423 x 465mm | 10kg |
| Heracell 150i CO₂ Incubators-Copper | 0 | 0 | 423 x 465mm | 10kg |
| Vios iDx 165 CO₂ incubator-Stainless | 0 | 0 | 423 x 465mm | 10kg |
| Vios iDx 165 CO₂ incubator-Copper | 0 | 0 | 423 x 465m | 10kg |
| Heracell 240i CO₂ incubator-Stainless | 1 | 10 | 560 x 500mm | 10kg |
| Heracell 240i CO₂ incubator-Copper | 1 | 10 | 560 x 500mm | 10kg |
| Vios iDx 255 CO₂ incubator-Stainless | 1 | 10 | 560 x 500mm | 10kg |
| Vios iDx 255 CO₂ incubator-Copper | 1 | 10 | 560 x 500mm | 10kg |
*Some incubators and environmental chambers will require reinforced shelving to support the weight of a shaker. Please refer to the loading limit per shelf indicated for your incubator or contact your sales representative for more information.
Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.