Search
Life Technologies offers a collection of TaqMan™ Drug Metabolism Genotyping Assays for pharmacogenomics applications. This product provides 2,700 unique assays to detect polymorphisms in 221 genes that code for various drug metabolism enzymes and drug transporters. View the Drug Metabolism Genotyping Assay Index in Excel (908 KB) or Text (905 KB) format.
TaqMan Drug Metabolism Genotyping Assays detect biologically important polymorphisms that reside in complex portions of the genome. These polymorphisms have been associated with certain diseases such as cancer, and have been shown to significantly impact drug efficacy. Where possible all assays have been mapped to the common public allele nomenclature. All TaqMan DME Genotyping Assays have proven performance across 180 unique DNA samples.
| Discrimination | 5' nuclease assay to discriminate between 2 alleles of a specific SNP for use in genotyping studies. |
| Assay Components | Each pre-formulated assay contains:
|
| Reactions | Sufficient for 750 5ul reactions in a 384-well plate |
| Expiration Date | 5 years from the date of manufacturing |
| Documentation | Available to download at thermofisher.com/taqmanfiles :
|
| Assay Information File | Includes sales order number, part number, assay ID, probe and primer concentrations, and Context Sequence. Also includes NCBI dbSNP ID, chromosome location, cytogenic location, gene association, SNP type, and Applied Biosystems® minor allele frequencies, if applicable. The Context Sequence is the nucleotide sequence surrounding the SNP site and is provided in the (+) genome strand orientation relative to the NCBI reference genome. The SNP alleles are included in brackets, where the order of the alleles corresponds to the association with probe reporter dyes, where [Allele 1 = VIC® dye / Allele 2 = FAM™ dye]. |
| Tracking/Identification |
|
| Shipping and Storage | Shipped at ambient temperature. |
Protocol: TaqMan® Drug Metabolism Genotyping Assays
Reference Guide: TaqMan® Drug Metabolism Genotyping Assays
Quick Reference Card: TaqMan® Genotyping Master Mix
Drug Metabolism Genotyping Assay Index: (XLS 908 KB) | (TXT 905 KB)
Poster: Coverage of the PharmaADME Consortium Core Marker List by TaqMan® DME and Copy Number Assays
Application Notes: TaqMan® Drug Metabolism Genotyping Assays for Triallelic SNPs
Bulletin: TaqMan® Drug Metabolism Genotyping Assays
Data Sheet: TaqMan® Drug Metabolism Genotyping Assays
Poster: Annotation of Polymorphisms in Drug Metabolizing Enzymes
How Well do the HapMap SNPs “Tag” Functional Variants in Drug Metabolizing Enzyme Genes?
| Discrimination | 5' nuclease assay to discriminate between 2 alleles of a specific SNP for use in genotyping studies. |
| Assay Components | Each pre-formulated assay contains:
|
| Reactions | Sufficient for 750 5ul reactions in a 384-well plate |
| Expiration Date | 5 years from the date of manufacturing |
| Documentation | Available to download at thermofisher.com/taqmanfiles :
|
| Assay Information File | Includes sales order number, part number, assay ID, probe and primer concentrations, and Context Sequence. Also includes NCBI dbSNP ID, chromosome location, cytogenic location, gene association, SNP type, and Applied Biosystems® minor allele frequencies, if applicable. The Context Sequence is the nucleotide sequence surrounding the SNP site and is provided in the (+) genome strand orientation relative to the NCBI reference genome. The SNP alleles are included in brackets, where the order of the alleles corresponds to the association with probe reporter dyes, where [Allele 1 = VIC® dye / Allele 2 = FAM™ dye]. |
| Tracking/Identification |
|
| Shipping and Storage | Shipped at ambient temperature. |
Protocol: TaqMan® Drug Metabolism Genotyping Assays
Reference Guide: TaqMan® Drug Metabolism Genotyping Assays
Quick Reference Card: TaqMan® Genotyping Master Mix
Drug Metabolism Genotyping Assay Index: (XLS 908 KB) | (TXT 905 KB)
Poster: Coverage of the PharmaADME Consortium Core Marker List by TaqMan® DME and Copy Number Assays
Application Notes: TaqMan® Drug Metabolism Genotyping Assays for Triallelic SNPs
Bulletin: TaqMan® Drug Metabolism Genotyping Assays
Data Sheet: TaqMan® Drug Metabolism Genotyping Assays
Poster: Annotation of Polymorphisms in Drug Metabolizing Enzymes
How Well do the HapMap SNPs “Tag” Functional Variants in Drug Metabolizing Enzyme Genes?
For Research Use Only. Not for use in diagnostic procedures.