Samarium • Lanthanide Rare Earth
Primary XPS region: Sm3d
Overlapping regions: N/A
Binding energies of common chemical states: N/A
Experimental information
N/A
Interpretation of XPS spectra
N/A
General comments
  • Readily changes valency.
About this element
Element Crystal rhom
Symbol: Sm
Date of discovery: 1879
Name origin: Samarskite
Appearance: silver
Discoverer: Paul Emile Lecoq de Boisbaudran
Obtained from: kernite
Melting point: 1345 K
Boiling point: 2173 K
Density[kg/m3]: 7353
Molar volume: 19.98 × 10-6 m3/mol
Protons/Electrons: 62
Neutrons: 88
Shell structure: 2,8,18,24,8,2
Electron configuration: [Xe]6s24f6
Oxidation state: 3
Crystal structure: rhombohedral
In 1853, a Swiss chemist by the name of J. C. Galissard de Marignac spectroscopically discovered samarium in the material dydimia. Very similar to praseodymium, samarium is one of the rare elements used to make carbon arc lights for use in the motion picture industry. Comprising about 1% of Misch metal, the element is also used to make flints for lighters. When combined with cobalt, the elements produce a powerful, permanent magnet with a high resistance to demagnetization. Samarium oxide is added to glass to sensitize the phosphors in infrared radiation and acts as a catalyst for the dehydration and dehydrogenation of ethanol. While samarium has no known biological role, it is purported to stimulate metabolism.
element_samarium
Products
Nexsa XPS
  • Módulo de basculación para mediciones de ARXPS
  • Fuente de iones de modo dual para capacidades de realización de perfiles de profundidad
  • Análisis de aislantes.
Hoja de estilo para tarjetas originales instrumentos
K-Alpha XPS
  • Espectroscopía de área seleccionable.
  • Monocromador microfocalizado.
  • Espectroscopía del estado químico de alta resolución.
ESCALAB Xi+ XPS
  • Espectroscopía de alta sensibilidad.
  • XPS con fuente de rayos X no monocromática.
  • Analizador hemisférico de 180°.
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Servicios de microscopía electrónica para
la ciencia de materiales
Para garantizar un rendimiento óptimo del sistema, le proporcionamos acceso a una red de expertos de primer nivel en servicios de campo, asistencia técnica y piezas de repuesto certificadas.