COL16316-hero
按照您希望的方式开展研究.
QuantStudio™实时荧光定量PCR系统及数字PCR系统
灵活、多功能、快速——每个用户都有着独特的需求,这也是ife Technologies进一步拓展其QuantStudio™系列实时荧光定量PCR系统及数字PCR系统的原因所在。现在您可以挑选最适于自身研究需求,为自己量身定制的qPCR平台。
立即询价
Take the quiz
QuantStudio™
1 System
Entry-level with proven reliability
Learn more
QuantStudio™
3 System
Easy to use, access, and share your results
Learn more
QuantStudio
5 System
Complete set of features to optimize your experience
Learn more
QuantStudio
6 Flex System
Flexibility designed to easily evolve with your research needs
Learn more
QuantStudio
7 Flex System
Move from application to application with ease
Learn more
QuantStudio
12K Flex System
High-throughput for maximum productivity with minimum effort
Learn more
QuantStudio 3D
Digital PCR System
Innovation designed to help you push the limits of PCR
Learn more
  QuantStudio™
3 System
Learn more ›
QuantStudio™
5 System
Learn more ›
QuantStudio™
6 Flex
Learn more ›
QuantStudio™
7 Flex
Learn more ›
QuantStudio™
12 Flex
Learn more ›
QuantStudio™
3D
Learn more ›
Reactions per Run 96 96 or 384 96 or 384 96 or 384 96 or 384 96, 384, or 12,228
Colors Up to 4
QuantStudio 5 System, Color 96-well up to 6 (21 combinations)
384-well up to 5
Up to 5
QuantStudio 7 Flex System, Color
Up to 6 (21 combinations)
QuantStudio 12k Flex System, Color Up to 6 (21 combinations)
2 (endpoint detection)
Available
blocks
96-well
96-well Fast
96-well
96-well Fast
384-well
96-well
96-well Fast
384-well
96-well
96-well Fast
384-well
TaqMan™ Array Card
96-well
96-well Fast
384-well
TaqMan™ Array Card,
OpenArray™ plates
20,000 partitions/chip
Automation
compatible
No No No Yes Yes No
Throughput Medium Medium Medium High Very high Low
Block
change
Fixed Fixed Block change from front in less than 1 minute, no tools required N/A
21CFR
Part 11
enablement
Security Security, auditing, e-signature package included Security, auditing, e-signature package available separately No
Wi-Fi Yes Yes No No No No
Thermo Fisher Cloud direct connection direct connection connect to cloud through computer connect to cloud through computer connect to cloud through computer connect to cloud through computer
Touch
Screen
Yes, interactive Yes, interactive Yes Yes Yes Yes
  Request a quote › Request a quote › Request a quote › Request a quote › Request a quote › Request a quote ›
{{column.ids[1]}}
  • {{value}}
Block Format Type Colors Product Full Specs
{{row["format+0"]}} {{row["type+1"]}} {{row["color+2"]}} {{row.title}} {{row.specs}}
根据您的需求进行配置
您和您的研究有着一样的独特性。无论您有何种需求,总有一款QuantStudio™实时荧光定量PCR系统或数字PCR系统能够满足您。
您当地的销售代表可根据您的需求帮助您量身定制出个性化解决方案,您也可采用我们的在线产品配置工具,轻松地自行配置最合适的QuantStudio®系统。
配置解决方案
Compare QuantStudio Real-Time PCR & Digital PCR Systems
  QuantStudio
3 System
Learn more ›
QuantStudio
5 System
Learn more ›
QuantStudio
6 Flex
Learn more ›
QuantStudio
7 Flex
Learn more ›
QuantStudio
12 Flex
Learn more ›
QuantStudio
3D
Learn more ›
Reactions per Run 96 96 or 384 96 or 384 96 or 384 96 or 384 96, 384, or 12,228
Colors Up to 4 96-well up to 21 
384-well up to 5
Up to 5 Up to 21 Up to 21 2 (endpoint detection)
Available
blocks
96-well
96-well Fast
96-well
96-well Fast
384-well
96-well
96-well Fast
384-well
96-well
96-well Fast
384-well
TaqMan Array Card
96-well
96-well Fast
384-well
TaqMan Array Card,
OpenArray plates
20,000 partitions/chip
Automation
compatible
No No No Yes Yes No
Throughput Medium Medium Medium High Very high Low
Block
change
Fixed Fixed Block change from front in less than 1 minute, no tools required N/A
21CFR
Part 11
enablement
Security Security, auditing, e-signature package included Security, auditing, e-signature package available separately No
Wi-Fi Yes Yes No No No No
Thermo Fisher Cloud direct connection direct connection connect to cloud through computer connect to cloud through computer connect to cloud through computer connect to cloud through computer
Touch
Screen
Yes, interactive Yes, interactive Yes Yes Yes Yes
  Request a quote › Request a quote › Request a quote › Request a quote › Request a quote › Request a quote ›
根据您的需求进行配置
您和您的研究有着一样的独特性。无论您有何种需求,总有一款QuantStudio™实时荧光定量PCR系统或数字PCR系统能够满足您。
您当地的销售代表可根据您的需求帮助您量身定制出个性化解决方案,您也可采用我们的在线产品配置工具,轻松地自行配置最合适的QuantStudio®系统。
配置解决方案
Compare QuantStudio Real-Time PCR & Digital PCR Systems
  QuantStudio
3 System
Learn more ›
QuantStudio
5 System
Learn more ›
QuantStudio
6 Flex
Learn more ›
QuantStudio
7 Flex
Learn more ›
QuantStudio
12 Flex
Learn more ›
QuantStudio
3D
Learn more ›
Reactions per Run 96 96 or 384 96 or 384 96 or 384 96 or 384 96, 384, or 12,228
Colors Up to 4 96-well up to 21 
384-well up to 5
Up to 5 Up to 21 Up to 21 2 (endpoint detection)
Available
blocks
96-well
96-well Fast
96-well
96-well Fast
384-well
96-well
96-well Fast
384-well
96-well
96-well Fast
384-well
TaqMan Array Card
96-well
96-well Fast
384-well
TaqMan Array Card,
OpenArray plates
20,000 partitions/chip
Automation
compatible
No No No Yes Yes No
Throughput Medium Medium Medium High Very high Low
Block
change
Fixed Fixed Block change from front in less than 1 minute, no tools required N/A
21CFR
Part 11
enablement
Security Security, auditing, e-signature package included Security, auditing, e-signature package available separately No
Wi-Fi Yes Yes No No No No
Thermo Fisher Cloud direct connection direct connection connect to cloud through computer connect to cloud through computer connect to cloud through computer connect to cloud through computer
Touch
Screen
Yes, interactive Yes, interactive Yes Yes Yes Yes
  Request a quote › Request a quote › Request a quote › Request a quote › Request a quote › Request a quote ›