Search
Lipopolysaccharide (LPS) serves as a potent method to activate various immune cells, such as B cells, monocytes, and macrophages, and other cells known to be responsive to LPS stimulation. By providing an immunogenic trigger, such as LPS, immune cells can initiate the production of crucial cytokines essential for the immune response. For instance, upon LPS activation, monocytes are stimulated to release cytokines such as interleukin-6 (IL-6), interleukin-10 (IL-10), and TNF-α, which can be evaluated using ELISA or flow cytometry assays. While LPS activates monocytes to release these cytokines, B cells respond via intricate signaling pathways, highlighting diverse immune cell activation mechanisms during LPS treatment. This LPS stimulation protocol provides a clear method for effective LPS treatment, helping ensure robust immune cell activation, enabling researchers to explore the cellular responses and cytokine production patterns essential for diverse research applications.
Search ELISA kits and multiplex immunoassays Search primary antibodies
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
Immunology at Work Resource Center
Find guides and protocols for immunology research.
Protocols for Immunology
Discover protocols and eLearning courses for various applications to study immunology.
Immune Cell Guide
Download this guide that provides detailed marker information for immune cell types and subtypes.
Cell Signaling Pathways
Investigate signaling pathways and signal transduction.
Cell Isolation and Expansion
Learn more about Dynabeads activation and expansion technology that mimics in vivo T cell activation.
Immunoassays
Explore a wide range of immunoassays for various research applications.
Flow Cytometry Solutions
Explore a wide range of immunoassays for various research applications.
Cell Analysis Support
Find technical information, tips and tricks, and answers to everyday problems.
For Research Use Only. Not for use in diagnostic procedures.