Promethium • Lanthanide Rare Earth
Primary XPS region: Pm3d
Overlapping regions: N/A
Binding energies of common chemical states: N/A
Experimental information
N/A
Interpretation of XPS spectra
N/A
About this element
Element Crystal hex
Symbol: Pm
Date of discovery: 1945
Name origin: Greek prometheus
Appearance: unknown
Discoverer: J.A. Marinsky, et al.
Obtained from: fission of U, Th or Pu
Melting point: 1371 K
Boiling point: 3273 K
Density[kg/m3]: 7264
Molar volume: 20.23 × 10-6 m3/mol
Protons/Electrons: 61
Neutrons: 84
Shell structure: 2,8,18,23,8,2
Electron configuration: [Xe]6s24f5
Oxidation state: 3
Crystal structure: hexagonal
Promethium has been found in the spectrum of the star HR465 in Andromeda, but does not occur naturally on earth. Little is known of metallic promethium’s properties; however, promethium salts glow in the dark with a pale blue or green color because they are highly radioactive. Because of its radioactivity, handling promethium requires great care, as it can emit X-rays during its beta decay. Promethium is used in a nuclear battery where photocells convert light into electric current, with a useful battery life of around five years. It is also a beta radiation source for thickness gauges. The element’s name originates from Prometheus of Greek mythology who stole fire from the gods and gave it to mankind.
element_promethium
Products
Nexsa XPS
  • Módulo de basculación para mediciones de ARXPS
  • Fuente de iones de modo dual para capacidades de realización de perfiles de profundidad
  • Análisis de aislantes.
Hoja de estilo para tarjetas originales instrumentos
K-Alpha XPS
  • Espectroscopía de área seleccionable.
  • Monocromador microfocalizado.
  • Espectroscopía del estado químico de alta resolución.
ESCALAB Xi+ XPS
  • Espectroscopía de alta sensibilidad.
  • XPS con fuente de rayos X no monocromática.
  • Analizador hemisférico de 180°.
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Hoja de estilo para las especificaciones de la tabla de productos
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Servicios de microscopía electrónica para
la ciencia de materiales
Para garantizar un rendimiento óptimo del sistema, le proporcionamos acceso a una red de expertos de primer nivel en servicios de campo, asistencia técnica y piezas de repuesto certificadas.