Search
Lentiviral and Adenoviral Delivery of shRNA and miR RNAi Vectors |
Having evolved to proficiently deliver nucleic acids to cells, viruses offer a means to reach hard-to-transfect cell types for protein overexpression or knockdown. Adenoviral, oncoretroviral, and lentiviral vectors have been used extensively for delivery in cell culture and in vivo (Table 1).
Adenoviruses are DNA viruses that can transiently transduce nearly any mammalian cell type. The adenovirus enters target cells by binding to the coxsackie adenovirus receptor (CAR). After binding to the CAR, the adenovirus is internalized via integrin-mediated endocytosis followed by active transport to the nucleus where its DNA is expressed episomally.
Oncoretroviruses and lentiviruses are positive-strand RNA viruses that stably integrate their genomes into host cell chromosomes. When pseudotyped with an envelope that has a broad tropism, such as vesicular stomatitis virus glycoprotein (VSV-G), these viruses can enter virtually any mammalian cell type. However, the oncoretroviruses depend upon nuclear membrane breakdown during cell division to transduce cells. In contrast, lentiviruses are more versatile tools, as they use an active nuclear import pathway to transduce non-dividing cells.
The choice of viral system depends on a variety of factors, including length of expression and which cells will be transduced.
| Viral system | Transient expression | Stable expression | ||||
|---|---|---|---|---|---|---|
| Dividing cells | Nondividing cells | Dividing cells | Neuronal cells | Drug- or growth- arrested cells | Contact- inhibited cells | |
| Adenovirus | ||||||
| Lentivirus | ||||||
| Retrovirus | ||||||
For many disease models, the most desirable cell types, such as immune system or primary cells, are not amenable to transfection. Viral delivery of RNAi vectors is a powerful alternative to transfection for these cell types as well as for in vivo applications. To accurately determine the efficacy of knockdown from an RNAi vector in a population of cells, it is critical to deliver the RNAi vector to as many cells as possible. Otherwise, when knockdown is measured by quantitative real-time PCR (qRT-PCR) or western analysis, the background of mRNA or protein in nontransfected cells will make the knockdown appear less effective than it is. Viral delivery can be the best option in virtually any mammalian cell type, including hard-to-transfect, primary, and even nondividing cell types. Conveniently, lentiviral delivery systems are available for both shRNA and miRNA vectors, and an adenoviral delivery system is available for shRNA vectors.
Figure 1. How the BLOCK-iT Lentiviral RNAi System works.
The product manuals for the adenoviral and lentiviral expression systems have more detailed protocols for generating and titering adenoviral and lentiviral stocks.
The first step for successful transductions is to choose the system for your experimental goals (Table 1). Find kits to clone and produce adenoviral and lentiviral stocks as well as services to clone these viral vectors and produce the viral stocks. The BLOCK-iT Pol II miR RNAi vector system and the shRNA vector system are not compatible with each other and employ different lentiviral and adenoviral vector backbones. Once you have your miR RNAi or shRNA sequence in a lentiviral backbone or adenoviral backbone, producing the viral stocks follows the same protocols for both vector systems.
| Product or service | When to use | Key advantages |
|---|---|---|
| Lentiviral delivery | ||
| BLOCK-iT HiPerform Lentiviral Pol II miR RNAi Expression System with EmGFP |
|
|
| BLOCK-iT Lentiviral Pol II miR RNAi Expression System |
|
|
| BLOCK-iT Lentiviral Pol II miR RNAi Expression System with EmGFP |
|
|
| BLOCK-iT Inducible H1 Lentiviral RNAi System |
|
|
| BLOCK-iT Lentiviral RNAi Expression System |
|
|
| Lentiviral cloning & lentiviral production services |
|
|
| ViraPower Lentiviral Packaging Mix |
|
|
| Adenoviral delivery | ||
| BLOCK-iT Adenoviral RNAi Expression System |
|
|
| ViraPower Adenoviral Gateway Expression Kit |
|
|
| Adenoviral cloning & adenoviral production services |
|
|
Technical inquires:
Our Technical Application Scientists are available to help assist you at techsupport@thermofisher.com
Ordering & Order Status inquires:
If you have questions about pre-designed RNAi orders and order status, please contact us at genomicorders@thermofisher.com
If you have any questions about Custom RNAi orders and order status, please contact us at RNAiSupport@thermofisher.com
For Research Use Only. Not for use in diagnostic procedures.