Discover and profile expression of small RNAs
From pure small RNA discovery to small RNA expression analysis, cutting-edge Ambion™ RNA isolation technology, combined with Ion Torrent™ next-generation sequencing is accessible to any lab interested in small RNA function.
Request a quote
View and order products >
Introduction to small RNA sequencing
Read more about the basics of small RNA sequencing and solutions for the Ion S5™ and Ion PGM™ systems
Informatics solutions
Learn about small RNA sequencing informatics solutions for the fastest way to get to your biological results
Small RNA sequencing workflow
Learn about small RNA sequencing solutions
Solutions for small RNA sequencing
Ion S5 System with Ion 520™, Ion 530™, and Ion 540™ chips
Ion Total RNA-Seq Kit v2 to purify small RNA and construct strand-specific libraries
PureLink™ miRNA Isolation Kit provides fast, flexible, and high-quality sequencing of small RNA
Small RNA sequencing workflow for Ion Torrent next-generation sequencing systems
1 Purify
Use the PureLink miRNA Isolation Kit to isolate small RNA from tissues and cells.
2 Construct
Construct library for RNA-Seq
Use the Ion Total RNA-Seq Kit v2 and optional Ion Xpress™ RNA-Seq Barcode 1-16 Kit to make small RNA libraries that preserve strand information.
3 Prepare
Prepare Template for RNA-Seq
Use the Ion Chef™ System for automated template preparation and chip loading. Suitable for users of all experience levels.
4 Run
Run Sequence for RNA Sequencing
Discover and profile small RNA on the Ion S5 or Ion PGM™ systems. Barcodes enable you to maximize the number of samples analyzed in a single sequencing run.
5 Analyze
Analyze data for RNA Sequencing
Torrent Suite™ Software automatically provides sequence reads in exportable FASTQ and BAM formats, simplifying further analysis using third-party software.
Small RNA sequencing informatics solutions
Ion Torrent semiconductor sequencing combines a simple, integrated wet-lab workflow with Torrent Suite™ Software and third-party solutions for fast identification, characterization, and reporting of small RNA expression.
Sequencing run reports are provided, and with expandable analysis plots and straightforward tables summarizing key results, are easily browsed to help ensure high-quality sequencing runs. With a simple right-click, Torrent Suite Software provides the sequence reads in exportable FASTQ and BAM file formats for direct upload to third-party analysis software.
Third-party software, such as Partek™ Flow™ Software packages, performs read alignment, microRNA level abundance measurements, and multi-sample differential microRNA expression analysis. Partek also provides quality-control metrics and interactive data visualization.
Introduction to small RNA sequencing
Since the discovery of microRNA (short ~22mer RNA sequences that bind to mRNA and inhibit translation), the study of gene expression regulation has expanded tremendously. With the advent of next-generation sequencing (NGS) in the last decade, researchers have harnessed the power of deep sequencing to further reveal the complexity of these small RNA species, which have expanded to include multiple classes of small RNA such as microRNA (miRNA), short-interfering RNA (siRNA), and piwi-interacting RNA (piRNA), among others.
Ambion RNA kits offer a perfect way to isolate and purify your samples, enabling high-quality capture of small RNA species. Leveraging the knowledge leadership of Thermo Fisher Scientific and integrating kits that isolate and purify small RNA species creates a complete solution that is optimized for small RNA analysis on the Ion S5 and Ion PGM systems. The scalability of these systems enables you to include a range of samples per run, giving you the throughput flexibility to match your experiment sizes. Although other NGS platforms can also analyze small RNA, their ultrahigh-throughput format requires batching hundreds of samples per run, potentially delaying your time to results.
Choose the Ion PGM System, which enables fast and accurate results for small RNA and targeted RNA studies, while the Ion S5 System offers higher throughput, enabling you to scale from small RNA to whole transcriptome analysis. Both systems surpass microarrays as a tool for expression analysis, with these advantages:
  • Superior sensitivity, with a greater dynamic range, to detect more transcripts and subtle changes in gene expression
  • Discrimination between sequences from closely related small RNA families
  • Detection of novel small RNA sequences
  • Digital output enabling results to be compared over time and between studies and labs
  • Close correlation with TaqMan™ Assay results