Search
Vacuum pumps are designed to reduce the pressure within a system thereby lowering the boiling point of a solvent. By reducing the boiling point, the solvent evaporates at a much lower temperature, which reduces sample degradation. Two pump styles are available from Thermo Scientific: traditional rotary vane oil pumps and the newer oil-free, maintenance free vacuum pumps.
Achieve maximum solute recovery with the Thermo Scientific™ Deep Vacuum Oil Pumps for Vacuum Concentrators, designed to work with all Savant™ SpeedVac™ concentrating and freeze drying systems.
Features:
All Rotary Vane Oil Pumps come standard with:
| Description | Pump Displacement | 115V/60Hz | 230V/50Hz |
|---|---|---|---|
| Oil Vacuum Pump for Freeze Drying | 76L/min throughput at 60 Hz | FDP80-115 | FDP80-230 |
| Oil Vacuum Pump for Freeze Drying | 195L/min throughput at 60 Hz | FDP200-115 | FDP200-230 |
| Oil Vacuum Pump for Freeze Drying | 195L/min throughput at 60 Hz | VLP195FD-115 | VLP195FD-230 |
| Deep Oil Vacuum Pump for Freeze Drying | 116L/min throughput at 60 Hz 97L/min throughput at 50 Hz | VLP120-115 | VLP120-230 |
| Deep Oil Vacuum Pump for Freeze Drying | 195L/min throughput at 60 Hz 162L/min throughput at 50 Hz | VLP200-115 | VLP200-230 |
| Deep Oil Vacuum Pump for Freeze Drying | 283L/min throughput at 60 Hz 237L/min throughput at 50 Hz | VLP285-115 | VLP285-230 |
| Deep Oil Vacuum Pump for Freeze Drying | 76L/min throughput at 60 Hz 62L/min throughput at 60 Hz | VLP80-115 | VLP80-230 |
Eliminate oil mist vapors with Thermo Scientific™ Oil-Free Vacuum Pumps for Vacuum Concentrators, designed for a wide variety of laboratory uses. These quiet-running pumps are highly chemical resistant, and do not require oil to operate.
Features:
| Description | Pump Displacement | 115V/60Hz | 230V/50Hz |
|---|---|---|---|
| Oil-Free FTE/PFA 4-headed Vacuum Pump | 30L/min throughput at 50 Hz | OFP400-115 | OFP400-230 |
| Oil-Free Scroll Pump | 96.7L/min throughput at 50 Hz | OFP5C-115 | OFP5C-230 |
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.