Search
For successful formation of new blood vessels, endothelial cells in an existing blood vessel must degrade the surrounding basement membrane. Upon activation by angiogenic stimuli, endothelial cells produce various matrix metalloproteinases (MMPs)–proteolytic enzymes that cleave the protein backbone of the basement membrane and surrounding extracellular matrix in order to facilitate cellular migration into the extracellular space.1
The matrix invasion assay is used to measure the ability of cells to degrade the basement membrane. Although it was originally developed to measure the invasive capability of tumor cells, it has been adapted for various cell types. This assay employs a modified Boyden chamber in which endothelial cells are placed in an upper compartment of a cell culture insert that contains a cell permeable membrane coated with a basement membrane extract. This extract occludes the pores of the membrane and prevents cell migration. Then a solution containing the test agent is placed below the cell permeable membrane. Following an incubation period (4–48 hours), the cells that have degraded the matrix and migrated through the membrane are stained and counted.
Variables affecting this assay that must be optimized by the investigator include cell type, cell seeding density, type and thickness of matrix barrier, incubation time, and type of angiogenic factor being tested. This assay has the advantage of being quantitative, flexible, and rapid. However, it can be difficult setting up the assay chamber. Commercially available cell culture inserts have alleviated this burden.
Primary cells—find primary cells from human, mouse, rat and other species.
Cell culture products—find media, fetal bovine serum (FBS), cell culture reagents and antibiotics, and cell culture supplements.
Extracellular matrices—find Geltrex and AlgiMatrix matrices as well as laminin, collagen, fibronectin, gelatin, and poly-D-lysine.
Cell culture plastics—find cell culture inserts, dishes, plates, flasks, tubes and other standard culture surfaces as well as cryoware and pipettes.
Cell dissociation solutions—find TrypLE solution as well as StemPro Accutase reagent and standard trypsin.
Cell analysis solutions—find microscopes, high-content platforms and microplate readers for cell analysis, review our portfolio of fluorescent probes for cell structure analysis, find products for cell tracing and tracking, and see options for determining cell viability and proliferation.
Growth factors and recombinant proteins—find growth factors and recombinant proteins for your cell biology investigations, including angiogenesis inducers and inhibitors.
ELISA kits—find ELISA kits by target.
Antibodies—search from thousands of Invitrogen primary and secondary antibodies validated and cited for flow cytometry, IHC/IF/ICC, western blotting, ELISA and other applications.
LT168 updated 6-Oct-2011
For Research Use Only. Not for use in diagnostic procedures.