TaqMan gene expression array cards and plates
High throughput gene expression analysis is a powerful technique that allows for the simultaneous examination of thousands of genes across multiple samples, providing comprehensive insights into cellular functions and biological processes. TaqMan Arrays contain TaqMan Gene Expression Assays dried down in three array formats: 96- and 384-well TaqMan Array plates, 384-well TaqMan Array cards (microfluidic cards), and OpenArray plates. These qPCR arrays are ideal for gene expression profiling and verification applications that require the analysis of tens to thousands of targets. The streamlined workflow of these arrays, which involves simply adding master mix and cDNA samples, helps ensure efficient and reproducible results, facilitating robust data acquisition and analysis. Additionally, their flexibility in format and customizable panel options make them suitable for a wide range of research needs, from small-scale studies to large-scale genomic projects.
TaqMan gene expression array content options
Predesigned TaqMan gene expression arrays
photo of single tube predesigned TaqMan gene expression assays
These products feature our most popular TaqMan gene expression assays targeting relevant genes for specific biological pathways, processes, or diseases. Use the TaqMan Assay search tool to search for TaqMan Array cards or plates or browse the complete list of inventoried TaqMan arrays.
Search all TaqMan gene expression arrays
Flexible (made-to-order) TaqMan gene expression arrays
TaqMan flexible gene expression array in card and plate formats
For scientists who prefer not to design their panels from scratch, flexible content TaqMan arrays are an excellent choice. Users can begin by selecting a panel of commonly studied pathways, diseases, biological processes, or cellular functions (human, mouse, or rat). Each panel is pre-populated with specifically chosen assays and serves as a starting point for modification using a simple online configuration tool found on the webpage linked below. Within the tool, users can swap out genes of interest, reposition assays, or change the layout to fit their experimental design. The minimum order quantity is six plates or ten cards.
Explore:Flexible TaqMan arrays
Custom TaqMan gene expression arrays
product photo of custom gene expression arrays
Customized cards and plates allow researchers to configure their panel from scratch by choosing from any of the 2.8 million predesigned TaqMan Gene Expression assays. By uploading their assay list, the simple-to-use online configuration tool facilitates easy drag-and-drop arrangement of assays. The minimum order quantity is six plates or ten cards.
Learn more:Custom TaqMan array plate (96-well 0.1 mL or 0.2 mL)
Learn more:Custom TaqMan array card (384-well)
Learn more:Custom OpenArray plate
Specialty TaqMan gene expression arrays
Product photo of specialty TaqMan gene expression assays tubes, plates and arrays
For a custom TaqMan Array that incorporates custom TaqMan gene expression assays, TaqMan SNP genotyping assays, TaqMan copy number assays, TaqMan miRNA assays, or a combination of these assay types, the Specialty TaqMan plating service is available. This service provides TaqMan assays in 96- or 384-well plates, as well as 384-well microfluidic cards, all configured to meet specific research needs.
Learn more:Specialty TaqMan array cards and plates
How to choose a gene expression array
Instrument compatibility table for gene expression arrays
Product format/InstrumentQuantStudio 3QuantStudio 5QuantStudio 12K Flex
96-well plate, standard standard 20 µL✓*✓**✓
96-well plate, fast 10 µL✓*✓**✓
384-well plate ✓**✓
TaqMan array card (TAC)  ✓
OpenArray  ✓
*QuantStudio 3 comes with either a 96-well standard OR 96-well fast block
**QuantStudio 5 comes with either a 96-well standard, 96-well fast block, OR 384-well block
Inventoried TaqMan gene expression arrays
High throughput gene expression resources
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments