Search
This new internal pump will be used in the models 42iQ, 42iQD, 42iQLS and 42iQHL (with bypass option). The new pump is the same form, fit and function and meets all Thermo Scientific specifications and applicable certifications to be used in the above listed 42iQ series of NOx analyzers.
This new internal pump will be used in the models 42i, 42iD, 42iLS. The new pump is the same form, fit and function and meets all Thermo Scientific specifications and applicable certifications to be used in the above listed 42i series of NOx analyzers. Please note: This Internal pump is also used in the 43iHL SO2 Analyzer.
Thermo Scientific 49iQ Ozone Primary Standards shipped prior to 06/01/2019 were manufactured with a minor zero air plumbing configuration error that can affect the operation of the calibrator depending on the zero air pressure supplied to the calibrator.
We have received feedback that some Thermo Scientific 49iQPS instruments are exhibiting erroneous ozone concentrations. This appears to be related to long pieces of tubing connected to the vent, resulting in back pressure that impedes flow through the unit. A solution has been identified and all new units have this modification installed. Units in the field can be retrofitted by installing an upgrade kit.
The USB firmware update method will not work on instruments currently running iQ Series firmware version 1.6.11.33737. To upgrade any of the iQ instruments from version V1.6.11.33737, you must use ePort software to do a firmware update.