shutterstock_239255980-(1)
TaqMan Gene Expression Assays enable discovery and verification of genomic biomarkers of well-understood checkpoint inhibitors and other immune-related genes.
Immuno-oncology is one of the most promising areas of research in the world of anti-cancer therapeutics. Rather than targeting the cancer cell directly, immuno-oncology, also known as cancer immunotherapy, studies how to manipulate the body’s own immune system to mount an attack against tumor cells.
The potential benefits of immuno-oncology include long-lasting tumor regression where surgery, radiotherapy, chemotherapy, and targeted therapy have proven less effective.
Within immuno-oncology, checkpoint inhibitor therapy is a focus that continues to garner widespread attention. By understanding the effects of immune checkpoints, therapies can be developed that leverage the body’s innate immune response to attack cancer.
Our Applied Biosystems TaqMan Gene Expression Assays enable researchers to reliably detect and quantitate the expression levels of these immune response related genes and checkpoint markers.
Explore our TaqMan solutions for immuno-oncology research
TaqMan Gene Expression Assay products
Download a list of genes and corresponding assay IDs for nearly 400 biomarkers involved in immuno-oncology research.
Commonly studied checkpoint markers: 
Already know your target of interest? 
Search for a predesigned TaqMan Assay for your target using the tool below. 
Assay search tool
TaqMan Array products
In addition to single tube assays, we also have a variety of both configurable and fixed content TaqMan Array products focused on immuno-oncology research. TaqMan Arrays are pre-spotted 96-well plates or 384-well microfluidic array cards, saving you the time of pipetting many assays into an empty plate. Just add your cDNA and master mix and you’re ready to go.
TaqMan Array Human Immune Response Plate
96-well (0.2 mL) plate format containing 96 common immune response biomarkers
TaqMan Array Human Immune Response Plate, Fast
96-well (0.1 mL) format containing the same set of 96 targets
TaqMan Array Human Immune Response Panel 
384-well array card format; test 96 targets in four samples in a single experiment with the efficient workflow advantage of TaqMan Array Cards; fill 384 wells in less than 15 minutes
TaqMan Array Mouse Immune Response Plate 
96-well (0.2 mL) plate format containing 96 common immune response biomarkers
TaqMan Array Mouse Immune Response Plate, Fast
96-well (0.1 mL) format containing the same set of 96 targets
TaqMan Array Mouse Immune Response Panel
384-well array card format; test 96 targets in four samples in a single experiment with the efficient workflow advantage of TaqMan Array Cards; fill 384 wells in less than 15 minutes
Looking for more Immune Response arrays? View all of our predesigned array plates and cards by searching a gene symbol here
For Research Use Only. Not for use in diagnostic procedures.