Applied Biosystems® GeneAmp® PCR System 9700 is specifically designed for reliable PCR. The GeneAmp® PCR System 9700 consists of a base module and one of many interchangeable sample block modules to provide you with the flexibility to meet your changing research needs.
Request a demo
Which GeneAmp® PCR System 9700 thermal cycler is right for you?
Thermal Cycler 60-Well 96-Well  Aluminum 96-Well Gold
96-Well Silver
Dual 96-Well Dual 384-Well
Block Description0.5 mL aluminum0.2 mL 96-well0.2 mL 96-wellDual block aluminium 0.2 mL 96-well blocks2 aluminium 0.2 mL 384-well blocks
FeaturesSupports 0.5 mL thin walled tubesStandard 0.2 mL format, more cost effectiveStandard 0.2 mL format, faster ramp speeds0.2 mL format, 192 samples per runDual blocks enable up to 768 samples per run, optional auto-lid
Temperature Accuracy±0.25°C from 35.0°C–99.9°C
Height26 cm (10 in.)
Width30 cm (12 in.)
Depth40.6 cm (16 in.)52 cm (20.5 in.)
The GeneAmp 9700 PCR system is being discontinued. See details
PCR reagents
We offer a range of PCR enzymes to fit with your applications. With products to choose from for Fast PCR, Hot Start specificity, High Fidelity in cloning and Long fragment amplification we have the enzyme for you! Select the product for you!
Plastics consumables
Optimized for use on Applied Biosystems Thermal Cyclers, MicroAmp® disposables provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification. See our compatibility chart to choose the plastics that will work for you.
Mitochondrial DNA typing screens with control region and coding region SNPs
Publication: Journal of Forensic Science 2005 Mar;50(2):377-85.
Authors: Kline MC, Vallone PM, Redman JW, Duewer DL et al.
Linkage Disequilibrium Mapping
The linkage disequilibrium maps of three human chromosomes across four populations reflect their demographic history and a common underlying recombination pattern.
Publication: Genome Research 2005 Apr;15(4):454-62. Epub 2005 Mar 21
Authors: De La Vega FM, Isaac H, Collins A, Scafe CR et al.