Search Thermo Fisher Scientific
Organism Type | Human | T-REx cell lines stably express the tetracycline repressor protein. They are functionally tested by transient transfection with the positive control vector pcDNA4/TO/lacZ. T-REx cell lines exhibit extremely low basal expression levels in the repressed state and high expression upon induction with tetracycline. |
Tissues | Kidney | |
Phenotype | Adherent | |
Primary | no | |
Application | Protein Expression,RNAi,Stable Cell Transfection |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||