Icon for Research Scale Purification
The research portfolio comprises of a unique set of affinity resins for purification of a variety of antibody formats and species, a number of resins for purification of biosimilars, an innovative tag system based on only four amino acids and a set of products for plasma sample preparation.
Request more information ›       Order Now
Struggling with mAbs purity and yield in downstream process?
Watch our webinar, "Building a Purification Toolkit for mAb Therapeutics," to learn about effective strategies, chromatography resins, and buffer integration.
  • Antibody purification
    Purify virtually any antibody format from any species using our diverse product portfolio
  • C-tag
    Smallest affinity tag system (four amino acid recognition) available on the market, offering high selectivity, affinity, and a variety of elution conditions
  • POROS metal chelate columns
    Prepacked Immobilized Metal Affinity Column is based on an imidodiacetate functional group designed for the purifiction of proteins that form co-ordination complexes via metal sites and specific amino acids, such as histidine-tagged fusion proteins.
  • Plasma sample preparation
    Single-step, single-use depletion of highly abundant proteins from human serum and plasma, 99% protein removal before 2D gel or mass spectrometry analysis.
  • POROS HQ 10 & 20 µm columns
    Prepacked Strong Anion Exchange Column is based on a quarternized polyethyleneimine functional group yielding a high capacity, Perfusion Chromatography media designed for the separation and purification of biomolecules.
  • POROS PI 20 µm columns
    Prepacked Weak Anion Exchange Column is based on a polyethyleneimine functional group yielding a high capacity, Perfusion Chromatography media designed for the separation and purification of biomolecules.
  • POROS HS 20 µm columns
    Prepacked Strong Cation Exchange Column is a high capacity, strong cation exchange Perfusion Chromatography media suitable for the separation and purification of biomolecules.
  • POROS S 10 & 20 µm columns
    Prepacked Strong Cation Exchange Column is a strong cation exchange Perfusion Chromatography media, suitable for the separation and purification of biomolecules.
  • POROS R2 10 & 20 µm columns
    Pre-packed Reversed-Phase Column is a 2,000 angstrom pore size poly (styrene-divinylbenzene) particle, suitable for the reversed-phase separation of biomolecules.
  • POROS R1 10 & 20 µm columns
    Pre-packed Reversed-Phase Column is a 4,000 angstrom pore size Poly (Styrene-Divinylbenzene) particle, suitable for the reversed-phase separation of biomolecules.
  • POROS HP2 20 µm columns
    Prepacked Hydrophobic Interaction Chromatography Column is a high-density phenyl, Perfusion Chromatography media suitable for the separation and purification of proteins.
  • Antibody purification
    Purify virtually any antibody format from any species using our diverse product portfolio
  • C-tag
    Smallest affinity tag system (four amino acid recognition) available on the market, offering high selectivity, affinity, and a variety of elution conditions
  • POROS metal chelate columns
    Prepacked Immobilized Metal Affinity Column is based on an imidodiacetate functional group designed for the purifiction of proteins that form co-ordination complexes via metal sites and specific amino acids, such as histidine-tagged fusion proteins.
  • Plasma sample preparation
    Single-step, single-use depletion of highly abundant proteins from human serum and plasma, 99% protein removal before 2D gel or mass spectrometry analysis.
  • POROS HQ 10 & 20 µm columns
    Prepacked Strong Anion Exchange Column is based on a quarternized polyethyleneimine functional group yielding a high capacity, Perfusion Chromatography media designed for the separation and purification of biomolecules.
  • POROS PI 20 µm columns
    Prepacked Weak Anion Exchange Column is based on a polyethyleneimine functional group yielding a high capacity, Perfusion Chromatography media designed for the separation and purification of biomolecules.
  • POROS HS 20 µm columns
    Prepacked Strong Cation Exchange Column is a high capacity, strong cation exchange Perfusion Chromatography media suitable for the separation and purification of biomolecules.
  • POROS S 10 & 20 µm columns
    Prepacked Strong Cation Exchange Column is a strong cation exchange Perfusion Chromatography media, suitable for the separation and purification of biomolecules.
  • POROS R2 10 & 20 µm columns
    Pre-packed Reversed-Phase Column is a 2,000 angstrom pore size poly (styrene-divinylbenzene) particle, suitable for the reversed-phase separation of biomolecules.
  • POROS R1 10 & 20 µm columns
    Pre-packed Reversed-Phase Column is a 4,000 angstrom pore size Poly (Styrene-Divinylbenzene) particle, suitable for the reversed-phase separation of biomolecules.
  • POROS HP2 20 µm columns
    Prepacked Hydrophobic Interaction Chromatography Column is a high-density phenyl, Perfusion Chromatography media suitable for the separation and purification of proteins.
Application Notes and Webinars
For Research Use Only. Not for use in diagnostic procedures.
Bioprocessing resources
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing: