Search
View and control the Applied Biosystems thermal cyclers in your facility with Applied Biosystems Thermal Cycler Fleet Control Software.
Fleet Control Software comes pre-installed on a server that sits securely behind your firewall giving you the security you need.
Your thermal cycler fleet control can be as restrictive or open as your work requires. Set up permissions in the custom rules menu for methods, users, and instruments. Restrict methods to specific instruments or users, or limit user access to specific methods or instruments.
Fleet Control Software arrives pre-installed on a Dell OptiPlex 3050 All-In-One server. Operate your Fleet Control Software from the server or from any computer or device on the same network through Chrome internet browser. A mouse, keyboard and 3-year warranty are also included.
| Operating System | Debian Linux |
| Processor | Intel Celeron G3930T (DC/2MB/2T/2.7GHz/35W) |
| Memory | 4GB (1X4GB) 2400MHz DDR4 Non-ECC |
| LCD | 19.5 inch TN Wide Viewing Angle WLED, HD+ 1600x900 resolution with anti-glare coating |
| Video Card | Integrated Intel HD Graphics |
| Hard Drive | 2.5 inch 128GB SATA Class 20 Solid State Drive |
| Network Interface Card | Ethernet LAN 10/100/1000 |
| Wireless | Intel Dual Band Wireless AC 3165 (802.11ac) 1x1 + Bluetooth |
| Power Supply | External Power Supply Unit; 130W adapter |
| Keyboard | US English Dell KB216 QuietKey USB Keyboard Black |
| Mouse | Dell USB Laser 6-Button Mouse |
Fleet Control Software comes pre-installed on a server that sits securely behind your firewall giving you the security you need.
Your thermal cycler fleet control can be as restrictive or open as your work requires. Set up permissions in the custom rules menu for methods, users, and instruments. Restrict methods to specific instruments or users, or limit user access to specific methods or instruments.
Fleet Control Software arrives pre-installed on a Dell OptiPlex 3050 All-In-One server. Operate your Fleet Control Software from the server or from any computer or device on the same network through Chrome internet browser. A mouse, keyboard and 3-year warranty are also included.
| Operating System | Debian Linux |
| Processor | Intel Celeron G3930T (DC/2MB/2T/2.7GHz/35W) |
| Memory | 4GB (1X4GB) 2400MHz DDR4 Non-ECC |
| LCD | 19.5 inch TN Wide Viewing Angle WLED, HD+ 1600x900 resolution with anti-glare coating |
| Video Card | Integrated Intel HD Graphics |
| Hard Drive | 2.5 inch 128GB SATA Class 20 Solid State Drive |
| Network Interface Card | Ethernet LAN 10/100/1000 |
| Wireless | Intel Dual Band Wireless AC 3165 (802.11ac) 1x1 + Bluetooth |
| Power Supply | External Power Supply Unit; 130W adapter |
| Keyboard | US English Dell KB216 QuietKey USB Keyboard Black |
| Mouse | Dell USB Laser 6-Button Mouse |