SuperScript IV Reverse Transcriptase (RT) is a proprietary MMLV (Moloney Murine Leukemia Virus) RT mutant designed for reduced RNase H activity, increased thermostability, and highly efficient full-length cDNA synthesis. Compared to previous generation SuperScript RTs, SuperScript IV reverse transcriptase has significantly improved processivity, which enables fast reaction speed, inhibitor resistance, and exceptional performance even with challenging RNA samples. SuperScript IV reverse transcriptase is available in multiple formats tailored to specific applications, and it is widely cited in peer reviewed journals.
Advantages of SuperScript IV Reverse Transcriptase
SuperScript IV Reverse Transcriptase is known for its efficiency, sensitivity, robustness, short-reaction time, and thermostability. For recommendations on choosing the reverse transcriptase fit for your application, use the selection tool.
  • Efficient—Up to 100x higher cDNA yield
  • Sensitive—Ct values reduced by as many as eight cycles for RT-qPCR
  • Robust—Transcribes degraded and inhibitor-containing RNA samples
  • Fast—10-minute cDNA synthesis time
  • Stable—High thermostability to transcribe structured templates
 Read white paper to see data
SuperScript IV Reverse Transcriptase formats: Selection guide
SuperScript IV RT is available in several formats (Table 1)
  • Stand-alone enzyme—Reverse transcriptase supplied with reaction buffer
  • First-strand synthesis system—All cDNA synthesis reaction components are in separate tubes for maximum flexibility of reaction conditions
  • Master mix—cDNA synthesis reaction components are premixed for exceptional efficiency and reduced variability in RT-qPCR applications
  • One-step RT-PCR system—RT and PCR reagents for one-step RT-PCR applications
  • Direct reverse transcription—Reagents for cell lysis and cDNA synthesis included; RNA purification is not required
Table 1. SuperScript IV RT formats.
Comparison of SuperScript IV Reverse Transcriptase with other RTs
Stand-alone enzymes offer maximum flexibility in reaction setup. Table 2 compares the Superscript IV Reverse Transcriptase with other generations of reverse transcriptases.
Table 2. Comparison of standalone reverse transcriptases.
 M-MLV-Reverse Transcriptase
SuperScript II Reverse TranscriptaseSuperscript III Reverse Transcriptase
Superscript IV Reverse Transcriptase
Key attribute
Recombinant M-MLV RT for routine, non-demanding applications
Engineered M-MLV RT with reduced RNase H activity
Engineered M-MLV RT with reduced RNase H activity and improved thermal stability
Latest generation engineered RT for exceptional cDNA synthesis performance
Optimal reaction temperature37°C42°C50°C50–55°C
Reaction time50 min50 min30–50 min10 min
Sensitivity1 ng1 ng10 pg10 pg
Max cDNA lengthUp to 7 kbUp to 12.3 kbUp to 12.3 kb>12 kb
Ability to work with degraded or inhibitor-containing RNALowLowMediumHigh
Reduced RNase H activityNoYesYesYes
Performance of SuperScript IV Reverse Transcriptase
High sensitivity
Low Ct values. Ct values reduced by 8 cycles compared to other reverse transcriptases (Figure 2).
Figure 2. Sensitivity and variability in cDNA synthesis using degraded RNA samples. Degraded Arabidopsis total RNA (RIN: 1–3), in amounts of 1, 10, and 100 ng were used as input RNA in 20 μL reverse transcription reactions with SuperScript IV Reverse Transcriptase and random hexamers according to the product protocol. RTs from other vendors were used according to the manufacturers’ protocols. For each tested enzyme and each RNA input, RT reactions were performed in triplicates. From each reverse transcription reaction, 10% of the cDNA product was added to TaqMan assays for two targets, Gln synthetase and WRKY TF 70. Three qPCR reactions were performed for each reverse transcription reaction and the average Ct values for each RNA input were plotted (standard deviation from 9 Ct values for each input RNA).
Improved inhibitor tolerance
Compounds that have inhibitory effects on RTs are commonly found in RNA samples even after thorough purification. These compounds may interfere with cDNA synthesis, produce false RT-PCR and RT-qPCR results, and cause results to be misinterpreted. RT inhibitors include reagents used during RNA extraction, and co-purified contaminants arising from biological samples (Table 3). SuperScript IV Reverse Transcriptase shows significantly improved resistance to contaminating inhibitors when compared to previous generation SuperScript III Reverse Transcriptase and other commercially available RTs (Figure 3).
Table 3. Common cDNA synthesis inhibitors and their sources.
InhibitorSource
Ethanol/isopropanol, salts, phenol/chloroform, detergentsSample preparation
Hematin, bile saltsBlood, feces
Humic acid, polyphenols, polysaccharidesSoil, plants
Formalin, paraffinFFPE
Figure 3. Higher performance in cDNA synthesis in the presence of biological or sample prep inhibitors. A 0.5–10 kb RNA ladder was used in a 10 μL SuperScript IV Reverse Transcriptase reaction with oligo(dT)20 according to the product protocol. RTs from other vendors were used according to their respective protocols. Inhibitors were added to the RNA samples prior to primer annealing or addition of RT reaction mix. First-strand cDNAs were resolved by alkaline gel electrophoresis, and cDNA was stained using Invitrogen SYBR Gold Nucleic Acid Gel Stain. During electrophoresis, NaOH hydrolyzes all RNA, resulting in visualization of cDNA only.
Short reaction time
SuperScript IV Reverse Transcriptase has remarkable processivity and thereby generates long, full-length cDNA fragments within a short reaction time. Figure 4 demonstrates that SuperScript IV Reverse Transcriptase synthesized cDNAs of up to 9 kb in 10 minutes, while most other commercially available RTs were only able to synthesize cDNAs between 1.5–3 kb or less in the same duration.
Figure 4. Fast cDNA synthesis capability. The Invitrogen Millennium RNA Marker was used in a 10 μL reaction with SuperScript IV Reverse Transcriptase and oligo(dT) primer according to the product protocol. Other commercially available RTs were used according to manufacturers’ protocols, except for reaction times reduced to 10 minutes. First-strand cDNAs were resolved by alkaline gel electrophoresis, and cDNA was stained using SYBR Gold Nucleic Acid Gel Stain. During electrophoresis NaOH hydrolyzes all RNA, resulting in visualization of cDNA only.
High thermostability
When reverse transcription reactions are performed at low temperatures (less than 42°C), RNA secondary structures, especially GC-rich templates, may interfere with cDNA synthesis. SuperScript IV Reverse Transcriptase has high thermostability and can be used in reactions at 50°C or higher, which facilitates the successful transcription of highly structured RNA transcripts (Figure 5).
Figure 5. High thermostability of SuperScript IV Reverse Transcriptase. A 0.5–10 kb RNA ladder was used in a 10 μL SuperScript IV RT reaction with oligo(dT) according to the product protocol, with the exception that reaction temperature was varied between 50 and 65°C. First-strand cDNAs were resolved by alkaline gel electrophoresis, and cDNA was stained using SYBR Gold Nucleic Acid Gel Stain. During electrophoresis NaOH hydrolyzes all RNA, resulting in the visualization of cDNA only. cDNA bands were quantitated by TotalLab software for each reaction temperature. Percentage SuperScript IV RT activity was calculated by dividing values at each reaction temperature by values at 50°C.
High sensitivity
Low Ct values. Ct values reduced by 8 cycles compared to other reverse transcriptases (Figure 2).
Figure 2. Sensitivity and variability in cDNA synthesis using degraded RNA samples. Degraded Arabidopsis total RNA (RIN: 1–3), in amounts of 1, 10, and 100 ng were used as input RNA in 20 μL reverse transcription reactions with SuperScript IV Reverse Transcriptase and random hexamers according to the product protocol. RTs from other vendors were used according to the manufacturers’ protocols. For each tested enzyme and each RNA input, RT reactions were performed in triplicates. From each reverse transcription reaction, 10% of the cDNA product was added to TaqMan assays for two targets, Gln synthetase and WRKY TF 70. Three qPCR reactions were performed for each reverse transcription reaction and the average Ct values for each RNA input were plotted (standard deviation from 9 Ct values for each input RNA).
Improved inhibitor tolerance
Compounds that have inhibitory effects on RTs are commonly found in RNA samples even after thorough purification. These compounds may interfere with cDNA synthesis, produce false RT-PCR and RT-qPCR results, and cause results to be misinterpreted. RT inhibitors include reagents used during RNA extraction, and co-purified contaminants arising from biological samples (Table 3). SuperScript IV Reverse Transcriptase shows significantly improved resistance to contaminating inhibitors when compared to previous generation SuperScript III Reverse Transcriptase and other commercially available RTs (Figure 3).
Table 3. Common cDNA synthesis inhibitors and their sources.
InhibitorSource
Ethanol/isopropanol, salts, phenol/chloroform, detergentsSample preparation
Hematin, bile saltsBlood, feces
Humic acid, polyphenols, polysaccharidesSoil, plants
Formalin, paraffinFFPE
Figure 3. Higher performance in cDNA synthesis in the presence of biological or sample prep inhibitors. A 0.5–10 kb RNA ladder was used in a 10 μL SuperScript IV Reverse Transcriptase reaction with oligo(dT)20 according to the product protocol. RTs from other vendors were used according to their respective protocols. Inhibitors were added to the RNA samples prior to primer annealing or addition of RT reaction mix. First-strand cDNAs were resolved by alkaline gel electrophoresis, and cDNA was stained using Invitrogen SYBR Gold Nucleic Acid Gel Stain. During electrophoresis, NaOH hydrolyzes all RNA, resulting in visualization of cDNA only.
Short reaction time
SuperScript IV Reverse Transcriptase has remarkable processivity and thereby generates long, full-length cDNA fragments within a short reaction time. Figure 4 demonstrates that SuperScript IV Reverse Transcriptase synthesized cDNAs of up to 9 kb in 10 minutes, while most other commercially available RTs were only able to synthesize cDNAs between 1.5–3 kb or less in the same duration.
Figure 4. Fast cDNA synthesis capability. The Invitrogen Millennium RNA Marker was used in a 10 μL reaction with SuperScript IV Reverse Transcriptase and oligo(dT) primer according to the product protocol. Other commercially available RTs were used according to manufacturers’ protocols, except for reaction times reduced to 10 minutes. First-strand cDNAs were resolved by alkaline gel electrophoresis, and cDNA was stained using SYBR Gold Nucleic Acid Gel Stain. During electrophoresis NaOH hydrolyzes all RNA, resulting in visualization of cDNA only.
High thermostability
When reverse transcription reactions are performed at low temperatures (less than 42°C), RNA secondary structures, especially GC-rich templates, may interfere with cDNA synthesis. SuperScript IV Reverse Transcriptase has high thermostability and can be used in reactions at 50°C or higher, which facilitates the successful transcription of highly structured RNA transcripts (Figure 5).
Figure 5. High thermostability of SuperScript IV Reverse Transcriptase. A 0.5–10 kb RNA ladder was used in a 10 μL SuperScript IV RT reaction with oligo(dT) according to the product protocol, with the exception that reaction temperature was varied between 50 and 65°C. First-strand cDNAs were resolved by alkaline gel electrophoresis, and cDNA was stained using SYBR Gold Nucleic Acid Gel Stain. During electrophoresis NaOH hydrolyzes all RNA, resulting in the visualization of cDNA only. cDNA bands were quantitated by TotalLab software for each reaction temperature. Percentage SuperScript IV RT activity was calculated by dividing values at each reaction temperature by values at 50°C.
Resources for SuperScript IV Reverse Transcriptase
Interactive selection tool: Find the right reverse transcriptase for your experiments.
Video Player is loading.
Current Time 0:00
Duration 2:39
Loaded: 6.16%
Stream Type LIVE
Remaining Time 2:39
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Learn how our commitment to enzyme excellence, including the exceptional reliability and performance of SuperScript IV Reverse Transcriptases, are leading the way in the pursuit of science.
    Video Player is loading.
    Current Time 0:00
    Duration 0:53
    Loaded: 18.56%
    Stream Type LIVE
    Remaining Time 0:53
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Watch this video on the advantages of SuperScript IV Reverse Transcriptase.
      Video Player is loading.
      Current Time 0:00
      Duration 2:37
      Loaded: 6.28%
      Stream Type LIVE
      Remaining Time 2:37
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Understand how SuperScript IV Reverse Transcriptase consistently delivers low Ct values in qPCR reactions.
        Video Player is loading.
        Current Time 0:00
        Duration 1:43
        Loaded: 0.00%
        Stream Type LIVE
        Remaining Time 1:43
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Discover how SuperScript IV Reverse Transcriptase performs despite presence of inhibitors.
          Video Player is loading.
          Current Time 0:00
          Duration 1:02
          Loaded: 0%
          Stream Type LIVE
          Remaining Time 1:02
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • en (Main), selected
            Learn how the SuperScript IV UniPrime One-Step RT-PCR System makes it easy to set up reactions while bringing superior results.
            Brochure
            Manuals
            Protocol
            Technical articles
            SuperScript IV Reverse Transcriptase FAQs
            Find tips, troubleshooting help, and resources for common questions about SuperScript Reverse Transcriptases. Can’t find your question? Search the FAQ database
            The SuperScript IV enzyme has been engineered for higher thermostability, processivity, and cDNA yields. It performs better in the presence of inhibitors, and the reaction buffer has also been optimized for robust cDNA synthesis from a wide range of samples.
            When compared with SuperScript III RT (and other manufacturers’ RTs) in a synthesis reaction for a 9 kb cDNA, SuperScript IV RT performed successful synthesis in just 10 minutes and did so with comparable (or improved) yield.
            SuperScript IV RT sustains 100% activity at up to 56.4°C and 70% activity at up to 65°C, while wild type MMLV RT or MMLV RNase H RT enzymes usually display very low or no activity above 45°C. SuperScript IV RT’s ability to function at higher temperatures enables the reverse transcription of RNA targets with structural complexities.
            SuperScript IV Reverse Transcriptases: Citations
            SuperScript IV reverse transcriptases are highly cited in the several peer reviewed research publications. In three years, between 2022 and 2024, it has been cited over in 10K publications.
            Cancer
            UseReference
            Detect cancer biomarkers from extracellular vesicles by ddPCR (digital droplet).
            Lee KY, Beatson EL, Knechel MA et al. (2024). Detection of Extracellular Vesicle-Derived RNA as Potential Prostate Cancer Biomarkers: Role of Cancer-type SLCO1B3 and ABCC3. J Cancer 15(3):615–622.
            doi: 10.7150/jca.90836. PMID: 38213719.
            cDNA synthesis for preparation of a Poly(A)-seq library and also for nested RT-PCR to study changes in UTR length.
            Gabel AM, Belleville AE, Thomas JD et al. (2024). Multiplexed screening reveals how cancer-specific alternative polyadenylation shapes tumor growth in vivo.Nat Commun 15:959.
            doi: 10.1038/s41467-024-44931-x. PMID: 38302465
            Synthesize cDNA from FFPE samples for ddPCR.
            Look T, Puca E, Bühler M et al. (2023). Targeted delivery of tumor necrosis factor in combination with CCNU induces a T cell-dependent regression of glioblastoma. Sci Transl Med 15(697).
            doi:10.1126/scitranslmed.adf2281 PMID: 37224228
            Viral and infectious diseases
            UseReference
            Reverse transcribe viral RNA to cDNA prior to circularization, RCA, and nanopore sequencing.
            Misu M, Yoshikawa T, Sugimoto S (2023). Rapid whole genome sequencing methods for RNA viruses. Front Microbiol 23: 14.
            doi: 10.3389/fmicb.2023.1137086. PMID: 36910229
            Reverse transcribe avian viral RNA prior to amplification and library prep.
            Narvaez SA, Harrell TL, Oluwayinka (2023). Optimizing the Conditions for Whole-Genome Sequencing of Avian Reoviruses. Viruses 15(9):1938.
            doi: 10.3390/v15091938. PMID: 37766345
            Substituted SuperScript IV Reverse Transcriptase for RT in Illumina kit for library prep and had improved depth of sequencing.
            Soufi EG, Jorio DL, Gerber Z et al (2024). Highly efficient and sensitive membrane-based concentration process allows quantification, surveillance, and sequencing of viruses in large volumes of wastewater. Water Res 249.
            doi: 10.1016/j.watres.2023.120959. PMID: 38070350
            Measure concatemeric cDNA formation upon phage infection. This ccDNA encodes for a toxic protein.
            Wilkinson ME, Li D, Gao A et al. (2024). Phage-triggeredreverse transcription assembles a toxic repetitive gene from a noncoding RNA. Science 386 (6717).
            doi:10.1126/science.adq3977 PMID: 39208082
            Synthesize cDNA from HIV RNA from patients. The amplified DNA was analyzed by gel electrophoresis and NGS. It was also cloned and sequenced by Sanger sequencing.
            Esmaeilzadeh E, Etemad B, Lavine CL et al. (2023). Autologous neutralizing antibodies increase with early antiretroviral therapy and shape HIV rebound after treatment interruption. Sci Transl Med 5(695).
            doi:10.1126/scitranslmed.abq4490
            Immunology
            UseReference
            Characterize monoclonal antibodies from B-cells. Developed a switching mechanism at the 5′ ends of the RNA transcript (SMART)-based method for amplifying IgV genes from single RM B cells to capture Ig heavy and light chain pairs.
            Weinfurter JT, Bennett SN, Reynolds MR (2024). A SMART method for isolating monoclonal antibodies from individual rhesus macaque memory B cells.J Immunol Methods 525:113602.
            doi: 10.1016/j.jim.2023.113602 PMID: 38103783
            Synthesize cDNA for RT-qPCR to study changes to gene expression as a result of CRISPR knock out of transcription factor Mef2d.
            Szeto ACH, Clark PA, Ferreira ACF et al. (2024). Mef2d potentiates type-2 immune responses and allergic lung inflammation. Science 384(6703).
            doi:10.1126/science.adl0370 PMID: 38935708
            Synthesize cDNA to measure the expression of Epstein bar virus transcripts.
            Zhang TT, Cheng RY, Ott AR, et al (2024). BCR signaling is required for posttransplant lymphoproliferative disease in immunodeficient mice receiving human B cells. Sci Transl Med 16(742).
            doi:10.1126/scitranslmed.adh8846 PMID: 38598616
            Synthesize cDNA from human B cells. cDNA was amplified and sequenced.Grigoryan L, Feng Y, Bellusci L et al. (2024). AS03 adjuvant enhances the magnitude, persistence, and clonal breadth of memory B cell responses to a plant-based COVID-19 vaccine in humans. Sci Immunol 9(94).
            doi:10.1126/sciimmunol.adi8039 PMID: 38579013
            cDNA synthesis for RT-qPCR. Patient samples were used.
            Morot J, Del Duca E, Chastagner M et al. (2023). Hyperactivation of the JAK2/STAT5 signaling pathway and evaluation of baricitinib treatment among patients with eosinophilic cellulitis. JAMA Dermatol 159(8):820–829.
            doi: 10.1001/jamadermatol.2023.1651 PMID: 37342057
            Synthesize cDNA from human B cells. cDNA was amplified and sequenced.
            Dejoux A, Zhu Q, Ganneau C, et al. (2024). Rocuronium-specific antibodies drive perioperative anaphylaxis but can also function as reversal agents in preclinical models. Sci Transl Med 16(764).
            doi:10.1126/scitranslmed.ado4463 PMID: 39259810
            Ecology and Evolutionary Biology
            UseReference
            Study mRNA expression profiles in tissue and identify a unique pathway in a shark species.
            Cutler CP, Omoregie E, Ojo T. UT-1 Transporter Expression in the Spiny Dogfish (Squalus acanthias): UT-1 Protein Shows a Different Localization in Comparison to That of Other Sharks. Biomolecules 14(9):1151.
            doi: 10.3390/biom14091151. PMID: 39334917
            Used SuperScript IV Reverse Transcriptase with low input sample difficult sample (RNA from insect antennae).
            Johny J, Nihad M, Alharbi HA et al. (2024). Silencing sensory neuron membrane protein RferSNMPu1 impairs pheromone detection in the invasive Asian Palm Weevil.Sci Rep 14(1):16541.
            doi: 10.1038/s41598-024-67309-x. PMID: 39019908
            Synthesize cDNA from RNA isolated from yeast cells to measure gene expression.
            Montrose K, Lac DT, Burnetti AJ et al.(2024). Proteostatic tuning underpins the evolution of novel multicellular traits. Sci Adv 10(10).
            doi:10.1126/sciadv.adn2706 PMID: 38457507
            Other areas
            UseReference
            Convert RNA to cDNA prior to a qPCR reaction to measure the relative expression levels of target genes in wound healing assays.
            Rapp J, Ness J, Wolf J (2024). 2D and 3D in vitro angiogenesis assays highlight different aspects of angiogenesis. Biochim Biophys Acta Mol Basis Dis 1870(3).
            doi: 10.1016/j.bbadis PMID: 38244944
            Synthesize cDNA for the study of RNA splicing. Products were analyzed by agarose gel electrophoresis, qPCR, and Sanger sequencing.
            Žedaveinytė R, Meers C, Le HC et al. (2024). Antagonistic conflict between transposon-encoded introns and guide RNAs. Science 385(6705).
            doi:10.1126/science.adm8189 PMID: 38991068
            Synthesize cDNA from CRISPR modified cells for RNA-seq.
            Dudnyk K, Cai D, Shi C, Xu J et al. (2024). Sequence basis of transcription initiation in the human genome. Science 384(6694).
            doi:10.1126/science.adj0116 PMID: 38662817
            Cancer
            UseReference
            Detect cancer biomarkers from extracellular vesicles by ddPCR (digital droplet).
            Lee KY, Beatson EL, Knechel MA et al. (2024). Detection of Extracellular Vesicle-Derived RNA as Potential Prostate Cancer Biomarkers: Role of Cancer-type SLCO1B3 and ABCC3. J Cancer 15(3):615–622.
            doi: 10.7150/jca.90836. PMID: 38213719.
            cDNA synthesis for preparation of a Poly(A)-seq library and also for nested RT-PCR to study changes in UTR length.
            Gabel AM, Belleville AE, Thomas JD et al. (2024). Multiplexed screening reveals how cancer-specific alternative polyadenylation shapes tumor growth in vivo.Nat Commun 15:959.
            doi: 10.1038/s41467-024-44931-x. PMID: 38302465
            Synthesize cDNA from FFPE samples for ddPCR.
            Look T, Puca E, Bühler M et al. (2023). Targeted delivery of tumor necrosis factor in combination with CCNU induces a T cell-dependent regression of glioblastoma. Sci Transl Med 15(697).
            doi:10.1126/scitranslmed.adf2281 PMID: 37224228
            Viral and infectious diseases
            UseReference
            Reverse transcribe viral RNA to cDNA prior to circularization, RCA, and nanopore sequencing.
            Misu M, Yoshikawa T, Sugimoto S (2023). Rapid whole genome sequencing methods for RNA viruses. Front Microbiol 23: 14.
            doi: 10.3389/fmicb.2023.1137086. PMID: 36910229
            Reverse transcribe avian viral RNA prior to amplification and library prep.
            Narvaez SA, Harrell TL, Oluwayinka (2023). Optimizing the Conditions for Whole-Genome Sequencing of Avian Reoviruses. Viruses 15(9):1938.
            doi: 10.3390/v15091938. PMID: 37766345
            Substituted SuperScript IV Reverse Transcriptase for RT in Illumina kit for library prep and had improved depth of sequencing.
            Soufi EG, Jorio DL, Gerber Z et al (2024). Highly efficient and sensitive membrane-based concentration process allows quantification, surveillance, and sequencing of viruses in large volumes of wastewater. Water Res 249.
            doi: 10.1016/j.watres.2023.120959. PMID: 38070350
            Measure concatemeric cDNA formation upon phage infection. This ccDNA encodes for a toxic protein.
            Wilkinson ME, Li D, Gao A et al. (2024). Phage-triggeredreverse transcription assembles a toxic repetitive gene from a noncoding RNA. Science 386 (6717).
            doi:10.1126/science.adq3977 PMID: 39208082
            Synthesize cDNA from HIV RNA from patients. The amplified DNA was analyzed by gel electrophoresis and NGS. It was also cloned and sequenced by Sanger sequencing.
            Esmaeilzadeh E, Etemad B, Lavine CL et al. (2023). Autologous neutralizing antibodies increase with early antiretroviral therapy and shape HIV rebound after treatment interruption. Sci Transl Med 5(695).
            doi:10.1126/scitranslmed.abq4490
            Immunology
            UseReference
            Characterize monoclonal antibodies from B-cells. Developed a switching mechanism at the 5′ ends of the RNA transcript (SMART)-based method for amplifying IgV genes from single RM B cells to capture Ig heavy and light chain pairs.
            Weinfurter JT, Bennett SN, Reynolds MR (2024). A SMART method for isolating monoclonal antibodies from individual rhesus macaque memory B cells.J Immunol Methods 525:113602.
            doi: 10.1016/j.jim.2023.113602 PMID: 38103783
            Synthesize cDNA for RT-qPCR to study changes to gene expression as a result of CRISPR knock out of transcription factor Mef2d.
            Szeto ACH, Clark PA, Ferreira ACF et al. (2024). Mef2d potentiates type-2 immune responses and allergic lung inflammation. Science 384(6703).
            doi:10.1126/science.adl0370 PMID: 38935708
            Synthesize cDNA to measure the expression of Epstein bar virus transcripts.
            Zhang TT, Cheng RY, Ott AR, et al (2024). BCR signaling is required for posttransplant lymphoproliferative disease in immunodeficient mice receiving human B cells. Sci Transl Med 16(742).
            doi:10.1126/scitranslmed.adh8846 PMID: 38598616
            Synthesize cDNA from human B cells. cDNA was amplified and sequenced.Grigoryan L, Feng Y, Bellusci L et al. (2024). AS03 adjuvant enhances the magnitude, persistence, and clonal breadth of memory B cell responses to a plant-based COVID-19 vaccine in humans. Sci Immunol 9(94).
            doi:10.1126/sciimmunol.adi8039 PMID: 38579013
            cDNA synthesis for RT-qPCR. Patient samples were used.
            Morot J, Del Duca E, Chastagner M et al. (2023). Hyperactivation of the JAK2/STAT5 signaling pathway and evaluation of baricitinib treatment among patients with eosinophilic cellulitis. JAMA Dermatol 159(8):820–829.
            doi: 10.1001/jamadermatol.2023.1651 PMID: 37342057
            Synthesize cDNA from human B cells. cDNA was amplified and sequenced.
            Dejoux A, Zhu Q, Ganneau C, et al. (2024). Rocuronium-specific antibodies drive perioperative anaphylaxis but can also function as reversal agents in preclinical models. Sci Transl Med 16(764).
            doi:10.1126/scitranslmed.ado4463 PMID: 39259810
            Ecology and Evolutionary Biology
            UseReference
            Study mRNA expression profiles in tissue and identify a unique pathway in a shark species.
            Cutler CP, Omoregie E, Ojo T. UT-1 Transporter Expression in the Spiny Dogfish (Squalus acanthias): UT-1 Protein Shows a Different Localization in Comparison to That of Other Sharks. Biomolecules 14(9):1151.
            doi: 10.3390/biom14091151. PMID: 39334917
            Used SuperScript IV Reverse Transcriptase with low input sample difficult sample (RNA from insect antennae).
            Johny J, Nihad M, Alharbi HA et al. (2024). Silencing sensory neuron membrane protein RferSNMPu1 impairs pheromone detection in the invasive Asian Palm Weevil.Sci Rep 14(1):16541.
            doi: 10.1038/s41598-024-67309-x. PMID: 39019908
            Synthesize cDNA from RNA isolated from yeast cells to measure gene expression.
            Montrose K, Lac DT, Burnetti AJ et al.(2024). Proteostatic tuning underpins the evolution of novel multicellular traits. Sci Adv 10(10).
            doi:10.1126/sciadv.adn2706 PMID: 38457507
            Other areas
            UseReference
            Convert RNA to cDNA prior to a qPCR reaction to measure the relative expression levels of target genes in wound healing assays.
            Rapp J, Ness J, Wolf J (2024). 2D and 3D in vitro angiogenesis assays highlight different aspects of angiogenesis. Biochim Biophys Acta Mol Basis Dis 1870(3).
            doi: 10.1016/j.bbadis PMID: 38244944
            Synthesize cDNA for the study of RNA splicing. Products were analyzed by agarose gel electrophoresis, qPCR, and Sanger sequencing.
            Žedaveinytė R, Meers C, Le HC et al. (2024). Antagonistic conflict between transposon-encoded introns and guide RNAs. Science 385(6705).
            doi:10.1126/science.adm8189 PMID: 38991068
            Synthesize cDNA from CRISPR modified cells for RNA-seq.
            Dudnyk K, Cai D, Shi C, Xu J et al. (2024). Sequence basis of transcription initiation in the human genome. Science 384(6694).
            doi:10.1126/science.adj0116 PMID: 38662817
            Reverse transcriptase selection tool
            Find the right products for your cDNA synthesis needs.
            Reverse transcriptase webinars
            Stay up-to-date on current industry trends in cDNA synthesis applications with on-demand webinars.
            Reverse transcriptase videos
            Watch instructional videos on troubleshooting tips, how-to videos, and explainers related to cDNA synthesis.
            Reverse transcription education
            cDNA Synthesis Protocol in Five Steps
            Review a wealth of educational resources on cDNA synthesis.
            Reverse transcriptase literature
            Access white papers and technical and application notes for insights on cutting edge reverse transcription technology.
            OEM and custom solutions
            Contact us
            PCR and cDNA Synthesis Support Center
            Get support for your cDNA synthesis experiments.
            For Research Use Only. Not for use in diagnostic procedures.
            Stylesheet for Classic Wide Template adjustments