Search
Pulp and paper applications typically require total reduced sulfur (TRS) systems and the Thermo ScientificTM Continuous Emissions Monitoring System for Total Reduced Sulfur has been providing a unique solution for challenges facing the pulp and paper industry for over 20 years. Our systems have the unique ability to measure low TRS levels even in high sulfur dioxide (SO2) conditions and currently there are more than 300 TRS CEMS installed globally.
Sales and support for the Thermo Scientific C-series Gas Analyzers will be discontinued beginning August 1, 2015.
Unheated sample lines are more reliable and less expensive to run. The Thermo Scientific Continuous Emissions Monitoring System for Total Reduced Sulfur employs a dry basis dilution system which means no heated sample line is necessary, ever. Many competitive TRS systems use a wet dilution or a dry extractive technique which requires a heated umbilical line. Heated lines are often more costly and results in more downtime.
The Thermo Scientific Continuous Emissions Monitoring System for Total Reduced Sulfur uses the same working principle of system is based on EPA Reference Method 16C. As detailed on the US epa.gov website, Method 16C is a procedure for measuring total reduced sulfur (TRS) in stationary source emissions using a continuous instrumental analyzer. Quality assurance and quality control requirements are included to assure that data of known quality is collected. Documenting adherence to these specific requirements for equipment, supplies, sample collection and analysis, calculations, and data analysis is easier with the Thermo Scientific TRS CEMS.
The Thermo Scientific Continuous Emissions Monitoring System for Total Reduced Sulfur delivers the cleanest sample to the analyzer as possible because the system measures oxygen with a dilution probe. With other techniques, the sample is concentrated with pollutants and results in sludge and heavy contaminants being fed to the analyzer. Measuring oxygen with a dilution probe ensures you are drawing a cleaner sample ensures longer lifetime and more time between failures. Additionally, the dilution system ensures emissions compliance regulations.
The sulfur dioxide (SO2) scrubbing technique that is used by the Thermo Scientific Continuous Emissions Monitoring System for Total Reduced Sulfur is designed specifically for lime kilns and recovery boilers that burn black liquor fuels. Burning black liquor fuels emits mercaptans and sulfur compounds. The Thermo Scientific TRS CEMS uses an SO2 scrubbing technique, which is favorable over the differential subtraction technique. The SO2 scrubbing technique allows for the measurement of low TRS levels (up to 10,000 ppm of SO2) even in high SO2 conditions unlike the differential subtraction technique employed by other systems used when the ratio of TRS to SO2 is near unity.
Eliminate bias and error by calibrating with a gas that best represents the gas being measured. For example, it is best to calibrate with hydrogen sulfide (H2S) because from a chemical reaction standpoint, it is more efficient to convert H2S to sulfur dioxide (SO2) because the ratio between the molecules is 1:1.
Our technical and engineering staff share their product and application expertise for gas analyzers and systems.
Sign up today to receive Fast5 tips and tricks every month!
Schedule your analyzer, calibrator, probe controller or probe for a Return Material Authorization (RMA) to factory depot repair for complete maintenance and factory specification service.