Search
The ability to stain and detect intracellular molecules opens the door to identify distinct cell subsets as well as further characterize cell populations. Our resources, tools and products help you save time and effort and increase your efficiency when designing your flow cytometry experiments.
Save time and energy by using our streamlined approach to find the antibody that you need quickly and easily.
When performing intracellular staining for flow cytometry the selection of buffers used for fixation and permeabilization has a significant impact on the quality and accuracy of data. We provide optimized solutions for staining cytosolic, nuclear and secreted proteins for flow cytometry.
| eBioscience Intracellular Fixation & Permeabilization Buffer Set | FIX & PERM Cell Fixation & Cell Permeabilization Kit | |
|---|---|---|
| Use |
Learn more about the IC Fix & Perm buffer set |
|
| Contents |
|
|
| Size | 1 kit | 50 tests |
| Cat. No. | 88-8824-00 | GAS003 |
| Regulatory status | RUO–For Research Use Only. Not for Use in Diagnostic Procedures. | GPR–General Purpose Reagent. |
| When to use | For use in research only (i.e. research on any sample type). | For use in general laboratory application, that is used to collect, prepare, and examine specimens from the human body for diagnostic purposes. |
| eBioscience Foxp3 / Transcription Factor Staining Buffer Set | |
|---|---|
| Use |
|
| Contents |
|
| Size | 1 kit |
| Cat. No. | 00-5523-00 |
Save time and avoid inaccurate analysis of your cell population by removing dead cells and unwanted artifacts efficiently with the Invitrogen LIVE/DEAD Fixable Dead Cell Stains.
Find LIVE/DEAD kits for:UV & 405 nm lasers | 488 & 561 nm lasers | 633/635 laser
| LIVE/DEAD Fixable Blue stain | LIVE/DEAD Fixable Violet stain | LIVE/DEAD Fixable Aqua stain | LIVE/DEAD Fixable Yellow stain | |
|---|---|---|---|---|
| Basis of assay | LIVE/DEAD Fixable Dead Cell Stain kits are fixable viability dyes that distinguish live cells from dead cells based on cell membrane integrity and access to available amines. Cells can then be fixed for intracellular antigen detection without loss of original cell staining pattern. Allows easy exclusion of dead cells from flow cytometry readout. | |||
| Readout | Dead cells are highly fluorescent and easy to distinguish from live cells, which are stained with minimal fluorescence | |||
| Laser (nm) | UV | 405 | 405 | 405 |
| Ex/Em (nm) | 350/450 | 416/451 | 367/526 | 400/575 |
| Bibliography |
Citations
| |||
| Multiplexable |
Yes
|
Yes
|
Yes
|
Yes
|
| Dye compatibility | Cannot be used in combination with Invitrogen product line such as Indo-1 (blue), DAPI, or Hoechst 33342 | Cannot be used in combination with Invitrogen product line such as Pacific Blue dye, CellTrace Violet stain, FxCycle Violet stain, BV421 or eFluor 450 | Cannot be used in combination with Invitrogen product line such as Pacific Green dye, F2N12S apoptosis assay, AmCyan, or BV510 | Cannot be used in combination with Invitrogen product line such as Pacific Orange dye, Qdot 605, F2N12S apoptosis assay, or BV605 |
| Fixable |
Yes
|
Yes
|
Yes
|
Yes
|
| Sample type |
Live cells
| |||
| Size | 80 tests | |||
| Cat. No. | ||||
| LIVE/DEAD Fixable Green stain | LIVE/DEAD Fixable Red stain | ||
|---|---|---|---|
| Basis of assay | LIVE/DEAD Fixable Dead Cell Stain kits are fixable viability dyes that distinguish live cells from dead cells based on cell membrane integrity and access to available amines. Cells can then be fixed for intracellular antigen detection without loss of original cell staining pattern. Allows easy exclusion of dead cells from flow cytometry readout. | ||
| Readout | Dead cells are highly fluorescent and easy to distinguish from live cells, which are stained with minimal fluorescence | ||
| Laser (nm) | 488 | 488 | 561 |
| Ex/Em (nm) | 495/520 | 595/615 | |
| Bibliography |
Citations
| ||
| Multiplexable |
Yes
|
Yes
| |
| Dye compatibility | Cannot be used in combination with Invitrogen product line such as FITC, Alexa Fluor 488, GFP, or CellTrace CSFE stain | Cannot be used in combination with Invitrogen product line such as R-PE, propidium iodide, SYTOX AADvanced stain, 7-AAD, PE-Alexa Fluor 610 and PE-Texas Red tandems | |
| Fixable |
Yes
|
Yes
| |
| Sample type |
Live cells
| ||
| Size | 80 tests | ||
| Cat. No. | |||
| LIVE/DEAD Fixable Far Red stain | LIVE/DEAD Fixable Near-IR stain | |
|---|---|---|
| Basis of assay | LIVE/DEAD Fixable Dead Cell Stain kits are fixable viability dyes that distinguish live cells from dead cells based on cell membrane integrity and access to available amines. Cells can then be fixed for intracellular antigen detection without loss of original cell staining pattern. Allows easy exclusion of dead cells from flow cytometry readout. | |
| Readout | Dead cells are highly fluorescent and easy to distinguish from live cells, which are stained with minimal fluorescence | |
| Laser (nm) | 633/635 | 633/635 |
| Ex/Em (nm) | 650/665 | 750/775 |
| Bibliography |
Citations
| |
| Multiplexable |
Yes
|
Yes
|
| Dye compatibility | Cannot be used in combination with Invitrogen product line such as APC, Alexa Fluor 647 dye, FxCycle Far Red and CellTrace Far Red stains | Cannot be used in combination with Invitrogen product line such as APC-Alexa Fluor 750, APC-Cy7 or Vybrant DyeCycle Ruby stain |
| Fixable |
Yes
|
Yes
|
| Sample type |
Live cells
| |
| Size | 80 tests | |
| Cat. No. |
L34973
|
L34975
|
Staining intracellular targets for flow cytometry requires the use of multiple antibodies that often have overlapping fluorescence emission spectra. The emission spillover must be compensated for correctly in order to ensure that the fluorescent signal being measure is due to the fluorochrome of interest and thus the cell population of interest.
Learn more about eBeads microspheres for antibody compensation
| UltraComp eBeads | |
|---|---|
| Use | Designed for compensation of dyes in immunophenotyping experiments using fluorescently labeled antibodies, the beads can be stained with individual fluorochrome-conjugated antibodies for use as single-color compensation controls. |
| Reactivity | Hamster, mouse, and rat antibodies and recognition of the kappa & lambda chains |
| Format | One vial, dispense as a single drop |
| Compatible with most standard lasers, UV to 633 nm | |
| Size | 25 tests |
| Cat. No. | 01-2222-41 |
Save time and energy by using our streamlined approach to find the antibody that you need quickly and easily.
When performing intracellular staining for flow cytometry the selection of buffers used for fixation and permeabilization has a significant impact on the quality and accuracy of data. We provide optimized solutions for staining cytosolic, nuclear and secreted proteins for flow cytometry.
| eBioscience Intracellular Fixation & Permeabilization Buffer Set | FIX & PERM Cell Fixation & Cell Permeabilization Kit | |
|---|---|---|
| Use |
Learn more about the IC Fix & Perm buffer set |
|
| Contents |
|
|
| Size | 1 kit | 50 tests |
| Cat. No. | 88-8824-00 | GAS003 |
| Regulatory status | RUO–For Research Use Only. Not for Use in Diagnostic Procedures. | GPR–General Purpose Reagent. |
| When to use | For use in research only (i.e. research on any sample type). | For use in general laboratory application, that is used to collect, prepare, and examine specimens from the human body for diagnostic purposes. |
| eBioscience Foxp3 / Transcription Factor Staining Buffer Set | |
|---|---|
| Use |
|
| Contents |
|
| Size | 1 kit |
| Cat. No. | 00-5523-00 |
Save time and avoid inaccurate analysis of your cell population by removing dead cells and unwanted artifacts efficiently with the Invitrogen LIVE/DEAD Fixable Dead Cell Stains.
Find LIVE/DEAD kits for:UV & 405 nm lasers | 488 & 561 nm lasers | 633/635 laser
| LIVE/DEAD Fixable Blue stain | LIVE/DEAD Fixable Violet stain | LIVE/DEAD Fixable Aqua stain | LIVE/DEAD Fixable Yellow stain | |
|---|---|---|---|---|
| Basis of assay | LIVE/DEAD Fixable Dead Cell Stain kits are fixable viability dyes that distinguish live cells from dead cells based on cell membrane integrity and access to available amines. Cells can then be fixed for intracellular antigen detection without loss of original cell staining pattern. Allows easy exclusion of dead cells from flow cytometry readout. | |||
| Readout | Dead cells are highly fluorescent and easy to distinguish from live cells, which are stained with minimal fluorescence | |||
| Laser (nm) | UV | 405 | 405 | 405 |
| Ex/Em (nm) | 350/450 | 416/451 | 367/526 | 400/575 |
| Bibliography |
Citations
| |||
| Multiplexable |
Yes
|
Yes
|
Yes
|
Yes
|
| Dye compatibility | Cannot be used in combination with Invitrogen product line such as Indo-1 (blue), DAPI, or Hoechst 33342 | Cannot be used in combination with Invitrogen product line such as Pacific Blue dye, CellTrace Violet stain, FxCycle Violet stain, BV421 or eFluor 450 | Cannot be used in combination with Invitrogen product line such as Pacific Green dye, F2N12S apoptosis assay, AmCyan, or BV510 | Cannot be used in combination with Invitrogen product line such as Pacific Orange dye, Qdot 605, F2N12S apoptosis assay, or BV605 |
| Fixable |
Yes
|
Yes
|
Yes
|
Yes
|
| Sample type |
Live cells
| |||
| Size | 80 tests | |||
| Cat. No. | ||||
| LIVE/DEAD Fixable Green stain | LIVE/DEAD Fixable Red stain | ||
|---|---|---|---|
| Basis of assay | LIVE/DEAD Fixable Dead Cell Stain kits are fixable viability dyes that distinguish live cells from dead cells based on cell membrane integrity and access to available amines. Cells can then be fixed for intracellular antigen detection without loss of original cell staining pattern. Allows easy exclusion of dead cells from flow cytometry readout. | ||
| Readout | Dead cells are highly fluorescent and easy to distinguish from live cells, which are stained with minimal fluorescence | ||
| Laser (nm) | 488 | 488 | 561 |
| Ex/Em (nm) | 495/520 | 595/615 | |
| Bibliography |
Citations
| ||
| Multiplexable |
Yes
|
Yes
| |
| Dye compatibility | Cannot be used in combination with Invitrogen product line such as FITC, Alexa Fluor 488, GFP, or CellTrace CSFE stain | Cannot be used in combination with Invitrogen product line such as R-PE, propidium iodide, SYTOX AADvanced stain, 7-AAD, PE-Alexa Fluor 610 and PE-Texas Red tandems | |
| Fixable |
Yes
|
Yes
| |
| Sample type |
Live cells
| ||
| Size | 80 tests | ||
| Cat. No. | |||
| LIVE/DEAD Fixable Far Red stain | LIVE/DEAD Fixable Near-IR stain | |
|---|---|---|
| Basis of assay | LIVE/DEAD Fixable Dead Cell Stain kits are fixable viability dyes that distinguish live cells from dead cells based on cell membrane integrity and access to available amines. Cells can then be fixed for intracellular antigen detection without loss of original cell staining pattern. Allows easy exclusion of dead cells from flow cytometry readout. | |
| Readout | Dead cells are highly fluorescent and easy to distinguish from live cells, which are stained with minimal fluorescence | |
| Laser (nm) | 633/635 | 633/635 |
| Ex/Em (nm) | 650/665 | 750/775 |
| Bibliography |
Citations
| |
| Multiplexable |
Yes
|
Yes
|
| Dye compatibility | Cannot be used in combination with Invitrogen product line such as APC, Alexa Fluor 647 dye, FxCycle Far Red and CellTrace Far Red stains | Cannot be used in combination with Invitrogen product line such as APC-Alexa Fluor 750, APC-Cy7 or Vybrant DyeCycle Ruby stain |
| Fixable |
Yes
|
Yes
|
| Sample type |
Live cells
| |
| Size | 80 tests | |
| Cat. No. |
L34973
|
L34975
|
Staining intracellular targets for flow cytometry requires the use of multiple antibodies that often have overlapping fluorescence emission spectra. The emission spillover must be compensated for correctly in order to ensure that the fluorescent signal being measure is due to the fluorochrome of interest and thus the cell population of interest.
Learn more about eBeads microspheres for antibody compensation
| UltraComp eBeads | |
|---|---|
| Use | Designed for compensation of dyes in immunophenotyping experiments using fluorescently labeled antibodies, the beads can be stained with individual fluorochrome-conjugated antibodies for use as single-color compensation controls. |
| Reactivity | Hamster, mouse, and rat antibodies and recognition of the kappa & lambda chains |
| Format | One vial, dispense as a single drop |
| Compatible with most standard lasers, UV to 633 nm | |
| Size | 25 tests |
| Cat. No. | 01-2222-41 |
.
We have a team of technical support scientists available to help you. Our team provides this panel design service for those who want help with building their panel every step of the way.
Contact our technical sales specialists for help choosing antibodies, fluorophores, and reagents for success on the flow cytometer you’ll be using.
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.