Search
TaqMan miRNA Assays are innovative tools for miRNA research—from isolation through discovery, profiling, quantitation, validation, and functional analysis. The newer TaqMan Advanced miRNA Assays use ligation-based universal reverse transcription for a streamlined and highly sensitive workflow. The standard TaqMan miRNA Assays, which employ target-specific stem-loop reverse transcription primers for 3’ extended templates, continue to cover a range of species using standard TaqMan Assay–based real-time PCR.
| TaqMan MicroRNA Assays | TaqMan Advanced miRNA Assays | |
| Description | Employ a novel target-specific stem-loop primer during cDNA synthesis that produces a template for real-time PCR | Employ a universal RT step for a streamlined workflow and a universal miR-Amp step |
| RT chemistry | miRNA-specific RT | Universal RT |
| Throughput | Best for 1–10 targets | Best for >10 targets |
| Coverage | 205 species available; coverage for miRBase v.21 | All human, mouse, and rat miRNAs; coverage for miRBase v.21 |
| Formats | Available in individual tubes, TaqMan array cards and plates, and OpenArray formats | Available in individual tubes, TaqMan array cards and plates, and OpenArray formats; inquire for custom plating options |
TaqMan Advanced miRNA Assays utilize a universal primer to reverse transcribe all of the miRNAs in the sample. They enable highly sensitive and specific quantification of mature miRNAs using qPCR. Together with the TaqMan Advanced miRNA cDNA Synthesis Kit, this solution is designed to provide a streamlined workflow that allows for exceptional detection of multiple miRNA targets from a single sample.
mirVana miRNA Isolation kits use a rapid procedure to isolate total RNA, including small RNAs such as miRNA, small interfering RNA (siRNA), and small nuclear RNA (snRNA), from tissues and cells.
TaqMan Advanced miRNA cDNA Synthesis Kit
The TaqMan Advanced miRNA cDNA Synthesis Kit is used for preparation of cDNA templates from the entire pool of mature miRNAs in a sample.
miR-Amp Universal Amplification
miR-Amp uses universal primers and master mix to uniformly increase the amount of cDNA for each target. miR-Amp reagents are included in the TaqMan Advanced miRNA cDNA Synthesis Kit.
Applied Biosystems real-time PCR instruments
With Applied Biosystems real-time PCR platforms, you get true value with excellent performance, reliability, and world-class support. Designed for compatibility with Applied Biosystems TaqMan Assays and the flexibility of unlimited real-time chemistry choices, we make it easy for you to get started with your experiments.
mirVana miRNA Isolation kits use a rapid procedure to isolate total RNA, including small RNAs such as miRNA, small interfering RNA (siRNA), and small nuclear RNA (snRNA), from tissues and cells.
TaqMan Advanced miRNA cDNA Synthesis Kit
The TaqMan Advanced miRNA cDNA Synthesis Kit is used for preparation of cDNA templates from the entire pool of mature miRNAs in a sample.
miR-Amp Universal Amplification
miR-Amp uses universal primers and master mix to uniformly increase the amount of cDNA for each target. miR-Amp reagents are included in the TaqMan Advanced miRNA cDNA Synthesis Kit.
Applied Biosystems real-time PCR instruments
With Applied Biosystems real-time PCR platforms, you get true value with excellent performance, reliability, and world-class support. Designed for compatibility with Applied Biosystems TaqMan Assays and the flexibility of unlimited real-time chemistry choices, we make it easy for you to get started with your experiments.
TaqMan miRNA Assays employ an innovative target-specific stem-loop reverse transcription primer to address the challenge of the short length of mature miRNA. The primer extends the 3' end of the target to produce a template that can be used in standard TaqMan Assay-based real-time PCR. Also, the stem-loop structure in the tail of the primer confers a key advantage to these assays: specific detection of the mature, biologically active miRNA.
We offer a variety of kits for the isolation and analysis of miRNA and other small RNAs, each is ideal for use in miRNA analysis because they enable quantitative recovery of small RNA (<200 nt) and maintenance of representative amounts of small RNA (eliminating experimental bias).
| TaqMan miRNA ABC Purification Kit Purify high-quality miRNA from difficult sample types without hazardous chemicals. This kit enables single-tube isolation of specific miRNA from small inputs of all human sample types, including blood, serum, plasma, FFPE samples, solid tissues, cultured cells, and saliva in 75 minutes. | ||
mirVana miRNA Isolation Kit | ||
RecoverAll Total Nucleic Acid Isolation Kit for FFPE | ||
TaqMan MicroRNA Cells-to-CT Kit |
Two-step TaqMan MicroRNA Assays, containing the TaqMan Assay and reverse transcription primer specific for the target of interest, are designed for individual miRNA targets. The TaqMan MicroRNA Reverese Transcription Kit provides optimal TaqMan MicroRNA Assay performance. This kit is used with a single RT primer provided with each individual TaqMan MicroRNA Assays or with Megaplex RT Primers to convert miRNA to cDNA prior to real-time PCR quantitation.
TaqMan MicroRNA Reverse Transcription Kit | ||
TaqMan MicroRNA Assays (RT tube) |
For the final quantitation step, the cDNA product generated in step 2 is mixed with TaqMan Universal Master Mix II and added to the matched TaqMan MicroRNA Assay.
TaqMan MicroRNA Assays (individual assays) | ||
TaqMan Universal Master Mix II |
Applied Biosystems analysis apps are innovative, cloud-based secondary data analysis solutions that bring together multiple data sets in one convenient place. This online solution makes it easier to view, store, and analyze qPCR and Sanger sequencing data. These apps take advantage of cloud computing to provide highly versatile analysis tools that are flexible, fast, and easy to use.
Learn more ›
We offer a variety of kits for the isolation and analysis of miRNA and other small RNAs, each is ideal for use in miRNA analysis because they enable quantitative recovery of small RNA (<200 nt) and maintenance of representative amounts of small RNA (eliminating experimental bias).
| TaqMan miRNA ABC Purification Kit Purify high-quality miRNA from difficult sample types without hazardous chemicals. This kit enables single-tube isolation of specific miRNA from small inputs of all human sample types, including blood, serum, plasma, FFPE samples, solid tissues, cultured cells, and saliva in 75 minutes. | ||
mirVana miRNA Isolation Kit | ||
RecoverAll Total Nucleic Acid Isolation Kit for FFPE | ||
TaqMan MicroRNA Cells-to-CT Kit |
Two-step TaqMan MicroRNA Assays, containing the TaqMan Assay and reverse transcription primer specific for the target of interest, are designed for individual miRNA targets. The TaqMan MicroRNA Reverese Transcription Kit provides optimal TaqMan MicroRNA Assay performance. This kit is used with a single RT primer provided with each individual TaqMan MicroRNA Assays or with Megaplex RT Primers to convert miRNA to cDNA prior to real-time PCR quantitation.
TaqMan MicroRNA Reverse Transcription Kit | ||
TaqMan MicroRNA Assays (RT tube) |
For the final quantitation step, the cDNA product generated in step 2 is mixed with TaqMan Universal Master Mix II and added to the matched TaqMan MicroRNA Assay.
TaqMan MicroRNA Assays (individual assays) | ||
TaqMan Universal Master Mix II |
Applied Biosystems analysis apps are innovative, cloud-based secondary data analysis solutions that bring together multiple data sets in one convenient place. This online solution makes it easier to view, store, and analyze qPCR and Sanger sequencing data. These apps take advantage of cloud computing to provide highly versatile analysis tools that are flexible, fast, and easy to use.
Learn more ›
A technical guide to identifying miRNA normalizers using TaqMan Advanced miRNA Assays
TaqMan Advanced miRNA Assays—superior performance for miRNA detection and quantification
A complete workflow for high-throughput isolation of serum miRNAs and downstream analysis by qRT-PCR: application for cancer research and biomarker discovery
Simultaneous detection of miRNA and mRNA on TaqMan Array Cards using the TaqMan Advanced miRNA workflow
For Research Use Only. Not for use in diagnostic procedures.